Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10461366-10461929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD3
Entrez Gene ID
272
Gene Name
adenosine monophosphate deaminase 3
Gene Aliases
-
UniGene
Hs.501890
Gene Chromosome Location
Chr.11: 10450321-10507579 on Build GRCh38
Transcripts
NM_000480.2 NM_001025389.1 NM_001025390.1 NM_001172430.1 NM_001172431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297582 c.31C>T p.P11S Chr.11: 10461523-10461523 on Build GRCh38
1350527 c.225G>T p.Q75H Chr.11: 10461717-10461717 on Build GRCh38
4017410 c.123G>T p.E41D Chr.11: 10461615-10461615 on Build GRCh38
4802316 c.87G>T p.L29L Chr.11: 10461579-10461579 on Build GRCh38
4017411 c.139C>A p.L47M Chr.11: 10461631-10461631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776041673 Chr.11: 10461720-10461720 on Build GRCh38
rs767955616 Chr.11: 10461458-10461458 on Build GRCh38
rs749940953 Chr.11: 10461581-10461581 on Build GRCh38
rs910172760 Chr.11: 10461534-10461534 on Build GRCh38
rs540683532 Chr.11: 10461509-10461509 on Build GRCh38
rs145604316 Chr.11: 10461524-10461524 on Build GRCh38
rs935689688 Chr.11: 10461639-10461639 on Build GRCh38
rs776533422 Chr.11: 10461408-10461408 on Build GRCh38
rs745366158 Chr.11: 10461481-10461481 on Build GRCh38
rs773226493 Chr.11: 10461515-10461515 on Build GRCh38
rs373716662 Chr.11: 10461608-10461608 on Build GRCh38
rs746879134 Chr.11: 10461489-10461489 on Build GRCh38
rs763501693 Chr.11: 10461407-10461407 on Build GRCh38
rs561207052 Chr.11: 10461525-10461525 on Build GRCh38
rs778629569 Chr.11: 10461462-10461462 on Build GRCh38
rs1045502991 Chr.11: 10461705-10461705 on Build GRCh38
rs760184620 Chr.11: 10461669-10461669 on Build GRCh38
rs545122288 Chr.11: 10461447-10461447 on Build GRCh38
rs866323898 Chr.11: 10461580-10461580 on Build GRCh38
rs781089888 Chr.11: 10461779-10461779 on Build GRCh38
rs529947316 Chr.11: 10461564-10461564 on Build GRCh38
rs774226348 Chr.11: 10461667-10461667 on Build GRCh38
rs75353752 Chr.11: 10461765-10461765 on Build GRCh38
rs571424441 Chr.11: 10461764-10461764 on Build GRCh38
rs370410880 Chr.11: 10461791-10461791 on Build GRCh38
rs778505193 Chr.11: 10461412-10461412 on Build GRCh38
rs961581096 Chr.11: 10461800-10461800 on Build GRCh38
rs372156334 Chr.11: 10461752-10461752 on Build GRCh38
rs186250080 Chr.11: 10461429-10461429 on Build GRCh38
rs369669044 Chr.11: 10461570-10461570 on Build GRCh38
rs966976087 Chr.11: 10461617-10461617 on Build GRCh38
rs148904594 Chr.11: 10461526-10461526 on Build GRCh38
rs527260375 Chr.11: 10461503-10461503 on Build GRCh38
rs899212838 Chr.11: 10461860-10461860 on Build GRCh38
rs955758494 Chr.11: 10461852-10461852 on Build GRCh38
rs779381182 Chr.11: 10461595-10461595 on Build GRCh38
rs889445553 Chr.11: 10461636-10461636 on Build GRCh38
rs190670467 Chr.11: 10461802-10461802 on Build GRCh38
rs933555146 Chr.11: 10461735-10461735 on Build GRCh38
rs143425698 Chr.11: 10461565-10461565 on Build GRCh38
rs199953746 Chr.11: 10461732-10461732 on Build GRCh38
rs757169910 Chr.11: 10461628-10461628 on Build GRCh38
rs376360074 Chr.11: 10461505-10461505 on Build GRCh38
rs200620189 Chr.11: 10461728-10461728 on Build GRCh38
rs761179959 Chr.11: 10461486-10461486 on Build GRCh38
rs745794355 Chr.11: 10461414-10461414 on Build GRCh38
rs368833888 Chr.11: 10461780-10461780 on Build GRCh38
rs922765795 Chr.11: 10461845-10461845 on Build GRCh38
rs768905591 Chr.11: 10461427-10461427 on Build GRCh38
rs1037888890 Chr.11: 10461796-10461796 on Build GRCh38
rs12418097 Chr.11: 10461739-10461739 on Build GRCh38
rs201661559 Chr.11: 10461437-10461437 on Build GRCh38
rs777385535 Chr.11: 10461787-10461787 on Build GRCh38
rs751921390 Chr.11: 10461446-10461446 on Build GRCh38
rs762457397 Chr.11: 10461733-10461733 on Build GRCh38
rs372846179 Chr.11: 10461424-10461424 on Build GRCh38
rs138403236 Chr.11: 10461664-10461664 on Build GRCh38
rs576749734 Chr.11: 10461445-10461445 on Build GRCh38
rs959396325 Chr.11: 10461749-10461749 on Build GRCh38
rs141052158 Chr.11: 10461726-10461726 on Build GRCh38
rs746982689 Chr.11: 10461607-10461607 on Build GRCh38
rs144704411 Chr.11: 10461616-10461616 on Build GRCh38
rs78155698 Chr.11: 10461461-10461461 on Build GRCh38
rs141926261 Chr.11: 10461666-10461666 on Build GRCh38
rs770055051 Chr.11: 10461646-10461646 on Build GRCh38
rs867446911 Chr.11: 10461523-10461523 on Build GRCh38
rs775206482 Chr.11: 10461416-10461416 on Build GRCh38
rs372097330 Chr.11: 10461487-10461487 on Build GRCh38
rs773456324 Chr.11: 10461652-10461652 on Build GRCh38
rs911002202 Chr.11: 10461870-10461870 on Build GRCh38
rs763070134 Chr.11: 10461519-10461519 on Build GRCh38
rs149625321 Chr.11: 10461428-10461428 on Build GRCh38
rs150672366 Chr.11: 10461696-10461696 on Build GRCh38
rs935662279 Chr.11: 10461890-10461890 on Build GRCh38
rs137905602 Chr.11: 10461572-10461572 on Build GRCh38
rs147321717 Chr.11: 10461434-10461434 on Build GRCh38
rs765920579 Chr.11: 10461747-10461747 on Build GRCh38
rs985209078 Chr.11: 10461854-10461854 on Build GRCh38
rs1001914767 Chr.11: 10461552-10461552 on Build GRCh38
rs767996085 Chr.11: 10461561-10461561 on Build GRCh38
rs747924930 Chr.11: 10461635-10461635 on Build GRCh38
rs375814372 Chr.11: 10461629-10461629 on Build GRCh38
rs753551502 Chr.11: 10461460-10461460 on Build GRCh38
rs940161890 Chr.11: 10461550-10461550 on Build GRCh38
rs933731212 Chr.11: 10461600-10461600 on Build GRCh38
rs142453229 Chr.11: 10461582-10461582 on Build GRCh38
rs138271082 Chr.11: 10461406-10461406 on Build GRCh38

View Full Product Details