Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 71348881-71349454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF638
Entrez Gene ID
27332
Gene Name
zinc finger protein 638
Gene Aliases
NP220, ZFML, Zfp638
UniGene
Hs.434401
Gene Chromosome Location
Chr.2: 71276543-71435061 on Build GRCh38
Transcripts
NM_001014972.2 NM_001252612.1 NM_001252613.1 NM_014497.4 XM_005264263.1 XM_006711989.1 XM_011532768.2 XM_017003809.1 XM_017003810.1 XM_017003811.1 XM_017003812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4415808 c.328A>G p.I110V Chr.2: 71349282-71349282 on Build GRCh38
6300327 c.436A>T p.S146C Chr.2: 71349390-71349390 on Build GRCh38
2997310 c.291A>T p.L97F Chr.2: 71348927-71348927 on Build GRCh38
5658398 c.470T>C p.I157T Chr.2: 71349106-71349106 on Build GRCh38
3991427 c.300T>G p.H100Q Chr.2: 71349254-71349254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35601433 Chr.2: 71349092-71349092 on Build GRCh38
rs375591943 Chr.2: 71348922-71348922 on Build GRCh38
rs747048683 Chr.2: 71349212-71349212 on Build GRCh38
rs748329873 Chr.2: 71349201-71349201 on Build GRCh38
rs200279719 Chr.2: 71349315-71349315 on Build GRCh38
rs765518639 Chr.2: 71349279-71349279 on Build GRCh38
rs758857804 Chr.2: 71349284-71349284 on Build GRCh38
rs146722075 Chr.2: 71349087-71349087 on Build GRCh38
rs941029561 Chr.2: 71349002-71349002 on Build GRCh38
rs757260719 Chr.2: 71349310-71349310 on Build GRCh38
rs202042555 Chr.2: 71348980-71348980 on Build GRCh38
rs762639298 Chr.2: 71349053-71349053 on Build GRCh38
rs777855142 Chr.2: 71349287-71349287 on Build GRCh38
rs751321911 Chr.2: 71349058-71349058 on Build GRCh38
rs751129686 Chr.2: 71348975-71348975 on Build GRCh38
rs758554465 Chr.2: 71349208-71349208 on Build GRCh38
rs199581978 Chr.2: 71349283-71349283 on Build GRCh38
rs751234024 Chr.2: 71349133-71349133 on Build GRCh38
rs896369725 Chr.2: 71348919-71348919 on Build GRCh38
rs753315788 Chr.2: 71349383-71349383 on Build GRCh38
rs764789883 Chr.2: 71349084-71349084 on Build GRCh38
rs1008359631 Chr.2: 71349039-71349039 on Build GRCh38
rs147124716 Chr.2: 71349288-71349288 on Build GRCh38
rs748421915 Chr.2: 71349111-71349111 on Build GRCh38
rs758156786 Chr.2: 71348959-71348959 on Build GRCh38
rs1020287160 Chr.2: 71349045-71349045 on Build GRCh38
rs770168685 Chr.2: 71348983-71348983 on Build GRCh38
rs142437672 Chr.2: 71349179-71349179 on Build GRCh38
rs756606982 Chr.2: 71349085-71349085 on Build GRCh38
rs780396533 Chr.2: 71349092-71349092 on Build GRCh38
rs751681811 Chr.2: 71349426-71349426 on Build GRCh38
rs753787540 Chr.2: 71348926-71348926 on Build GRCh38
rs774315651 Chr.2: 71349249-71349249 on Build GRCh38
rs779079673 Chr.2: 71349108-71349108 on Build GRCh38
rs192501942 Chr.2: 71349416-71349416 on Build GRCh38
rs1000190140 Chr.2: 71349238-71349238 on Build GRCh38
rs754774700 Chr.2: 71348939-71348939 on Build GRCh38
rs767009161 Chr.2: 71349137-71349137 on Build GRCh38
rs777668320 Chr.2: 71349198-71349198 on Build GRCh38
rs1056261791 Chr.2: 71349267-71349267 on Build GRCh38
rs771106816 Chr.2: 71349229-71349229 on Build GRCh38
rs777853207 Chr.2: 71349210-71349210 on Build GRCh38
rs527383849 Chr.2: 71349131-71349131 on Build GRCh38
rs12612365 Chr.2: 71349282-71349282 on Build GRCh38
rs756033721 Chr.2: 71348923-71348923 on Build GRCh38
rs772087476 Chr.2: 71349021-71349021 on Build GRCh38
rs765720870 Chr.2: 71349377-71349377 on Build GRCh38
rs781444067 Chr.2: 71349313-71349313 on Build GRCh38
rs772720043 Chr.2: 71349354-71349354 on Build GRCh38
rs201124084 Chr.2: 71349345-71349345 on Build GRCh38
rs747880058 Chr.2: 71349337-71349337 on Build GRCh38
rs777607525 Chr.2: 71348929-71348930 on Build GRCh38
rs139959335 Chr.2: 71349063-71349063 on Build GRCh38
rs370334156 Chr.2: 71349261-71349261 on Build GRCh38
rs756884103 Chr.2: 71348978-71348978 on Build GRCh38
rs772146866 Chr.2: 71349113-71349113 on Build GRCh38
rs917848633 Chr.2: 71349269-71349269 on Build GRCh38
rs898600748 Chr.2: 71348938-71348938 on Build GRCh38
rs775536403 Chr.2: 71349230-71349230 on Build GRCh38
rs1033088307 Chr.2: 71348942-71348942 on Build GRCh38
rs755513799 Chr.2: 71349277-71349277 on Build GRCh38
rs373047103 Chr.2: 71348921-71348921 on Build GRCh38
rs35950410 Chr.2: 71349103-71349104 on Build GRCh38
rs775749985 Chr.2: 71348905-71348905 on Build GRCh38
rs368965334 Chr.2: 71349350-71349350 on Build GRCh38
rs761390533 Chr.2: 71349136-71349136 on Build GRCh38
rs763207310 Chr.2: 71348910-71348910 on Build GRCh38
rs61344341 Chr.2: 71349033-71349033 on Build GRCh38
rs760537038 Chr.2: 71348920-71348920 on Build GRCh38
rs768868859 Chr.2: 71348990-71348990 on Build GRCh38
rs774616770 Chr.2: 71349006-71349006 on Build GRCh38
rs754386185 Chr.2: 71349151-71349151 on Build GRCh38
rs749902397 Chr.2: 71349144-71349144 on Build GRCh38
rs767087150 Chr.2: 71349256-71349256 on Build GRCh38
rs145739027 Chr.2: 71349262-71349262 on Build GRCh38
rs774443569 Chr.2: 71348916-71348916 on Build GRCh38
rs763194025 Chr.2: 71349388-71349388 on Build GRCh38
rs754298277 Chr.2: 71349272-71349272 on Build GRCh38
rs772973354 Chr.2: 71349259-71349259 on Build GRCh38
rs780615135 Chr.2: 71349071-71349071 on Build GRCh38
rs769633167 Chr.2: 71349125-71349125 on Build GRCh38
rs773197512 Chr.2: 71349120-71349120 on Build GRCh38
rs201188979 Chr.2: 71349344-71349344 on Build GRCh38
rs1056777895 Chr.2: 71348917-71348917 on Build GRCh38
rs777353569 Chr.2: 71348960-71348960 on Build GRCh38
rs867183650 Chr.2: 71349007-71349007 on Build GRCh38
rs781719825 Chr.2: 71348979-71348979 on Build GRCh38
rs759979187 Chr.2: 71349374-71349374 on Build GRCh38
rs140659649 Chr.2: 71348953-71348953 on Build GRCh38
rs201231738 Chr.2: 71349187-71349187 on Build GRCh38
rs765925772 Chr.2: 71349271-71349271 on Build GRCh38
rs760789054 Chr.2: 71349242-71349242 on Build GRCh38
rs752345066 Chr.2: 71348955-71348955 on Build GRCh38
rs749286173 Chr.2: 71349231-71349231 on Build GRCh38
rs764567426 Chr.2: 71349406-71349406 on Build GRCh38
rs368733519 Chr.2: 71349355-71349355 on Build GRCh38
rs955295011 Chr.2: 71349323-71349323 on Build GRCh38
rs149932567 Chr.2: 71349099-71349099 on Build GRCh38
rs566104595 Chr.2: 71349124-71349124 on Build GRCh38
rs148746042 Chr.2: 71349042-71349042 on Build GRCh38
rs764107972 Chr.2: 71348911-71348911 on Build GRCh38
rs141395815 Chr.2: 71349055-71349055 on Build GRCh38
rs375345890 Chr.2: 71349217-71349217 on Build GRCh38
rs143340083 Chr.2: 71348994-71348994 on Build GRCh38
rs756584071 Chr.2: 71349147-71349147 on Build GRCh38
rs761459300 Chr.2: 71349250-71349250 on Build GRCh38
rs755317686 Chr.2: 71349171-71349171 on Build GRCh38
rs186891171 Chr.2: 71349362-71349362 on Build GRCh38
rs776123680 Chr.2: 71349368-71349368 on Build GRCh38
rs776612839 Chr.2: 71349027-71349027 on Build GRCh38
rs766794013 Chr.2: 71349149-71349149 on Build GRCh38
rs775628567 Chr.2: 71349127-71349127 on Build GRCh38

View Full Product Details