Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106093871-106094268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UXS1
Entrez Gene ID
80146
Gene Name
UDP-glucuronate decarboxylase 1
Gene Aliases
SDR6E1, UGD
UniGene
Hs.469561
Gene Chromosome Location
Chr.2: 106093303-106194339 on Build GRCh38
Transcripts
NM_001253875.1 NM_001253876.1 NM_025076.4 NR_045607.1 XM_006712775.3 XM_011511902.2 XM_011511903.2 XM_011511904.2 XM_017005008.1 XM_017005009.1 XM_017005010.1 XM_017005011.1 XM_017005012.1 XM_017005013.1 XM_017005014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6819386 c.1253G>A p.R418H Chr.2: 106094036-106094036 on Build GRCh38
714886 c.1169A>T p.Y390F Chr.2: 106094120-106094120 on Build GRCh38
6819390 c.1250C>G p.T417S Chr.2: 106094039-106094039 on Build GRCh38
6819387 c.1268G>A p.R423H Chr.2: 106094036-106094036 on Build GRCh38
714887 c.1184A>T p.Y395F Chr.2: 106094120-106094120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748615729 Chr.2: 106094171-106094174 on Build GRCh38
rs750718890 Chr.2: 106093978-106093978 on Build GRCh38
rs758535566 Chr.2: 106094069-106094069 on Build GRCh38
rs760985922 Chr.2: 106094047-106094047 on Build GRCh38
rs765652023 Chr.2: 106093958-106093959 on Build GRCh38
rs943833139 Chr.2: 106093987-106093987 on Build GRCh38
rs904453181 Chr.2: 106094063-106094063 on Build GRCh38
rs548322158 Chr.2: 106094091-106094091 on Build GRCh38
rs565697301 Chr.2: 106094000-106094000 on Build GRCh38
rs79415522 Chr.2: 106094104-106094104 on Build GRCh38
rs755557725 Chr.2: 106094019-106094021 on Build GRCh38
rs375530882 Chr.2: 106094246-106094246 on Build GRCh38
rs993291500 Chr.2: 106093911-106093911 on Build GRCh38
rs376706115 Chr.2: 106094207-106094207 on Build GRCh38
rs915942487 Chr.2: 106093929-106093929 on Build GRCh38
rs769897233 Chr.2: 106094175-106094178 on Build GRCh38
rs1010666517 Chr.2: 106094113-106094113 on Build GRCh38
rs762499899 Chr.2: 106094133-106094133 on Build GRCh38
rs200449146 Chr.2: 106094208-106094208 on Build GRCh38
rs752895591 Chr.2: 106094059-106094059 on Build GRCh38
rs768124349 Chr.2: 106094152-106094152 on Build GRCh38
rs775117759 Chr.2: 106094116-106094116 on Build GRCh38
rs753156533 Chr.2: 106094094-106094094 on Build GRCh38
rs773625425 Chr.2: 106094153-106094153 on Build GRCh38
rs754061917 Chr.2: 106093989-106093989 on Build GRCh38
rs867543431 Chr.2: 106094107-106094107 on Build GRCh38
rs765616979 Chr.2: 106094187-106094187 on Build GRCh38
rs772072034 Chr.2: 106094005-106094005 on Build GRCh38
rs768079305 Chr.2: 106094042-106094042 on Build GRCh38
rs756488276 Chr.2: 106093979-106093979 on Build GRCh38
rs769314727 Chr.2: 106094022-106094022 on Build GRCh38
rs781384322 Chr.2: 106094073-106094074 on Build GRCh38
rs386648924 Chr.2: 106094210-106094212 on Build GRCh38
rs199726062 Chr.2: 106094212-106094212 on Build GRCh38
rs752065603 Chr.2: 106093984-106093990 on Build GRCh38
rs769369773 Chr.2: 106094115-106094115 on Build GRCh38
rs370584220 Chr.2: 106094017-106094017 on Build GRCh38
rs770734463 Chr.2: 106094014-106094014 on Build GRCh38
rs372232366 Chr.2: 106094075-106094075 on Build GRCh38
rs779091344 Chr.2: 106093999-106093999 on Build GRCh38
rs11268588 Chr.2: 106094212-106094213 on Build GRCh38
rs386390823 Chr.2: 106094238-106094239 on Build GRCh38
rs779140829 Chr.2: 106094053-106094053 on Build GRCh38
rs375972008 Chr.2: 106094013-106094013 on Build GRCh38
rs750908852 Chr.2: 106094043-106094043 on Build GRCh38
rs777851982 Chr.2: 106094011-106094011 on Build GRCh38
rs377734042 Chr.2: 106094210-106094210 on Build GRCh38
rs1006495791 Chr.2: 106093997-106093997 on Build GRCh38
rs367574318 Chr.2: 106094186-106094186 on Build GRCh38
rs374259230 Chr.2: 106094036-106094036 on Build GRCh38
rs183741987 Chr.2: 106094209-106094209 on Build GRCh38
rs973718014 Chr.2: 106093942-106093942 on Build GRCh38
rs577101765 Chr.2: 106094213-106094213 on Build GRCh38
rs762311959 Chr.2: 106094037-106094037 on Build GRCh38
rs36008693 Chr.2: 106094213-106094214 on Build GRCh38
rs371810370 Chr.2: 106094197-106094197 on Build GRCh38
rs532882503 Chr.2: 106093931-106093931 on Build GRCh38
rs547771755 Chr.2: 106093952-106093952 on Build GRCh38
rs776989891 Chr.2: 106094185-106094185 on Build GRCh38
rs912037631 Chr.2: 106093974-106093974 on Build GRCh38
rs766728762 Chr.2: 106094049-106094049 on Build GRCh38
rs928969763 Chr.2: 106093954-106093954 on Build GRCh38
rs368427904 Chr.2: 106094074-106094074 on Build GRCh38
rs571271122 Chr.2: 106093995-106093995 on Build GRCh38
rs970115733 Chr.2: 106094151-106094151 on Build GRCh38
rs200751439 Chr.2: 106094161-106094161 on Build GRCh38
rs373893940 Chr.2: 106093981-106093981 on Build GRCh38
rs766669334 Chr.2: 106094167-106094167 on Build GRCh38
rs970445342 Chr.2: 106093940-106093943 on Build GRCh38
rs1021658251 Chr.2: 106094125-106094125 on Build GRCh38
rs1040835128 Chr.2: 106094221-106094222 on Build GRCh38
rs767881403 Chr.2: 106093977-106093977 on Build GRCh38
rs745518393 Chr.2: 106094114-106094114 on Build GRCh38
rs1991 Chr.2: 106094084-106094084 on Build GRCh38
rs755174060 Chr.2: 106094050-106094050 on Build GRCh38
rs1023465976 Chr.2: 106094128-106094128 on Build GRCh38

View Full Product Details