Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29699976-29700528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TJP1
Entrez Gene ID
7082
Gene Name
tight junction protein 1
Gene Aliases
ZO-1
UniGene
Hs.743990
Gene Chromosome Location
Chr.15: 29699367-29968919 on Build GRCh38
Transcripts
NM_001301025.1 NM_001301026.1 NM_003257.4 NM_175610.3 XM_005254617.3 XM_005254618.3 XM_005254619.3 XM_005254620.3 XM_011521972.2 XM_017022521.1 XM_017022522.1 XM_017022523.1 XM_017022524.1 XM_017022525.1 XM_017022526.1 XM_017022527.1 XM_017022528.1 XM_017022529.1 XM_017022530.1 XM_017022531.1

SNP Detail

SNP ID Chromosome Location
rs914579317 Chr.15: 29700083-29700083 on Build GRCh38
rs531302604 Chr.15: 29700040-29700040 on Build GRCh38
rs147528387 Chr.15: 29700182-29700182 on Build GRCh38
rs922557512 Chr.15: 29700221-29700221 on Build GRCh38
rs913401251 Chr.15: 29700058-29700058 on Build GRCh38
rs557879200 Chr.15: 29700315-29700315 on Build GRCh38
rs1005923828 Chr.15: 29700253-29700253 on Build GRCh38
rs578163155 Chr.15: 29700348-29700348 on Build GRCh38
rs935144282 Chr.15: 29700146-29700146 on Build GRCh38
rs531281823 Chr.15: 29700450-29700450 on Build GRCh38
rs764687619 Chr.15: 29700317-29700318 on Build GRCh38
rs117015824 Chr.15: 29700015-29700015 on Build GRCh38
rs1041063742 Chr.15: 29700193-29700193 on Build GRCh38
rs777571101 Chr.15: 29700445-29700445 on Build GRCh38
rs905619523 Chr.15: 29700305-29700306 on Build GRCh38
rs527594169 Chr.15: 29700140-29700140 on Build GRCh38
rs953643760 Chr.15: 29700057-29700057 on Build GRCh38
rs962519296 Chr.15: 29700026-29700026 on Build GRCh38
rs924760606 Chr.15: 29700138-29700138 on Build GRCh38
rs753668889 Chr.15: 29700375-29700375 on Build GRCh38
rs761014390 Chr.15: 29700160-29700161 on Build GRCh38
rs1006390525 Chr.15: 29700014-29700014 on Build GRCh38
rs969122046 Chr.15: 29700452-29700452 on Build GRCh38
rs571478099 Chr.15: 29700071-29700071 on Build GRCh38
rs41280054 Chr.15: 29700304-29700304 on Build GRCh38
rs1047617769 Chr.15: 29700260-29700260 on Build GRCh38
rs969318668 Chr.15: 29700454-29700454 on Build GRCh38
rs924659627 Chr.15: 29700475-29700475 on Build GRCh38
rs764243134 Chr.15: 29700122-29700122 on Build GRCh38
rs538206506 Chr.15: 29700272-29700272 on Build GRCh38
rs41412345 Chr.15: 29700266-29700266 on Build GRCh38
rs139873144 Chr.15: 29700227-29700227 on Build GRCh38
rs911348156 Chr.15: 29700486-29700486 on Build GRCh38
rs540608291 Chr.15: 29700416-29700416 on Build GRCh38
rs1015998165 Chr.15: 29700034-29700034 on Build GRCh38
rs977476179 Chr.15: 29700473-29700473 on Build GRCh38
rs567164813 Chr.15: 29700206-29700206 on Build GRCh38
rs986071609 Chr.15: 29700067-29700067 on Build GRCh38
rs960285649 Chr.15: 29700043-29700043 on Build GRCh38
rs562445175 Chr.15: 29700039-29700039 on Build GRCh38
rs182915652 Chr.15: 29700241-29700241 on Build GRCh38
rs535013702 Chr.15: 29700121-29700122 on Build GRCh38
rs978944379 Chr.15: 29700469-29700469 on Build GRCh38
rs887432941 Chr.15: 29700230-29700230 on Build GRCh38
rs947463190 Chr.15: 29700092-29700092 on Build GRCh38
rs1056322767 Chr.15: 29700151-29700151 on Build GRCh38
rs779837619 Chr.15: 29700273-29700273 on Build GRCh38
rs187932012 Chr.15: 29700459-29700459 on Build GRCh38
rs979276292 Chr.15: 29700114-29700114 on Build GRCh38
rs942711330 Chr.15: 29700161-29700161 on Build GRCh38
rs565683119 Chr.15: 29700453-29700454 on Build GRCh38
rs953591093 Chr.15: 29700428-29700428 on Build GRCh38
rs947952683 Chr.15: 29700205-29700205 on Build GRCh38
rs1042187984 Chr.15: 29700107-29700107 on Build GRCh38
rs45452501 Chr.15: 29700042-29700042 on Build GRCh38

View Full Product Details