Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1747301-1747900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINF2
Entrez Gene ID
5345
Gene Name
serpin family F member 2
Gene Aliases
A2AP, AAP, ALPHA-2-PI, API, PLI
UniGene
Hs.159509
Gene Chromosome Location
Chr.17: 1742808-1755265 on Build GRCh38
Transcripts
NM_000934.3 NM_001165920.1 NM_001165921.1 XM_005256701.4 XM_017024765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
704887 c.526G>C p.E176Q Chr.17: 1747323-1747323 on Build GRCh38
5385767 c.641G>A p.G214E Chr.17: 1747438-1747438 on Build GRCh38
7230324 c.672G>A p.P224P Chr.17: 1747469-1747469 on Build GRCh38
5972065 c.546C>T p.S182S Chr.17: 1747343-1747343 on Build GRCh38
7327657 c.671C>G p.P224R Chr.17: 1747468-1747468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773144356 Chr.17: 1747546-1747546 on Build GRCh38
rs898312344 Chr.17: 1747608-1747608 on Build GRCh38
rs889916484 Chr.17: 1747627-1747627 on Build GRCh38
rs771886951 Chr.17: 1747528-1747528 on Build GRCh38
rs556652068 Chr.17: 1747633-1747633 on Build GRCh38
rs909653557 Chr.17: 1747479-1747479 on Build GRCh38
rs1043051958 Chr.17: 1747562-1747562 on Build GRCh38
rs201169280 Chr.17: 1747454-1747454 on Build GRCh38
rs1012335904 Chr.17: 1747869-1747869 on Build GRCh38
rs541732263 Chr.17: 1747595-1747595 on Build GRCh38
rs770506258 Chr.17: 1747467-1747467 on Build GRCh38
rs148076491 Chr.17: 1747352-1747352 on Build GRCh38
rs767167105 Chr.17: 1747368-1747368 on Build GRCh38
rs1014074888 Chr.17: 1747697-1747697 on Build GRCh38
rs778254669 Chr.17: 1747526-1747526 on Build GRCh38
rs112409977 Chr.17: 1747844-1747844 on Build GRCh38
rs760773008 Chr.17: 1747376-1747376 on Build GRCh38
rs969787874 Chr.17: 1747753-1747753 on Build GRCh38
rs931129132 Chr.17: 1747843-1747843 on Build GRCh38
rs774020894 Chr.17: 1747469-1747469 on Build GRCh38
rs533358132 Chr.17: 1747657-1747657 on Build GRCh38
rs121965061 Chr.17: 1747323-1747325 on Build GRCh38
rs928509327 Chr.17: 1747569-1747569 on Build GRCh38
rs112549101 Chr.17: 1747516-1747516 on Build GRCh38
rs182062385 Chr.17: 1747796-1747796 on Build GRCh38
rs367789714 Chr.17: 1747658-1747658 on Build GRCh38
rs1008331473 Chr.17: 1747853-1747853 on Build GRCh38
rs958321590 Chr.17: 1747862-1747862 on Build GRCh38
rs567943316 Chr.17: 1747816-1747816 on Build GRCh38
rs1031755033 Chr.17: 1747646-1747646 on Build GRCh38
rs939966779 Chr.17: 1747574-1747574 on Build GRCh38
rs1033030591 Chr.17: 1747710-1747710 on Build GRCh38
rs369175580 Chr.17: 1747867-1747867 on Build GRCh38
rs373646983 Chr.17: 1747334-1747334 on Build GRCh38
rs967334132 Chr.17: 1747638-1747638 on Build GRCh38
rs746955915 Chr.17: 1747535-1747535 on Build GRCh38
rs533821579 Chr.17: 1747817-1747817 on Build GRCh38
rs915658977 Chr.17: 1747750-1747750 on Build GRCh38
rs541645773 Chr.17: 1747855-1747855 on Build GRCh38
rs747440872 Chr.17: 1747433-1747433 on Build GRCh38
rs978629835 Chr.17: 1747464-1747464 on Build GRCh38
rs748952197 Chr.17: 1747460-1747460 on Build GRCh38
rs200102868 Chr.17: 1747533-1747533 on Build GRCh38
rs572431803 Chr.17: 1747626-1747626 on Build GRCh38
rs373848993 Chr.17: 1747539-1747539 on Build GRCh38
rs778971451 Chr.17: 1747415-1747415 on Build GRCh38
rs753898740 Chr.17: 1747391-1747391 on Build GRCh38
rs999964227 Chr.17: 1747859-1747859 on Build GRCh38
rs995370504 Chr.17: 1747617-1747617 on Build GRCh38
rs775122469 Chr.17: 1747490-1747490 on Build GRCh38
rs761986448 Chr.17: 1747540-1747540 on Build GRCh38
rs768601607 Chr.17: 1747517-1747518 on Build GRCh38
rs781699275 Chr.17: 1747520-1747520 on Build GRCh38
rs561718644 Chr.17: 1747625-1747625 on Build GRCh38
rs772083726 Chr.17: 1747487-1747487 on Build GRCh38
rs765557830 Chr.17: 1747543-1747543 on Build GRCh38
rs371366332 Chr.17: 1747532-1747532 on Build GRCh38
rs1032588676 Chr.17: 1747860-1747860 on Build GRCh38
rs868831118 Chr.17: 1747366-1747366 on Build GRCh38
rs780242777 Chr.17: 1747428-1747428 on Build GRCh38
rs757000660 Chr.17: 1747524-1747524 on Build GRCh38
rs370865079 Chr.17: 1747436-1747436 on Build GRCh38
rs750552019 Chr.17: 1747515-1747515 on Build GRCh38
rs928340069 Chr.17: 1747762-1747762 on Build GRCh38
rs60292203 Chr.17: 1747559-1747559 on Build GRCh38
rs10438725 Chr.17: 1747871-1747871 on Build GRCh38
rs768384244 Chr.17: 1747538-1747538 on Build GRCh38
rs563880121 Chr.17: 1747691-1747691 on Build GRCh38
rs376980433 Chr.17: 1747517-1747517 on Build GRCh38
rs759528674 Chr.17: 1747367-1747367 on Build GRCh38
rs990981963 Chr.17: 1747738-1747738 on Build GRCh38
rs202070354 Chr.17: 1747419-1747419 on Build GRCh38
rs56230135 Chr.17: 1747724-1747724 on Build GRCh38
rs774362776 Chr.17: 1747365-1747365 on Build GRCh38
rs762039372 Chr.17: 1747504-1747504 on Build GRCh38
rs758970141 Chr.17: 1747421-1747421 on Build GRCh38
rs1019618102 Chr.17: 1747634-1747634 on Build GRCh38
rs377667967 Chr.17: 1747343-1747343 on Build GRCh38
rs376742506 Chr.17: 1747856-1747856 on Build GRCh38
rs775919506 Chr.17: 1747340-1747340 on Build GRCh38
rs375195777 Chr.17: 1747508-1747508 on Build GRCh38
rs915821280 Chr.17: 1747384-1747384 on Build GRCh38
rs796097753 Chr.17: 1747833-1747833 on Build GRCh38
rs752567596 Chr.17: 1747372-1747372 on Build GRCh38
rs1041421726 Chr.17: 1747854-1747854 on Build GRCh38
rs536156123 Chr.17: 1747348-1747348 on Build GRCh38
rs981485658 Chr.17: 1747754-1747754 on Build GRCh38
rs78558219 Chr.17: 1747379-1747379 on Build GRCh38
rs202133429 Chr.17: 1747542-1747542 on Build GRCh38
rs138097660 Chr.17: 1747497-1747497 on Build GRCh38
rs564643549 Chr.17: 1747650-1747650 on Build GRCh38
rs904618141 Chr.17: 1747582-1747582 on Build GRCh38
rs748899037 Chr.17: 1747523-1747523 on Build GRCh38
rs745429351 Chr.17: 1747527-1747527 on Build GRCh38
rs553862155 Chr.17: 1747478-1747478 on Build GRCh38
rs575887048 Chr.17: 1747563-1747563 on Build GRCh38
rs746257917 Chr.17: 1747773-1747774 on Build GRCh38
rs150369383 Chr.17: 1747496-1747496 on Build GRCh38
rs909518009 Chr.17: 1747773-1747773 on Build GRCh38
rs771299356 Chr.17: 1747674-1747674 on Build GRCh38
rs1020280805 Chr.17: 1747378-1747378 on Build GRCh38
rs889842909 Chr.17: 1747849-1747849 on Build GRCh38
rs753995965 Chr.17: 1747498-1747498 on Build GRCh38
rs757397709 Chr.17: 1747406-1747406 on Build GRCh38

View Full Product Details