Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41184717-41185271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP4-1
Entrez Gene ID
85285
Gene Name
keratin associated protein 4-1
Gene Aliases
KAP4.1, KAP4.10, KRTAP4-10, KRTAP4.10
UniGene
Hs.437079
Gene Chromosome Location
Chr.17: 41184100-41184895 on Build GRCh38
Transcripts
NM_033060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349324 c.37C>A p.Q13K Chr.17: 41184818-41184818 on Build GRCh38
7250594 c.23C>G p.S8C Chr.17: 41184832-41184832 on Build GRCh38
4066304 c.113C>A p.S38Y Chr.17: 41184730-41184730 on Build GRCh38
4066306 c.32C>A p.S11Y Chr.17: 41184823-41184823 on Build GRCh38
979084 c.6T>C p.V2V Chr.17: 41184849-41184849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757046245 Chr.17: 41184804-41184805 on Build GRCh38
rs965174824 Chr.17: 41185198-41185198 on Build GRCh38
rs748214633 Chr.17: 41184931-41184931 on Build GRCh38
rs781162300 Chr.17: 41185163-41185163 on Build GRCh38
rs773563652 Chr.17: 41184935-41184935 on Build GRCh38
rs539460060 Chr.17: 41185044-41185044 on Build GRCh38
rs549268569 Chr.17: 41184986-41184986 on Build GRCh38
rs757999294 Chr.17: 41184871-41184871 on Build GRCh38
rs909097771 Chr.17: 41185103-41185103 on Build GRCh38
rs189088133 Chr.17: 41184891-41184891 on Build GRCh38
rs373729561 Chr.17: 41185042-41185042 on Build GRCh38
rs771294601 Chr.17: 41184751-41184753 on Build GRCh38
rs766976352 Chr.17: 41184828-41184828 on Build GRCh38
rs765063028 Chr.17: 41184807-41184812 on Build GRCh38
rs940613001 Chr.17: 41185110-41185110 on Build GRCh38
rs894261765 Chr.17: 41185216-41185216 on Build GRCh38
rs771143053 Chr.17: 41184942-41184942 on Build GRCh38
rs764662520 Chr.17: 41184852-41184852 on Build GRCh38
rs760686634 Chr.17: 41184792-41184793 on Build GRCh38
rs1055950694 Chr.17: 41185213-41185213 on Build GRCh38
rs764248874 Chr.17: 41184798-41184799 on Build GRCh38
rs746882953 Chr.17: 41184741-41184741 on Build GRCh38
rs976558880 Chr.17: 41185010-41185010 on Build GRCh38
rs12449316 Chr.17: 41185221-41185221 on Build GRCh38
rs369645187 Chr.17: 41184778-41184778 on Build GRCh38
rs760915633 Chr.17: 41184824-41184824 on Build GRCh38
rs753397895 Chr.17: 41184784-41184784 on Build GRCh38
rs145814064 Chr.17: 41184817-41184817 on Build GRCh38
rs538892734 Chr.17: 41185142-41185142 on Build GRCh38
rs1042732248 Chr.17: 41185231-41185231 on Build GRCh38
rs932028941 Chr.17: 41184786-41184786 on Build GRCh38
rs966087954 Chr.17: 41185000-41185000 on Build GRCh38
rs894116499 Chr.17: 41184881-41184881 on Build GRCh38
rs909941353 Chr.17: 41184869-41184869 on Build GRCh38
rs1022962150 Chr.17: 41184899-41184899 on Build GRCh38
rs776990643 Chr.17: 41184838-41184838 on Build GRCh38
rs772461380 Chr.17: 41184932-41184932 on Build GRCh38
rs761213270 Chr.17: 41184771-41184771 on Build GRCh38
rs1036130282 Chr.17: 41185116-41185116 on Build GRCh38
rs755113155 Chr.17: 41184921-41184921 on Build GRCh38
rs185774815 Chr.17: 41184779-41184779 on Build GRCh38
rs760022177 Chr.17: 41184767-41184767 on Build GRCh38
rs1003699755 Chr.17: 41184930-41184930 on Build GRCh38
rs61750924 Chr.17: 41184796-41184796 on Build GRCh38
rs764999546 Chr.17: 41184787-41184787 on Build GRCh38
rs560219723 Chr.17: 41184914-41184914 on Build GRCh38
rs774769135 Chr.17: 41184764-41184778 on Build GRCh38
rs201534080 Chr.17: 41185099-41185099 on Build GRCh38
rs754440596 Chr.17: 41184785-41184785 on Build GRCh38
rs748002876 Chr.17: 41184736-41184736 on Build GRCh38
rs751400050 Chr.17: 41184882-41184882 on Build GRCh38
rs1055268196 Chr.17: 41184962-41184962 on Build GRCh38
rs752336099 Chr.17: 41184876-41184876 on Build GRCh38
rs774716171 Chr.17: 41184816-41184816 on Build GRCh38
rs978216857 Chr.17: 41184924-41184924 on Build GRCh38
rs554560020 Chr.17: 41184802-41184802 on Build GRCh38
rs775592922 Chr.17: 41184749-41184749 on Build GRCh38
rs452531 Chr.17: 41184906-41184906 on Build GRCh38
rs752379914 Chr.17: 41184854-41184854 on Build GRCh38
rs1007409 Chr.17: 41184759-41184759 on Build GRCh38
rs762278055 Chr.17: 41184765-41184765 on Build GRCh38
rs372374744 Chr.17: 41184753-41184753 on Build GRCh38
rs752289958 Chr.17: 41184793-41184793 on Build GRCh38
rs759100768 Chr.17: 41184840-41184840 on Build GRCh38
rs768965840 Chr.17: 41184755-41184755 on Build GRCh38
rs769798959 Chr.17: 41184744-41184744 on Build GRCh38
rs768139539 Chr.17: 41184776-41184776 on Build GRCh38
rs895402863 Chr.17: 41184984-41184984 on Build GRCh38
rs547955212 Chr.17: 41185181-41185181 on Build GRCh38
rs758100896 Chr.17: 41184878-41184878 on Build GRCh38
rs769318596 Chr.17: 41185003-41185003 on Build GRCh38
rs767924599 Chr.17: 41184768-41184768 on Build GRCh38
rs181038520 Chr.17: 41185045-41185045 on Build GRCh38
rs775549382 Chr.17: 41184790-41184791 on Build GRCh38
rs999009800 Chr.17: 41185119-41185119 on Build GRCh38
rs449817 Chr.17: 41185014-41185014 on Build GRCh38
rs750444393 Chr.17: 41184831-41184832 on Build GRCh38
rs745720941 Chr.17: 41184915-41184915 on Build GRCh38
rs550482171 Chr.17: 41185160-41185161 on Build GRCh38
rs200327404 Chr.17: 41184821-41184821 on Build GRCh38
rs780964584 Chr.17: 41184813-41184813 on Build GRCh38
rs777703211 Chr.17: 41184737-41184737 on Build GRCh38
rs571636030 Chr.17: 41184811-41184811 on Build GRCh38
rs148996298 Chr.17: 41185132-41185132 on Build GRCh38
rs767645960 Chr.17: 41184787-41184788 on Build GRCh38
rs759302145 Chr.17: 41185055-41185055 on Build GRCh38
rs385122 Chr.17: 41185042-41185042 on Build GRCh38
rs984840927 Chr.17: 41185066-41185066 on Build GRCh38
rs938544138 Chr.17: 41185183-41185183 on Build GRCh38
rs373830142 Chr.17: 41184941-41184941 on Build GRCh38
rs61750930 Chr.17: 41184815-41184815 on Build GRCh38
rs778690929 Chr.17: 41184853-41184853 on Build GRCh38
rs754053739 Chr.17: 41184801-41184802 on Build GRCh38
rs757270485 Chr.17: 41184809-41184809 on Build GRCh38
rs968441328 Chr.17: 41185088-41185088 on Build GRCh38

View Full Product Details