Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89923021-89923611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMRN1
Entrez Gene ID
22915
Gene Name
multimerin 1
Gene Aliases
ECM, EMILIN4, GPIa*, MMRN
UniGene
Hs.268107
Gene Chromosome Location
Chr.4: 89879539-89954629 on Build GRCh38
Transcripts
NM_007351.2 XM_005262856.2 XM_017007893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186534 c.851C>T p.A284V Chr.4: 89923168-89923168 on Build GRCh38
6207678 c.927G>T p.Q309H Chr.4: 89923244-89923244 on Build GRCh38
5671362 c.955G>T p.E319* Chr.4: 89923272-89923272 on Build GRCh38
4625365 c.921T>C p.A307A Chr.4: 89923238-89923238 on Build GRCh38
3826438 c.878C>T p.T293I Chr.4: 89923195-89923195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542905278 Chr.4: 89923115-89923115 on Build GRCh38
rs894675941 Chr.4: 89923068-89923068 on Build GRCh38
rs948924607 Chr.4: 89923498-89923498 on Build GRCh38
rs927547799 Chr.4: 89923510-89923510 on Build GRCh38
rs762829732 Chr.4: 89923123-89923123 on Build GRCh38
rs761344505 Chr.4: 89923050-89923050 on Build GRCh38
rs994495320 Chr.4: 89923281-89923281 on Build GRCh38
rs971598389 Chr.4: 89923461-89923461 on Build GRCh38
rs907184540 Chr.4: 89923145-89923145 on Build GRCh38
rs868044004 Chr.4: 89923443-89923443 on Build GRCh38
rs1027354745 Chr.4: 89923367-89923367 on Build GRCh38
rs768679686 Chr.4: 89923127-89923127 on Build GRCh38
rs35595279 Chr.4: 89923049-89923050 on Build GRCh38
rs936006020 Chr.4: 89923070-89923070 on Build GRCh38
rs528835721 Chr.4: 89923169-89923169 on Build GRCh38
rs759807625 Chr.4: 89923287-89923287 on Build GRCh38
rs917449670 Chr.4: 89923043-89923043 on Build GRCh38
rs907469192 Chr.4: 89923520-89923520 on Build GRCh38
rs977740429 Chr.4: 89923405-89923405 on Build GRCh38
rs991011336 Chr.4: 89923447-89923447 on Build GRCh38
rs767126678 Chr.4: 89923054-89923054 on Build GRCh38
rs139544662 Chr.4: 89923225-89923225 on Build GRCh38
rs774347853 Chr.4: 89923237-89923237 on Build GRCh38
rs75121139 Chr.4: 89923583-89923583 on Build GRCh38
rs1054366951 Chr.4: 89923559-89923559 on Build GRCh38
rs369184053 Chr.4: 89923410-89923410 on Build GRCh38
rs760368389 Chr.4: 89923267-89923267 on Build GRCh38
rs916070007 Chr.4: 89923498-89923499 on Build GRCh38
rs755892330 Chr.4: 89923246-89923246 on Build GRCh38
rs913253091 Chr.4: 89923574-89923574 on Build GRCh38
rs866135923 Chr.4: 89923389-89923389 on Build GRCh38
rs772972478 Chr.4: 89923266-89923266 on Build GRCh38
rs760070578 Chr.4: 89923097-89923097 on Build GRCh38
rs1047764818 Chr.4: 89923563-89923563 on Build GRCh38
rs34848559 Chr.4: 89923073-89923074 on Build GRCh38
rs764717626 Chr.4: 89923170-89923170 on Build GRCh38
rs143280923 Chr.4: 89923232-89923232 on Build GRCh38
rs890307548 Chr.4: 89923200-89923200 on Build GRCh38
rs746138565 Chr.4: 89923197-89923197 on Build GRCh38
rs41284765 Chr.4: 89923160-89923160 on Build GRCh38
rs966325861 Chr.4: 89923348-89923349 on Build GRCh38
rs927265069 Chr.4: 89923065-89923065 on Build GRCh38
rs147134600 Chr.4: 89923132-89923132 on Build GRCh38
rs753424302 Chr.4: 89923168-89923168 on Build GRCh38
rs751806264 Chr.4: 89923173-89923173 on Build GRCh38
rs1054637559 Chr.4: 89923049-89923049 on Build GRCh38
rs775948921 Chr.4: 89923275-89923275 on Build GRCh38
rs984014100 Chr.4: 89923041-89923041 on Build GRCh38
rs936285441 Chr.4: 89923540-89923540 on Build GRCh38
rs200640234 Chr.4: 89923166-89923166 on Build GRCh38
rs368377681 Chr.4: 89923153-89923153 on Build GRCh38
rs764506443 Chr.4: 89923317-89923317 on Build GRCh38
rs1045959500 Chr.4: 89923499-89923499 on Build GRCh38
rs936426130 Chr.4: 89923435-89923435 on Build GRCh38
rs749756859 Chr.4: 89923129-89923130 on Build GRCh38
rs980723363 Chr.4: 89923481-89923481 on Build GRCh38
rs144508424 Chr.4: 89923550-89923550 on Build GRCh38
rs2870032 Chr.4: 89923109-89923109 on Build GRCh38
rs1013161136 Chr.4: 89923086-89923086 on Build GRCh38
rs750109004 Chr.4: 89923158-89923158 on Build GRCh38
rs770969212 Chr.4: 89923239-89923240 on Build GRCh38
rs774462590 Chr.4: 89923240-89923242 on Build GRCh38
rs74718110 Chr.4: 89923432-89923432 on Build GRCh38
rs535288773 Chr.4: 89923268-89923268 on Build GRCh38
rs1042589290 Chr.4: 89923185-89923185 on Build GRCh38
rs146692577 Chr.4: 89923254-89923254 on Build GRCh38
rs555578984 Chr.4: 89923370-89923370 on Build GRCh38
rs757686466 Chr.4: 89923181-89923181 on Build GRCh38
rs781379902 Chr.4: 89923191-89923191 on Build GRCh38
rs761760888 Chr.4: 89923136-89923136 on Build GRCh38
rs772047269 Chr.4: 89923256-89923256 on Build GRCh38
rs759092654 Chr.4: 89923307-89923307 on Build GRCh38
rs374985221 Chr.4: 89923211-89923211 on Build GRCh38
rs983240667 Chr.4: 89923392-89923392 on Build GRCh38
rs779956739 Chr.4: 89923214-89923214 on Build GRCh38

View Full Product Details