Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 150583825-150584373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRBA
Entrez Gene ID
987
Gene Name
LPS responsive beige-like anchor protein
Gene Aliases
BGL, CDC4L, CVID8, LAB300, LBA
UniGene
Hs.480938
Gene Chromosome Location
Chr.4: 150264515-151015725 on Build GRCh38
Transcripts
NM_001199282.2 NM_006726.4 XM_005263372.3 XM_005263373.2 XM_005263374.3 XM_005263375.3 XM_011532434.2 XM_017008872.1 NM_006439.4

Gene Symbol
MAB21L2
Entrez Gene ID
10586
Gene Name
mab-21 like 2
Gene Aliases
MCOPS14
UniGene
Hs.584852
Gene Chromosome Location
Chr.4: 150581925-150584693 on Build GRCh38
Transcripts
NM_001199282.2 NM_006726.4 XM_005263372.3 XM_005263373.2 XM_005263374.3 XM_005263375.3 XM_011532434.2 XM_017008872.1 NM_006439.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4122714 c.889G>T p.G297C Chr.4: 150583918-150583918 on Build GRCh38
1052005 c.832C>T p.L278L Chr.4: 150583861-150583861 on Build GRCh38
1052006 c.895C>T p.R299W Chr.4: 150583924-150583924 on Build GRCh38
3127585 c.835C>T p.L279L Chr.4: 150583864-150583864 on Build GRCh38
4330100 c.981T>C p.F327F Chr.4: 150584010-150584010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781395062 Chr.4: 150583972-150583972 on Build GRCh38
rs1027462535 Chr.4: 150584142-150584142 on Build GRCh38
rs761277103 Chr.4: 150584040-150584040 on Build GRCh38
rs372689984 Chr.4: 150584145-150584145 on Build GRCh38
rs1053934101 Chr.4: 150583898-150583898 on Build GRCh38
rs755146953 Chr.4: 150583953-150583953 on Build GRCh38
rs995966739 Chr.4: 150583942-150583942 on Build GRCh38
rs147719473 Chr.4: 150583929-150583929 on Build GRCh38
rs527720888 Chr.4: 150584197-150584197 on Build GRCh38
rs75417434 Chr.4: 150584353-150584353 on Build GRCh38
rs775358183 Chr.4: 150584027-150584027 on Build GRCh38
rs760139505 Chr.4: 150584103-150584103 on Build GRCh38
rs776306456 Chr.4: 150583897-150583897 on Build GRCh38
rs781360120 Chr.4: 150584110-150584110 on Build GRCh38
rs748153112 Chr.4: 150583916-150583916 on Build GRCh38
rs1050164716 Chr.4: 150584329-150584329 on Build GRCh38
rs768096574 Chr.4: 150584109-150584109 on Build GRCh38
rs932137000 Chr.4: 150584310-150584310 on Build GRCh38
rs1003204803 Chr.4: 150583911-150583911 on Build GRCh38
rs75096474 Chr.4: 150584192-150584192 on Build GRCh38
rs929998654 Chr.4: 150584255-150584255 on Build GRCh38
rs748185948 Chr.4: 150583974-150583974 on Build GRCh38
rs1008507357 Chr.4: 150584077-150584077 on Build GRCh38
rs776887007 Chr.4: 150584144-150584144 on Build GRCh38
rs186145445 Chr.4: 150584005-150584005 on Build GRCh38
rs200543214 Chr.4: 150584007-150584007 on Build GRCh38
rs752099479 Chr.4: 150583932-150583932 on Build GRCh38
rs766732897 Chr.4: 150584081-150584081 on Build GRCh38
rs13105060 Chr.4: 150584170-150584170 on Build GRCh38
rs1054432582 Chr.4: 150584257-150584257 on Build GRCh38
rs768204456 Chr.4: 150584028-150584028 on Build GRCh38
rs935567752 Chr.4: 150584240-150584240 on Build GRCh38
rs750789167 Chr.4: 150583926-150583926 on Build GRCh38
rs769949216 Chr.4: 150584171-150584171 on Build GRCh38
rs761433893 Chr.4: 150583901-150583901 on Build GRCh38
rs376960539 Chr.4: 150584157-150584157 on Build GRCh38
rs777985059 Chr.4: 150583984-150583984 on Build GRCh38
rs893995299 Chr.4: 150584286-150584286 on Build GRCh38
rs752884805 Chr.4: 150584127-150584128 on Build GRCh38
rs780702515 Chr.4: 150583905-150583905 on Build GRCh38
rs961899601 Chr.4: 150584119-150584119 on Build GRCh38
rs769443445 Chr.4: 150584046-150584046 on Build GRCh38
rs774309721 Chr.4: 150583849-150583849 on Build GRCh38
rs763591175 Chr.4: 150583895-150583895 on Build GRCh38
rs764824103 Chr.4: 150583906-150583906 on Build GRCh38
rs973664516 Chr.4: 150584134-150584134 on Build GRCh38
rs567573442 Chr.4: 150584235-150584235 on Build GRCh38
rs745635503 Chr.4: 150583857-150583857 on Build GRCh38
rs893558986 Chr.4: 150583915-150583915 on Build GRCh38
rs368334008 Chr.4: 150583872-150583872 on Build GRCh38
rs548849366 Chr.4: 150584352-150584352 on Build GRCh38
rs999696293 Chr.4: 150583861-150583861 on Build GRCh38
rs769158625 Chr.4: 150584001-150584001 on Build GRCh38
rs763291410 Chr.4: 150584073-150584073 on Build GRCh38
rs562705013 Chr.4: 150584055-150584055 on Build GRCh38
rs201523389 Chr.4: 150583983-150583983 on Build GRCh38
rs949400192 Chr.4: 150584208-150584208 on Build GRCh38
rs772060357 Chr.4: 150584316-150584316 on Build GRCh38
rs543223781 Chr.4: 150584029-150584029 on Build GRCh38
rs115384050 Chr.4: 150584223-150584223 on Build GRCh38
rs759032383 Chr.4: 150583885-150583885 on Build GRCh38
rs902733649 Chr.4: 150583848-150583848 on Build GRCh38
rs540652583 Chr.4: 150583869-150583869 on Build GRCh38

View Full Product Details