Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177090621-177091216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR4
Entrez Gene ID
2264
Gene Name
fibroblast growth factor receptor 4
Gene Aliases
CD334, JTK2, TKF
UniGene
Hs.165950
Gene Chromosome Location
Chr.5: 177086872-177098144 on Build GRCh38
Transcripts
NM_001291980.1 NM_002011.4 NM_022963.3 NM_213647.2 XM_005265838.3 XM_011534464.2 XM_011534465.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4497024 c.488C>T p.P163L Chr.5: 177090989-177090989 on Build GRCh38
7428022 c.537G>T p.T179T Chr.5: 177091038-177091038 on Build GRCh38
5608831 c.529A>C p.N177H Chr.5: 177091030-177091030 on Build GRCh38
1644607 c.577G>A p.G193R Chr.5: 177091078-177091078 on Build GRCh38
1066154 c.486A>G p.V162V Chr.5: 177090987-177090987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764916049 Chr.5: 177090974-177090974 on Build GRCh38
rs200285656 Chr.5: 177090702-177090702 on Build GRCh38
rs11549940 Chr.5: 177091071-177091071 on Build GRCh38
rs746832173 Chr.5: 177090782-177090782 on Build GRCh38
rs763005603 Chr.5: 177090683-177090683 on Build GRCh38
rs941187254 Chr.5: 177090913-177090913 on Build GRCh38
rs899749434 Chr.5: 177090940-177090940 on Build GRCh38
rs144853017 Chr.5: 177090648-177090648 on Build GRCh38
rs745405031 Chr.5: 177090896-177090897 on Build GRCh38
rs142584511 Chr.5: 177090999-177090999 on Build GRCh38
rs200854398 Chr.5: 177090679-177090679 on Build GRCh38
rs761185822 Chr.5: 177090942-177090942 on Build GRCh38
rs756467835 Chr.5: 177090728-177090728 on Build GRCh38
rs751043027 Chr.5: 177091070-177091070 on Build GRCh38
rs55675160 Chr.5: 177091036-177091036 on Build GRCh38
rs749977519 Chr.5: 177090864-177090864 on Build GRCh38
rs765645152 Chr.5: 177090707-177090707 on Build GRCh38
rs915910799 Chr.5: 177090706-177090706 on Build GRCh38
rs139464025 Chr.5: 177091012-177091012 on Build GRCh38
rs138995195 Chr.5: 177090686-177090686 on Build GRCh38
rs759266171 Chr.5: 177090699-177090699 on Build GRCh38
rs771235910 Chr.5: 177090895-177090895 on Build GRCh38
rs562836932 Chr.5: 177090981-177090981 on Build GRCh38
rs774681708 Chr.5: 177090900-177090900 on Build GRCh38
rs142240686 Chr.5: 177090768-177090768 on Build GRCh38
rs546558669 Chr.5: 177091069-177091069 on Build GRCh38
rs760059744 Chr.5: 177090697-177090697 on Build GRCh38
rs150920445 Chr.5: 177091003-177091003 on Build GRCh38
rs1033228084 Chr.5: 177091062-177091062 on Build GRCh38
rs773464620 Chr.5: 177090682-177090682 on Build GRCh38
rs1022951577 Chr.5: 177090998-177090998 on Build GRCh38
rs762538901 Chr.5: 177091065-177091065 on Build GRCh38
rs1019188558 Chr.5: 177090889-177090889 on Build GRCh38
rs765421192 Chr.5: 177090681-177090681 on Build GRCh38
rs987590875 Chr.5: 177090772-177090772 on Build GRCh38
rs1046287376 Chr.5: 177090746-177090746 on Build GRCh38
rs926007365 Chr.5: 177090649-177090649 on Build GRCh38
rs780979155 Chr.5: 177090887-177090887 on Build GRCh38
rs186636834 Chr.5: 177091184-177091184 on Build GRCh38
rs758945225 Chr.5: 177090889-177090890 on Build GRCh38
rs375497918 Chr.5: 177090708-177090708 on Build GRCh38
rs768336544 Chr.5: 177090885-177090885 on Build GRCh38
rs999035285 Chr.5: 177091194-177091194 on Build GRCh38
rs888774380 Chr.5: 177090874-177090874 on Build GRCh38
rs777645394 Chr.5: 177091088-177091088 on Build GRCh38
rs376618 Chr.5: 177090796-177090796 on Build GRCh38
rs777012486 Chr.5: 177090959-177090959 on Build GRCh38
rs959223871 Chr.5: 177091080-177091080 on Build GRCh38
rs184093905 Chr.5: 177090764-177090764 on Build GRCh38
rs745738782 Chr.5: 177090651-177090651 on Build GRCh38
rs753223974 Chr.5: 177090807-177090807 on Build GRCh38
rs116920044 Chr.5: 177090691-177090691 on Build GRCh38
rs147846257 Chr.5: 177091103-177091103 on Build GRCh38
rs773378049 Chr.5: 177090789-177090789 on Build GRCh38
rs775590700 Chr.5: 177091024-177091024 on Build GRCh38
rs757884615 Chr.5: 177090865-177090865 on Build GRCh38
rs200864630 Chr.5: 177091077-177091077 on Build GRCh38
rs768366360 Chr.5: 177090922-177090923 on Build GRCh38
rs201706535 Chr.5: 177090856-177090856 on Build GRCh38
rs762129323 Chr.5: 177090680-177090680 on Build GRCh38
rs777338486 Chr.5: 177090730-177090730 on Build GRCh38
rs747999569 Chr.5: 177091008-177091008 on Build GRCh38
rs760718603 Chr.5: 177091145-177091145 on Build GRCh38
rs746085347 Chr.5: 177091017-177091017 on Build GRCh38
rs1038266539 Chr.5: 177090930-177090930 on Build GRCh38
rs774203148 Chr.5: 177090954-177090954 on Build GRCh38
rs749013753 Chr.5: 177090920-177090920 on Build GRCh38
rs748583503 Chr.5: 177091127-177091127 on Build GRCh38
rs779938731 Chr.5: 177090777-177090777 on Build GRCh38
rs45526036 Chr.5: 177090990-177090990 on Build GRCh38
rs767542365 Chr.5: 177091085-177091085 on Build GRCh38
rs770660884 Chr.5: 177091148-177091148 on Build GRCh38
rs775928328 Chr.5: 177090906-177090906 on Build GRCh38
rs201257476 Chr.5: 177090962-177090962 on Build GRCh38
rs1016993541 Chr.5: 177090643-177090643 on Build GRCh38
rs111435837 Chr.5: 177091037-177091037 on Build GRCh38
rs772264286 Chr.5: 177090905-177090905 on Build GRCh38
rs920435389 Chr.5: 177091064-177091064 on Build GRCh38
rs376400659 Chr.5: 177090739-177090739 on Build GRCh38
rs765844308 Chr.5: 177090992-177090992 on Build GRCh38
rs759737475 Chr.5: 177090904-177090904 on Build GRCh38
rs2656910 Chr.5: 177090687-177090687 on Build GRCh38
rs779677763 Chr.5: 177090915-177090915 on Build GRCh38
rs752945069 Chr.5: 177090852-177090852 on Build GRCh38
rs751803683 Chr.5: 177091138-177091138 on Build GRCh38
rs755733945 Chr.5: 177090899-177090899 on Build GRCh38
rs752670660 Chr.5: 177091086-177091086 on Build GRCh38
rs908645579 Chr.5: 177091173-177091173 on Build GRCh38
rs779917772 Chr.5: 177090871-177090871 on Build GRCh38
rs764387996 Chr.5: 177090957-177090957 on Build GRCh38
rs55701137 Chr.5: 177090800-177090800 on Build GRCh38
rs74633199 Chr.5: 177090819-177090819 on Build GRCh38
rs1038618352 Chr.5: 177091195-177091195 on Build GRCh38
rs752961730 Chr.5: 177090714-177090714 on Build GRCh38
rs370636231 Chr.5: 177090855-177090855 on Build GRCh38
rs149737957 Chr.5: 177091049-177091049 on Build GRCh38
rs370322847 Chr.5: 177090765-177090765 on Build GRCh38
rs560823913 Chr.5: 177090798-177090798 on Build GRCh38
rs866476239 Chr.5: 177091102-177091102 on Build GRCh38
rs139194967 Chr.5: 177090993-177090993 on Build GRCh38
rs758770208 Chr.5: 177091016-177091016 on Build GRCh38
rs996490269 Chr.5: 177090950-177090950 on Build GRCh38
rs772050296 Chr.5: 177091020-177091020 on Build GRCh38
rs754899280 Chr.5: 177091002-177091002 on Build GRCh38
rs777186373 Chr.5: 177091110-177091110 on Build GRCh38
rs770153156 Chr.5: 177091128-177091128 on Build GRCh38
rs549359769 Chr.5: 177091160-177091160 on Build GRCh38
rs771868306 Chr.5: 177090672-177090672 on Build GRCh38
rs773856157 Chr.5: 177091154-177091154 on Build GRCh38
rs566495475 Chr.5: 177091048-177091048 on Build GRCh38
rs146400837 Chr.5: 177091099-177091099 on Build GRCh38
rs999400793 Chr.5: 177090896-177090896 on Build GRCh38
rs368065553 Chr.5: 177090806-177090806 on Build GRCh38
rs2656909 Chr.5: 177090707-177090707 on Build GRCh38
rs776978786 Chr.5: 177091038-177091038 on Build GRCh38
rs768489149 Chr.5: 177090788-177090788 on Build GRCh38
rs45444898 Chr.5: 177090884-177090884 on Build GRCh38
rs762547569 Chr.5: 177091139-177091139 on Build GRCh38
rs761002255 Chr.5: 177090820-177090820 on Build GRCh38
rs778099107 Chr.5: 177090892-177090893 on Build GRCh38
rs746114840 Chr.5: 177091091-177091091 on Build GRCh38
rs747160711 Chr.5: 177091105-177091105 on Build GRCh38
rs1040661095 Chr.5: 177091176-177091176 on Build GRCh38
rs764514463 Chr.5: 177090843-177090843 on Build GRCh38
rs45566036 Chr.5: 177090892-177090892 on Build GRCh38
rs199792541 Chr.5: 177090799-177090799 on Build GRCh38
rs1029375431 Chr.5: 177090767-177090767 on Build GRCh38

View Full Product Details