Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125262634-125263220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPD52L1
Entrez Gene ID
7164
Gene Name
tumor protein D52-like 1
Gene Aliases
D53
UniGene
Hs.591347
Gene Chromosome Location
Chr.6: 125153483-125264407 on Build GRCh38
Transcripts
NM_001003395.2 NM_001003396.2 NM_001003397.2 NM_001292026.2 NM_001300994.2 NM_001318903.1 NM_001318907.1 NM_003287.3 XM_005267122.2 XM_006715557.3 XM_017011239.1 XM_017011240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934220 c.583C>T p.R195W Chr.6: 125262930-125262930 on Build GRCh38
4398491 c.489G>A p.T163T Chr.6: 125262836-125262836 on Build GRCh38
3947975 c.498C>T p.G166G Chr.6: 125262845-125262845 on Build GRCh38
1072895 c.537C>T p.L179L Chr.6: 125262884-125262884 on Build GRCh38
4586191 c.613T>G p.*205E Chr.6: 125262960-125262960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780555981 Chr.6: 125262850-125262850 on Build GRCh38
rs572304870 Chr.6: 125263008-125263008 on Build GRCh38
rs1039169752 Chr.6: 125262772-125262772 on Build GRCh38
rs772348702 Chr.6: 125262846-125262846 on Build GRCh38
rs987315468 Chr.6: 125263048-125263048 on Build GRCh38
rs1032167227 Chr.6: 125263088-125263088 on Build GRCh38
rs752179870 Chr.6: 125262933-125262933 on Build GRCh38
rs776837951 Chr.6: 125262873-125262873 on Build GRCh38
rs1020911053 Chr.6: 125262988-125262988 on Build GRCh38
rs893551647 Chr.6: 125263101-125263101 on Build GRCh38
rs527280358 Chr.6: 125262974-125262974 on Build GRCh38
rs140651035 Chr.6: 125262931-125262931 on Build GRCh38
rs759383678 Chr.6: 125262928-125262928 on Build GRCh38
rs573282938 Chr.6: 125262860-125262860 on Build GRCh38
rs76969470 Chr.6: 125262851-125262851 on Build GRCh38
rs943125352 Chr.6: 125262751-125262751 on Build GRCh38
rs751443494 Chr.6: 125262923-125262923 on Build GRCh38
rs3799721 Chr.6: 125262663-125262663 on Build GRCh38
rs534330998 Chr.6: 125262881-125262881 on Build GRCh38
rs762758855 Chr.6: 125262894-125262894 on Build GRCh38
rs372792388 Chr.6: 125262802-125262802 on Build GRCh38
rs267600791 Chr.6: 125262836-125262836 on Build GRCh38
rs901174653 Chr.6: 125263043-125263043 on Build GRCh38
rs1051543950 Chr.6: 125262700-125262700 on Build GRCh38
rs375008815 Chr.6: 125262934-125262934 on Build GRCh38
rs1009311798 Chr.6: 125263140-125263140 on Build GRCh38
rs970952795 Chr.6: 125262880-125262880 on Build GRCh38
rs368984698 Chr.6: 125262882-125262882 on Build GRCh38
rs1007377130 Chr.6: 125263087-125263087 on Build GRCh38
rs369222385 Chr.6: 125262973-125262973 on Build GRCh38
rs930364107 Chr.6: 125263166-125263166 on Build GRCh38
rs765112877 Chr.6: 125263163-125263163 on Build GRCh38
rs933197549 Chr.6: 125263047-125263047 on Build GRCh38
rs544892765 Chr.6: 125263034-125263034 on Build GRCh38
rs888925802 Chr.6: 125263076-125263076 on Build GRCh38
rs758723323 Chr.6: 125262951-125262951 on Build GRCh38
rs780277801 Chr.6: 125262962-125262962 on Build GRCh38
rs748202040 Chr.6: 125262994-125262994 on Build GRCh38
rs753448082 Chr.6: 125262943-125262943 on Build GRCh38
rs567319547 Chr.6: 125262715-125262715 on Build GRCh38
rs560626136 Chr.6: 125263162-125263162 on Build GRCh38
rs577516158 Chr.6: 125262967-125262967 on Build GRCh38
rs766438084 Chr.6: 125262765-125262765 on Build GRCh38
rs769276376 Chr.6: 125262872-125262872 on Build GRCh38
rs754452123 Chr.6: 125262819-125262819 on Build GRCh38
rs762698186 Chr.6: 125262811-125262811 on Build GRCh38
rs372302467 Chr.6: 125262818-125262818 on Build GRCh38
rs559154046 Chr.6: 125262930-125262930 on Build GRCh38
rs199925644 Chr.6: 125262835-125262835 on Build GRCh38
rs777362446 Chr.6: 125262942-125262942 on Build GRCh38
rs542359213 Chr.6: 125263052-125263052 on Build GRCh38
rs770018708 Chr.6: 125262995-125262995 on Build GRCh38
rs911676543 Chr.6: 125262725-125262725 on Build GRCh38
rs773508230 Chr.6: 125263007-125263007 on Build GRCh38
rs1030537874 Chr.6: 125262655-125262655 on Build GRCh38
rs780685234 Chr.6: 125262821-125262821 on Build GRCh38
rs371270632 Chr.6: 125262899-125262899 on Build GRCh38
rs773602194 Chr.6: 125262893-125262893 on Build GRCh38
rs920319914 Chr.6: 125263112-125263112 on Build GRCh38
rs150120709 Chr.6: 125262845-125262845 on Build GRCh38
rs765613135 Chr.6: 125262800-125262800 on Build GRCh38
rs776647749 Chr.6: 125262888-125262888 on Build GRCh38

View Full Product Details