Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158111180-158111687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERAC1
Entrez Gene ID
84947
Gene Name
serine active site containing 1
Gene Aliases
-
UniGene
Hs.154706
Gene Chromosome Location
Chr.6: 158109504-158168280 on Build GRCh38
Transcripts
NM_032861.3 XM_006715586.2 XM_011536196.2 XM_011536198.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3829540 c.1829-1G>C p.? Chr.6: 158111503-158111503 on Build GRCh38
5459235 c.1916G>A p.R639H Chr.6: 158111415-158111415 on Build GRCh38
3022722 c.1933C>T p.R645C Chr.6: 158111398-158111398 on Build GRCh38
3022723 c.1915C>T p.R639C Chr.6: 158111416-158111416 on Build GRCh38
6874883 c.1846C>T p.L616L Chr.6: 158111485-158111485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780572089 Chr.6: 158111485-158111485 on Build GRCh38
rs200957270 Chr.6: 158111455-158111455 on Build GRCh38
rs751022622 Chr.6: 158111473-158111473 on Build GRCh38
rs980871833 Chr.6: 158111572-158111572 on Build GRCh38
rs775296113 Chr.6: 158111397-158111397 on Build GRCh38
rs191103232 Chr.6: 158111223-158111223 on Build GRCh38
rs745780427 Chr.6: 158111492-158111492 on Build GRCh38
rs199625765 Chr.6: 158111350-158111350 on Build GRCh38
rs754579027 Chr.6: 158111256-158111256 on Build GRCh38
rs879891959 Chr.6: 158111621-158111627 on Build GRCh38
rs201854885 Chr.6: 158111314-158111314 on Build GRCh38
rs769568348 Chr.6: 158111385-158111385 on Build GRCh38
rs747591237 Chr.6: 158111548-158111549 on Build GRCh38
rs948176078 Chr.6: 158111525-158111525 on Build GRCh38
rs535894128 Chr.6: 158111290-158111290 on Build GRCh38
rs574735577 Chr.6: 158111274-158111274 on Build GRCh38
rs756734323 Chr.6: 158111475-158111475 on Build GRCh38
rs749119853 Chr.6: 158111543-158111543 on Build GRCh38
rs143393289 Chr.6: 158111441-158111441 on Build GRCh38
rs139553840 Chr.6: 158111426-158111426 on Build GRCh38
rs370877945 Chr.6: 158111364-158111364 on Build GRCh38
rs756555539 Chr.6: 158111325-158111325 on Build GRCh38
rs762019418 Chr.6: 158111418-158111418 on Build GRCh38
rs977187207 Chr.6: 158111355-158111355 on Build GRCh38
rs1014696952 Chr.6: 158111570-158111570 on Build GRCh38
rs543781063 Chr.6: 158111612-158111612 on Build GRCh38
rs767893233 Chr.6: 158111419-158111419 on Build GRCh38
rs1053692836 Chr.6: 158111254-158111254 on Build GRCh38
rs920180428 Chr.6: 158111273-158111273 on Build GRCh38
rs375333121 Chr.6: 158111340-158111340 on Build GRCh38
rs928004689 Chr.6: 158111647-158111647 on Build GRCh38
rs80342743 Chr.6: 158111459-158111459 on Build GRCh38
rs758771323 Chr.6: 158111432-158111432 on Build GRCh38
rs775037012 Chr.6: 158111279-158111279 on Build GRCh38
rs939501674 Chr.6: 158111648-158111648 on Build GRCh38
rs1027291513 Chr.6: 158111337-158111337 on Build GRCh38
rs754950981 Chr.6: 158111347-158111348 on Build GRCh38
rs764905483 Chr.6: 158111261-158111261 on Build GRCh38
rs750748340 Chr.6: 158111315-158111315 on Build GRCh38
rs114624250 Chr.6: 158111398-158111398 on Build GRCh38
rs768913919 Chr.6: 158111415-158111415 on Build GRCh38
rs745806731 Chr.6: 158111372-158111372 on Build GRCh38
rs771709906 Chr.6: 158111277-158111277 on Build GRCh38
rs977303405 Chr.6: 158111215-158111215 on Build GRCh38
rs371513597 Chr.6: 158111252-158111252 on Build GRCh38
rs970749028 Chr.6: 158111620-158111621 on Build GRCh38
rs570786949 Chr.6: 158111206-158111206 on Build GRCh38
rs575982148 Chr.6: 158111416-158111416 on Build GRCh38
rs890108087 Chr.6: 158111535-158111535 on Build GRCh38
rs776179182 Chr.6: 158111659-158111659 on Build GRCh38
rs754306187 Chr.6: 158111452-158111452 on Build GRCh38
rs757141911 Chr.6: 158111360-158111360 on Build GRCh38
rs370788378 Chr.6: 158111401-158111401 on Build GRCh38
rs200964065 Chr.6: 158111345-158111345 on Build GRCh38
rs924499658 Chr.6: 158111220-158111220 on Build GRCh38
rs1004623209 Chr.6: 158111228-158111228 on Build GRCh38
rs963090745 Chr.6: 158111575-158111575 on Build GRCh38
rs537130188 Chr.6: 158111260-158111260 on Build GRCh38
rs779801675 Chr.6: 158111529-158111529 on Build GRCh38
rs996081087 Chr.6: 158111576-158111576 on Build GRCh38
rs898039954 Chr.6: 158111291-158111291 on Build GRCh38
rs552627077 Chr.6: 158111250-158111250 on Build GRCh38
rs752327642 Chr.6: 158111341-158111341 on Build GRCh38
rs1027150704 Chr.6: 158111579-158111579 on Build GRCh38
rs1004528134 Chr.6: 158111555-158111555 on Build GRCh38
rs182817906 Chr.6: 158111342-158111342 on Build GRCh38
rs980548491 Chr.6: 158111292-158111292 on Build GRCh38
rs779005858 Chr.6: 158111367-158111368 on Build GRCh38
rs756069913 Chr.6: 158111500-158111500 on Build GRCh38

View Full Product Details