Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 33904528-33905016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPER
Entrez Gene ID
168667
Gene Name
BMP binding endothelial regulator
Gene Aliases
CRIM3, CV-2, CV2
UniGene
Hs.660998
Gene Chromosome Location
Chr.7: 33904911-34155872 on Build GRCh38
Transcripts
NM_133468.4

SNP Detail

SNP ID Chromosome Location
rs370275346 Chr.7: 33904694-33904694 on Build GRCh38
rs776868586 Chr.7: 33904574-33904574 on Build GRCh38
rs906400468 Chr.7: 33904726-33904726 on Build GRCh38
rs781546542 Chr.7: 33904792-33904792 on Build GRCh38
rs768791113 Chr.7: 33904570-33904570 on Build GRCh38
rs1015068427 Chr.7: 33904941-33904941 on Build GRCh38
rs973614386 Chr.7: 33904997-33904997 on Build GRCh38
rs144748548 Chr.7: 33904923-33904923 on Build GRCh38
rs990661686 Chr.7: 33904954-33904954 on Build GRCh38
rs759528478 Chr.7: 33904593-33904593 on Build GRCh38
rs996423458 Chr.7: 33904856-33904856 on Build GRCh38
rs907564610 Chr.7: 33904676-33904676 on Build GRCh38
rs1050904274 Chr.7: 33904795-33904795 on Build GRCh38
rs1046063040 Chr.7: 33904662-33904662 on Build GRCh38
rs114417992 Chr.7: 33904972-33904972 on Build GRCh38
rs566454069 Chr.7: 33904910-33904910 on Build GRCh38
rs974932153 Chr.7: 33904592-33904592 on Build GRCh38
rs113005525 Chr.7: 33904657-33904657 on Build GRCh38
rs1020222636 Chr.7: 33904832-33904832 on Build GRCh38
rs757508614 Chr.7: 33904665-33904666 on Build GRCh38
rs940358789 Chr.7: 33904996-33904996 on Build GRCh38
rs13224548 Chr.7: 33904867-33904867 on Build GRCh38
rs552998020 Chr.7: 33904786-33904786 on Build GRCh38
rs996364215 Chr.7: 33904739-33904739 on Build GRCh38
rs1010292694 Chr.7: 33904797-33904797 on Build GRCh38
rs1009433442 Chr.7: 33904830-33904830 on Build GRCh38
rs747304896 Chr.7: 33904547-33904547 on Build GRCh38
rs988025490 Chr.7: 33904932-33904932 on Build GRCh38
rs903220389 Chr.7: 33904841-33904841 on Build GRCh38
rs970485530 Chr.7: 33904959-33904959 on Build GRCh38
rs185493634 Chr.7: 33904618-33904618 on Build GRCh38
rs542270725 Chr.7: 33904925-33904925 on Build GRCh38
rs987215128 Chr.7: 33904623-33904623 on Build GRCh38
rs765740189 Chr.7: 33904587-33904587 on Build GRCh38
rs368333354 Chr.7: 33904779-33904779 on Build GRCh38
rs1057241420 Chr.7: 33904765-33904765 on Build GRCh38
rs890535469 Chr.7: 33904811-33904811 on Build GRCh38
rs569489652 Chr.7: 33904585-33904585 on Build GRCh38
rs962112735 Chr.7: 33904948-33904948 on Build GRCh38
rs939156279 Chr.7: 33904737-33904737 on Build GRCh38
rs916335148 Chr.7: 33904629-33904629 on Build GRCh38
rs899310191 Chr.7: 33904717-33904717 on Build GRCh38
rs1023929237 Chr.7: 33904956-33904956 on Build GRCh38
rs762181544 Chr.7: 33904586-33904586 on Build GRCh38
rs891895173 Chr.7: 33904789-33904789 on Build GRCh38
rs954872273 Chr.7: 33904606-33904606 on Build GRCh38
rs879539271 Chr.7: 33904978-33904978 on Build GRCh38
rs536280897 Chr.7: 33904708-33904708 on Build GRCh38
rs557412277 Chr.7: 33904949-33904949 on Build GRCh38
rs1037973641 Chr.7: 33904711-33904711 on Build GRCh38
rs907510189 Chr.7: 33904977-33904977 on Build GRCh38
rs920242393 Chr.7: 33904666-33904666 on Build GRCh38
rs558346470 Chr.7: 33904599-33904599 on Build GRCh38
rs764205179 Chr.7: 33904624-33904624 on Build GRCh38
rs968956460 Chr.7: 33904584-33904584 on Build GRCh38

View Full Product Details