Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167478666-167479256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR49
Entrez Gene ID
151790
Gene Name
WD repeat domain 49
Gene Aliases
-
UniGene
Hs.213762
Gene Chromosome Location
Chr.3: 167478684-167653501 on Build GRCh38
Transcripts
NM_178824.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6821923 c.2083A>C p.N695H Chr.3: 167478889-167478889 on Build GRCh38
1632961 c.2086A>G p.K696E Chr.3: 167478886-167478886 on Build GRCh38
1632962 c.1981A>G p.K661E Chr.3: 167478886-167478886 on Build GRCh38
3203271 c.2036G>A p.R679Q Chr.3: 167478936-167478936 on Build GRCh38
3203277 c.1997A>C p.E666A Chr.3: 167478975-167478975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542915661 Chr.3: 167479142-167479142 on Build GRCh38
rs747672557 Chr.3: 167478848-167478848 on Build GRCh38
rs142044741 Chr.3: 167478891-167478891 on Build GRCh38
rs751512910 Chr.3: 167478831-167478831 on Build GRCh38
rs746736143 Chr.3: 167478838-167478838 on Build GRCh38
rs751376132 Chr.3: 167478894-167478894 on Build GRCh38
rs776951263 Chr.3: 167479003-167479004 on Build GRCh38
rs761240339 Chr.3: 167478714-167478714 on Build GRCh38
rs375411249 Chr.3: 167478863-167478863 on Build GRCh38
rs145156716 Chr.3: 167478959-167478959 on Build GRCh38
rs759835741 Chr.3: 167478884-167478884 on Build GRCh38
rs539256562 Chr.3: 167479066-167479066 on Build GRCh38
rs918643254 Chr.3: 167479069-167479069 on Build GRCh38
rs774113932 Chr.3: 167478893-167478897 on Build GRCh38
rs771249100 Chr.3: 167478862-167478862 on Build GRCh38
rs73878718 Chr.3: 167478784-167478784 on Build GRCh38
rs111344697 Chr.3: 167479009-167479009 on Build GRCh38
rs1012564834 Chr.3: 167479144-167479144 on Build GRCh38
rs151139703 Chr.3: 167478907-167478907 on Build GRCh38
rs778966147 Chr.3: 167478887-167478887 on Build GRCh38
rs771741698 Chr.3: 167478907-167478907 on Build GRCh38
rs997315308 Chr.3: 167478918-167478918 on Build GRCh38
rs781333884 Chr.3: 167478904-167478904 on Build GRCh38
rs1026615791 Chr.3: 167479215-167479215 on Build GRCh38
rs770648647 Chr.3: 167478937-167478937 on Build GRCh38
rs575505305 Chr.3: 167479100-167479100 on Build GRCh38
rs112630566 Chr.3: 167478941-167478941 on Build GRCh38
rs199579482 Chr.3: 167479005-167479005 on Build GRCh38
rs770856492 Chr.3: 167478878-167478881 on Build GRCh38
rs769263978 Chr.3: 167479039-167479040 on Build GRCh38
rs750600341 Chr.3: 167479041-167479041 on Build GRCh38
rs946983186 Chr.3: 167479083-167479083 on Build GRCh38
rs754057980 Chr.3: 167478886-167478886 on Build GRCh38
rs1051805135 Chr.3: 167478731-167478731 on Build GRCh38
rs761736168 Chr.3: 167479025-167479025 on Build GRCh38
rs754765752 Chr.3: 167478900-167478900 on Build GRCh38
rs1055098448 Chr.3: 167479219-167479219 on Build GRCh38
rs182070065 Chr.3: 167479118-167479118 on Build GRCh38
rs545895979 Chr.3: 167479121-167479121 on Build GRCh38
rs1022858219 Chr.3: 167478949-167478949 on Build GRCh38
rs554901435 Chr.3: 167479153-167479153 on Build GRCh38
rs557148388 Chr.3: 167479086-167479086 on Build GRCh38
rs527438547 Chr.3: 167479047-167479047 on Build GRCh38
rs376125490 Chr.3: 167478839-167478839 on Build GRCh38
rs765512728 Chr.3: 167479036-167479036 on Build GRCh38
rs200634274 Chr.3: 167478789-167478793 on Build GRCh38
rs973678093 Chr.3: 167478982-167478982 on Build GRCh38
rs533197939 Chr.3: 167478852-167478852 on Build GRCh38
rs768015953 Chr.3: 167478868-167478868 on Build GRCh38
rs761148435 Chr.3: 167478984-167478984 on Build GRCh38
rs541129354 Chr.3: 167478811-167478811 on Build GRCh38
rs998615016 Chr.3: 167478881-167478881 on Build GRCh38
rs73878717 Chr.3: 167478730-167478730 on Build GRCh38
rs369114443 Chr.3: 167478872-167478872 on Build GRCh38
rs918780571 Chr.3: 167478745-167478745 on Build GRCh38
rs769412412 Chr.3: 167478850-167478850 on Build GRCh38
rs902685809 Chr.3: 167479105-167479105 on Build GRCh38
rs1041463948 Chr.3: 167478877-167478877 on Build GRCh38
rs772423470 Chr.3: 167478950-167478950 on Build GRCh38
rs201391497 Chr.3: 167478873-167478873 on Build GRCh38
rs937637101 Chr.3: 167479168-167479168 on Build GRCh38
rs759395835 Chr.3: 167478906-167478907 on Build GRCh38
rs953312344 Chr.3: 167478953-167478953 on Build GRCh38
rs1022577304 Chr.3: 167479145-167479145 on Build GRCh38
rs534799971 Chr.3: 167478943-167478943 on Build GRCh38
rs371672430 Chr.3: 167478880-167478880 on Build GRCh38
rs911951690 Chr.3: 167479012-167479012 on Build GRCh38
rs551677010 Chr.3: 167478865-167478865 on Build GRCh38
rs937005698 Chr.3: 167478819-167478819 on Build GRCh38
rs919363074 Chr.3: 167478718-167478718 on Build GRCh38
rs1056825653 Chr.3: 167479136-167479136 on Build GRCh38
rs777312643 Chr.3: 167478909-167478909 on Build GRCh38
rs545096297 Chr.3: 167478821-167478821 on Build GRCh38
rs994766888 Chr.3: 167479176-167479176 on Build GRCh38
rs1039036619 Chr.3: 167479062-167479062 on Build GRCh38
rs748734746 Chr.3: 167478864-167478864 on Build GRCh38
rs76736291 Chr.3: 167478936-167478936 on Build GRCh38

View Full Product Details