Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 22550410-22550994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIP4K2A
Entrez Gene ID
5305
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 alpha
Gene Aliases
PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK
UniGene
Hs.57079
Gene Chromosome Location
Chr.10: 22534837-22714574 on Build GRCh38
Transcripts
NM_005028.4 XM_006717450.2 XM_011519496.1 XM_017016330.1 XM_017016331.1 XM_017016332.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4643450 c.792G>T p.E264D Chr.10: 22550659-22550659 on Build GRCh38
3741605 c.718A>T p.N240Y Chr.10: 22550733-22550733 on Build GRCh38
3751689 c.752A>G p.N251S Chr.10: 22550699-22550699 on Build GRCh38
3867001 c.762C>T p.V254V Chr.10: 22550689-22550689 on Build GRCh38
4807895 c.739A>C p.I247L Chr.10: 22550712-22550712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189874652 Chr.10: 22550452-22550452 on Build GRCh38
rs367563584 Chr.10: 22550627-22550627 on Build GRCh38
rs779587709 Chr.10: 22550610-22550610 on Build GRCh38
rs751485369 Chr.10: 22550686-22550686 on Build GRCh38
rs919347058 Chr.10: 22550632-22550632 on Build GRCh38
rs543105128 Chr.10: 22550943-22550943 on Build GRCh38
rs902919757 Chr.10: 22550858-22550858 on Build GRCh38
rs761027794 Chr.10: 22550628-22550628 on Build GRCh38
rs374939921 Chr.10: 22550805-22550805 on Build GRCh38
rs1026846891 Chr.10: 22550964-22550964 on Build GRCh38
rs541786882 Chr.10: 22550907-22550907 on Build GRCh38
rs2230469 Chr.10: 22550699-22550699 on Build GRCh38
rs148391473 Chr.10: 22550709-22550709 on Build GRCh38
rs1012806499 Chr.10: 22550929-22550929 on Build GRCh38
rs1035634280 Chr.10: 22550464-22550464 on Build GRCh38
rs747231511 Chr.10: 22550643-22550644 on Build GRCh38
rs531364565 Chr.10: 22550571-22550571 on Build GRCh38
rs374481001 Chr.10: 22550542-22550542 on Build GRCh38
rs58255702 Chr.10: 22550543-22550543 on Build GRCh38
rs895918902 Chr.10: 22550455-22550455 on Build GRCh38
rs571793397 Chr.10: 22550547-22550547 on Build GRCh38
rs775509689 Chr.10: 22550812-22550812 on Build GRCh38
rs905864089 Chr.10: 22550468-22550468 on Build GRCh38
rs930333287 Chr.10: 22550661-22550661 on Build GRCh38
rs752810739 Chr.10: 22550652-22550652 on Build GRCh38
rs376344827 Chr.10: 22550637-22550637 on Build GRCh38
rs531121600 Chr.10: 22550551-22550551 on Build GRCh38
rs113575582 Chr.10: 22550838-22550838 on Build GRCh38
rs775615577 Chr.10: 22550879-22550879 on Build GRCh38
rs759762848 Chr.10: 22550643-22550643 on Build GRCh38
rs755258492 Chr.10: 22550525-22550525 on Build GRCh38
rs777861486 Chr.10: 22550804-22550804 on Build GRCh38
rs748649312 Chr.10: 22550612-22550612 on Build GRCh38
rs181485100 Chr.10: 22550874-22550874 on Build GRCh38
rs776637685 Chr.10: 22550958-22550958 on Build GRCh38
rs373216893 Chr.10: 22550672-22550672 on Build GRCh38
rs185941375 Chr.10: 22550698-22550698 on Build GRCh38
rs369310691 Chr.10: 22550645-22550645 on Build GRCh38
rs147575486 Chr.10: 22550664-22550664 on Build GRCh38
rs771230836 Chr.10: 22550778-22550778 on Build GRCh38
rs575315179 Chr.10: 22550461-22550461 on Build GRCh38
rs779485845 Chr.10: 22550716-22550716 on Build GRCh38
rs765271244 Chr.10: 22550650-22550650 on Build GRCh38
rs549641449 Chr.10: 22550566-22550566 on Build GRCh38
rs768083736 Chr.10: 22550732-22550732 on Build GRCh38
rs748343687 Chr.10: 22550614-22550614 on Build GRCh38
rs1000217022 Chr.10: 22550930-22550930 on Build GRCh38
rs892074948 Chr.10: 22550948-22550948 on Build GRCh38
rs1012773491 Chr.10: 22550494-22550494 on Build GRCh38
rs141028548 Chr.10: 22550481-22550481 on Build GRCh38
rs115001136 Chr.10: 22550431-22550431 on Build GRCh38
rs201809681 Chr.10: 22550819-22550820 on Build GRCh38
rs763052531 Chr.10: 22550655-22550655 on Build GRCh38
rs577797161 Chr.10: 22550841-22550841 on Build GRCh38
rs181087198 Chr.10: 22550633-22550633 on Build GRCh38
rs543581761 Chr.10: 22550873-22550873 on Build GRCh38
rs1041878955 Chr.10: 22550843-22550844 on Build GRCh38
rs1044453522 Chr.10: 22550906-22550906 on Build GRCh38
rs369574294 Chr.10: 22550634-22550634 on Build GRCh38
rs748789808 Chr.10: 22550727-22550727 on Build GRCh38
rs10047326 Chr.10: 22550534-22550534 on Build GRCh38
rs563790047 Chr.10: 22550447-22550447 on Build GRCh38
rs539428684 Chr.10: 22550520-22550520 on Build GRCh38
rs902886487 Chr.10: 22550459-22550459 on Build GRCh38
rs750085723 Chr.10: 22550706-22550706 on Build GRCh38
rs965808217 Chr.10: 22550594-22550594 on Build GRCh38
rs777020636 Chr.10: 22550794-22550794 on Build GRCh38
rs1022946609 Chr.10: 22550467-22550467 on Build GRCh38
rs796283275 Chr.10: 22550901-22550901 on Build GRCh38
rs747465159 Chr.10: 22550769-22550769 on Build GRCh38
rs778262322 Chr.10: 22550754-22550754 on Build GRCh38
rs2230469 Chr.10: 22550699-22550699 on Build GRCh38
rs773630344 Chr.10: 22550623-22550623 on Build GRCh38
rs770052021 Chr.10: 22550807-22550807 on Build GRCh38

View Full Product Details