Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60921436-60921966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM109
Entrez Gene ID
79073
Gene Name
transmembrane protein 109
Gene Aliases
-
UniGene
Hs.13662
Gene Chromosome Location
Chr.11: 60913899-60923443 on Build GRCh38
Transcripts
NM_024092.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2036636 c.352G>A p.A118T Chr.11: 60921785-60921785 on Build GRCh38
3869672 c.393C>T p.F131F Chr.11: 60921826-60921826 on Build GRCh38
6552165 c.447C>T p.L149L Chr.11: 60921880-60921880 on Build GRCh38
4574535 c.521delC p.L175fs*1 Chr.11: 60921954-60921954 on Build GRCh38
7256168 c.437C>T p.S146F Chr.11: 60921870-60921870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749838629 Chr.11: 60921928-60921928 on Build GRCh38
rs903718687 Chr.11: 60921863-60921863 on Build GRCh38
rs770302461 Chr.11: 60921815-60921815 on Build GRCh38
rs1002841736 Chr.11: 60921641-60921641 on Build GRCh38
rs199887708 Chr.11: 60921854-60921854 on Build GRCh38
rs752335637 Chr.11: 60921749-60921749 on Build GRCh38
rs945171811 Chr.11: 60921839-60921839 on Build GRCh38
rs34881294 Chr.11: 60921848-60921848 on Build GRCh38
rs779606958 Chr.11: 60921811-60921811 on Build GRCh38
rs1019394532 Chr.11: 60921662-60921662 on Build GRCh38
rs536170140 Chr.11: 60921784-60921784 on Build GRCh38
rs896297995 Chr.11: 60921481-60921481 on Build GRCh38
rs919484947 Chr.11: 60921485-60921485 on Build GRCh38
rs919465857 Chr.11: 60921678-60921678 on Build GRCh38
rs147978277 Chr.11: 60921943-60921943 on Build GRCh38
rs190543238 Chr.11: 60921672-60921672 on Build GRCh38
rs774594944 Chr.11: 60921728-60921728 on Build GRCh38
rs530772093 Chr.11: 60921613-60921613 on Build GRCh38
rs758015533 Chr.11: 60921794-60921794 on Build GRCh38
rs757035036 Chr.11: 60921781-60921781 on Build GRCh38
rs771374233 Chr.11: 60921843-60921843 on Build GRCh38
rs953873881 Chr.11: 60921621-60921621 on Build GRCh38
rs753579365 Chr.11: 60921774-60921774 on Build GRCh38
rs778279542 Chr.11: 60921826-60921826 on Build GRCh38
rs907200000 Chr.11: 60921599-60921599 on Build GRCh38
rs200374043 Chr.11: 60921756-60921756 on Build GRCh38
rs536638844 Chr.11: 60921858-60921858 on Build GRCh38
rs764065933 Chr.11: 60921769-60921769 on Build GRCh38
rs375042502 Chr.11: 60921875-60921875 on Build GRCh38
rs148897091 Chr.11: 60921874-60921874 on Build GRCh38
rs760748768 Chr.11: 60921760-60921760 on Build GRCh38
rs143462464 Chr.11: 60921934-60921934 on Build GRCh38
rs201721653 Chr.11: 60921944-60921944 on Build GRCh38
rs747248259 Chr.11: 60921645-60921645 on Build GRCh38
rs929494426 Chr.11: 60921502-60921502 on Build GRCh38
rs943696609 Chr.11: 60921470-60921470 on Build GRCh38
rs765962512 Chr.11: 60921906-60921906 on Build GRCh38
rs185779880 Chr.11: 60921624-60921624 on Build GRCh38
rs761635924 Chr.11: 60921880-60921880 on Build GRCh38
rs368906004 Chr.11: 60921881-60921881 on Build GRCh38
rs547201895 Chr.11: 60921717-60921717 on Build GRCh38
rs750316636 Chr.11: 60921474-60921474 on Build GRCh38
rs758105259 Chr.11: 60921905-60921905 on Build GRCh38
rs149621483 Chr.11: 60921663-60921663 on Build GRCh38
rs759653504 Chr.11: 60921731-60921731 on Build GRCh38
rs952330236 Chr.11: 60921716-60921716 on Build GRCh38
rs1027980364 Chr.11: 60921609-60921609 on Build GRCh38
rs776580158 Chr.11: 60921879-60921879 on Build GRCh38
rs750181700 Chr.11: 60921882-60921882 on Build GRCh38
rs1044473672 Chr.11: 60921588-60921588 on Build GRCh38
rs1007963293 Chr.11: 60921657-60921657 on Build GRCh38
rs751077165 Chr.11: 60921913-60921913 on Build GRCh38
rs753100422 Chr.11: 60921550-60921550 on Build GRCh38
rs7951857 Chr.11: 60921524-60921524 on Build GRCh38
rs138262235 Chr.11: 60921553-60921553 on Build GRCh38
rs199565578 Chr.11: 60921832-60921832 on Build GRCh38
rs746512873 Chr.11: 60921812-60921812 on Build GRCh38
rs746173819 Chr.11: 60921940-60921940 on Build GRCh38
rs757697622 Chr.11: 60921931-60921931 on Build GRCh38
rs1042250222 Chr.11: 60921547-60921547 on Build GRCh38
rs774546009 Chr.11: 60921845-60921845 on Build GRCh38
rs866063054 Chr.11: 60921942-60921942 on Build GRCh38
rs746000733 Chr.11: 60921785-60921785 on Build GRCh38
rs754447668 Chr.11: 60921920-60921920 on Build GRCh38
rs778168545 Chr.11: 60921926-60921926 on Build GRCh38

View Full Product Details