Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10629440-10629999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH3
Entrez Gene ID
4621
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
Gene Aliases
DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE
UniGene
Hs.440895
Gene Chromosome Location
Chr.17: 10628526-10679657 on Build GRCh38
Transcripts
NM_002470.3 XM_011523870.2 XM_011523871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3514002 c.5749C>T p.Q1917* Chr.17: 10629644-10629644 on Build GRCh38
7127214 c.5772G>T p.K1924N Chr.17: 10629621-10629621 on Build GRCh38
3889265 c.5739C>T p.I1913I Chr.17: 10629654-10629654 on Build GRCh38
3514001 c.5789C>T p.S1930F Chr.17: 10629604-10629604 on Build GRCh38
6415839 c.5792G>A p.S1931N Chr.17: 10629601-10629601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199619639 Chr.17: 10629585-10629585 on Build GRCh38
rs761915974 Chr.17: 10629769-10629769 on Build GRCh38
rs778100400 Chr.17: 10629959-10629959 on Build GRCh38
rs770016592 Chr.17: 10629956-10629956 on Build GRCh38
rs761623039 Chr.17: 10629695-10629695 on Build GRCh38
rs201452885 Chr.17: 10629771-10629771 on Build GRCh38
rs769456541 Chr.17: 10629566-10629567 on Build GRCh38
rs368978437 Chr.17: 10629834-10629834 on Build GRCh38
rs758392734 Chr.17: 10629703-10629703 on Build GRCh38
rs749488693 Chr.17: 10629976-10629976 on Build GRCh38
rs775534998 Chr.17: 10629672-10629672 on Build GRCh38
rs878873684 Chr.17: 10629615-10629615 on Build GRCh38
rs549327504 Chr.17: 10629886-10629886 on Build GRCh38
rs779685889 Chr.17: 10629597-10629597 on Build GRCh38
rs759371633 Chr.17: 10629879-10629879 on Build GRCh38
rs150103893 Chr.17: 10629464-10629464 on Build GRCh38
rs199803615 Chr.17: 10629746-10629746 on Build GRCh38
rs537972961 Chr.17: 10629513-10629513 on Build GRCh38
rs766391547 Chr.17: 10629515-10629515 on Build GRCh38
rs754458917 Chr.17: 10629718-10629718 on Build GRCh38
rs537407236 Chr.17: 10629566-10629566 on Build GRCh38
rs549573582 Chr.17: 10629565-10629566 on Build GRCh38
rs138477550 Chr.17: 10629793-10629793 on Build GRCh38
rs370760565 Chr.17: 10629762-10629762 on Build GRCh38
rs1027765435 Chr.17: 10629484-10629484 on Build GRCh38
rs781722886 Chr.17: 10629929-10629929 on Build GRCh38
rs765090438 Chr.17: 10629698-10629698 on Build GRCh38
rs778783927 Chr.17: 10629557-10629557 on Build GRCh38
rs375046787 Chr.17: 10629806-10629806 on Build GRCh38
rs1040636980 Chr.17: 10629577-10629577 on Build GRCh38
rs952913518 Chr.17: 10629506-10629506 on Build GRCh38
rs556678852 Chr.17: 10629514-10629514 on Build GRCh38
rs527661230 Chr.17: 10629817-10629817 on Build GRCh38
rs187637244 Chr.17: 10629572-10629572 on Build GRCh38
rs771722680 Chr.17: 10629559-10629559 on Build GRCh38
rs1048702846 Chr.17: 10629673-10629673 on Build GRCh38
rs748859267 Chr.17: 10629582-10629582 on Build GRCh38
rs777469983 Chr.17: 10629810-10629810 on Build GRCh38
rs753515010 Chr.17: 10629893-10629893 on Build GRCh38
rs774239787 Chr.17: 10629864-10629864 on Build GRCh38
rs775588285 Chr.17: 10629570-10629570 on Build GRCh38
rs758726114 Chr.17: 10629902-10629902 on Build GRCh38
rs762980743 Chr.17: 10629804-10629805 on Build GRCh38
rs1011279441 Chr.17: 10629654-10629654 on Build GRCh38
rs147304568 Chr.17: 10629733-10629733 on Build GRCh38
rs749608195 Chr.17: 10629642-10629642 on Build GRCh38
rs973494741 Chr.17: 10629850-10629850 on Build GRCh38
rs775984807 Chr.17: 10629760-10629760 on Build GRCh38
rs533007781 Chr.17: 10629490-10629490 on Build GRCh38
rs773118635 Chr.17: 10629742-10629743 on Build GRCh38
rs776875348 Chr.17: 10629765-10629765 on Build GRCh38
rs781043150 Chr.17: 10629605-10629605 on Build GRCh38
rs1005826926 Chr.17: 10629531-10629531 on Build GRCh38
rs750723128 Chr.17: 10629811-10629811 on Build GRCh38
rs562465234 Chr.17: 10629479-10629479 on Build GRCh38
rs745315197 Chr.17: 10629558-10629558 on Build GRCh38
rs768744561 Chr.17: 10629564-10629565 on Build GRCh38
rs765210931 Chr.17: 10629580-10629580 on Build GRCh38
rs755432897 Chr.17: 10629925-10629925 on Build GRCh38
rs780712811 Chr.17: 10629618-10629618 on Build GRCh38
rs199982682 Chr.17: 10629911-10629911 on Build GRCh38
rs796496261 Chr.17: 10629565-10629566 on Build GRCh38
rs780846542 Chr.17: 10629719-10629719 on Build GRCh38
rs891566988 Chr.17: 10629596-10629596 on Build GRCh38
rs369516937 Chr.17: 10629574-10629574 on Build GRCh38
rs200888700 Chr.17: 10629887-10629887 on Build GRCh38
rs147333978 Chr.17: 10629565-10629565 on Build GRCh38
rs370355816 Chr.17: 10629782-10629782 on Build GRCh38
rs942089350 Chr.17: 10629472-10629472 on Build GRCh38
rs368693549 Chr.17: 10629805-10629805 on Build GRCh38
rs200347390 Chr.17: 10629591-10629591 on Build GRCh38
rs79747866 Chr.17: 10629728-10629728 on Build GRCh38
rs748323203 Chr.17: 10629848-10629848 on Build GRCh38
rs374929480 Chr.17: 10629833-10629833 on Build GRCh38
rs571875801 Chr.17: 10629537-10629537 on Build GRCh38
rs762786602 Chr.17: 10629853-10629853 on Build GRCh38
rs772563939 Chr.17: 10629752-10629752 on Build GRCh38
rs190434808 Chr.17: 10629565-10629565 on Build GRCh38
rs748177595 Chr.17: 10629932-10629932 on Build GRCh38
rs768643300 Chr.17: 10629764-10629764 on Build GRCh38
rs570950857 Chr.17: 10629568-10629568 on Build GRCh38
rs751472661 Chr.17: 10629797-10629797 on Build GRCh38
rs764314073 Chr.17: 10629681-10629681 on Build GRCh38
rs747504488 Chr.17: 10629818-10629818 on Build GRCh38
rs12940161 Chr.17: 10629567-10629567 on Build GRCh38
rs770925491 Chr.17: 10629974-10629974 on Build GRCh38
rs145624953 Chr.17: 10629716-10629716 on Build GRCh38
rs867200333 Chr.17: 10629607-10629607 on Build GRCh38
rs771220671 Chr.17: 10629658-10629658 on Build GRCh38
rs371770372 Chr.17: 10629807-10629807 on Build GRCh38
rs774802588 Chr.17: 10629664-10629664 on Build GRCh38
rs746571273 Chr.17: 10629601-10629601 on Build GRCh38
rs760737851 Chr.17: 10629678-10629678 on Build GRCh38
rs746345098 Chr.17: 10629665-10629665 on Build GRCh38
rs369612990 Chr.17: 10629671-10629671 on Build GRCh38
rs770880099 Chr.17: 10629858-10629858 on Build GRCh38
rs916629361 Chr.17: 10629855-10629855 on Build GRCh38
rs752145848 Chr.17: 10629884-10629884 on Build GRCh38
rs886052578 Chr.17: 10629724-10629724 on Build GRCh38
rs776734948 Chr.17: 10629684-10629684 on Build GRCh38
rs983492513 Chr.17: 10629829-10629829 on Build GRCh38
rs764281649 Chr.17: 10629573-10629573 on Build GRCh38
rs371260049 Chr.17: 10629851-10629851 on Build GRCh38
rs770070846 Chr.17: 10629852-10629852 on Build GRCh38
rs766360657 Chr.17: 10629794-10629794 on Build GRCh38
rs767550622 Chr.17: 10629880-10629880 on Build GRCh38
rs766486500 Chr.17: 10629710-10629710 on Build GRCh38
rs961662614 Chr.17: 10629761-10629761 on Build GRCh38
rs750289236 Chr.17: 10629701-10629701 on Build GRCh38

View Full Product Details