Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81301365-81301932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NR_034121.1 NR_034122.1 NR_034123.1 NM_001131035.1 NM_001131036.1 NM_032280.2 XM_005248616.3

Gene Symbol
ZCCHC9
Entrez Gene ID
84240
Gene Name
zinc finger CCHC-type containing 9
Gene Aliases
PPP1R41
UniGene
Hs.15536
Gene Chromosome Location
Chr.5: 81301583-81313147 on Build GRCh38
Transcripts
NR_034121.1 NR_034122.1 NR_034123.1 NM_001131035.1 NM_001131036.1 NM_032280.2 XM_005248616.3

SNP Detail

SNP ID Chromosome Location
rs2291942 Chr.5: 81301544-81301544 on Build GRCh38
rs916879234 Chr.5: 81301828-81301828 on Build GRCh38
rs974434214 Chr.5: 81301456-81301456 on Build GRCh38
rs540929802 Chr.5: 81301723-81301723 on Build GRCh38
rs936800609 Chr.5: 81301889-81301889 on Build GRCh38
rs188855817 Chr.5: 81301396-81301396 on Build GRCh38
rs975695537 Chr.5: 81301579-81301579 on Build GRCh38
rs1016124419 Chr.5: 81301718-81301718 on Build GRCh38
rs1004949882 Chr.5: 81301707-81301707 on Build GRCh38
rs945809693 Chr.5: 81301637-81301637 on Build GRCh38
rs948599780 Chr.5: 81301652-81301652 on Build GRCh38
rs1000015022 Chr.5: 81301671-81301671 on Build GRCh38
rs544628820 Chr.5: 81301512-81301512 on Build GRCh38
rs921400189 Chr.5: 81301582-81301582 on Build GRCh38
rs760266812 Chr.5: 81301631-81301631 on Build GRCh38
rs576596060 Chr.5: 81301673-81301673 on Build GRCh38
rs575103674 Chr.5: 81301566-81301566 on Build GRCh38
rs1023196046 Chr.5: 81301546-81301546 on Build GRCh38
rs988179749 Chr.5: 81301468-81301468 on Build GRCh38
rs987577758 Chr.5: 81301812-81301812 on Build GRCh38
rs116142117 Chr.5: 81301486-81301486 on Build GRCh38
rs1055266871 Chr.5: 81301654-81301654 on Build GRCh38
rs562880039 Chr.5: 81301395-81301395 on Build GRCh38
rs904150059 Chr.5: 81301505-81301505 on Build GRCh38
rs527571500 Chr.5: 81301482-81301482 on Build GRCh38
rs921581187 Chr.5: 81301457-81301457 on Build GRCh38
rs936936130 Chr.5: 81301645-81301645 on Build GRCh38
rs10036022 Chr.5: 81301451-81301451 on Build GRCh38
rs569444490 Chr.5: 81301832-81301832 on Build GRCh38
rs969848531 Chr.5: 81301444-81301444 on Build GRCh38
rs983390924 Chr.5: 81301409-81301409 on Build GRCh38
rs570546471 Chr.5: 81301502-81301502 on Build GRCh38
rs185202728 Chr.5: 81301689-81301689 on Build GRCh38
rs548520472 Chr.5: 81301431-81301431 on Build GRCh38
rs564311950 Chr.5: 81301759-81301759 on Build GRCh38
rs968582156 Chr.5: 81301552-81301552 on Build GRCh38
rs534221627 Chr.5: 81301588-81301588 on Build GRCh38
rs1000472780 Chr.5: 81301553-81301553 on Build GRCh38
rs1046385359 Chr.5: 81301687-81301687 on Build GRCh38
rs1031951009 Chr.5: 81301681-81301681 on Build GRCh38
rs765883118 Chr.5: 81301632-81301632 on Build GRCh38
rs1028875625 Chr.5: 81301739-81301739 on Build GRCh38
rs984290792 Chr.5: 81301791-81301791 on Build GRCh38
rs753337802 Chr.5: 81301640-81301640 on Build GRCh38
rs193039358 Chr.5: 81301452-81301452 on Build GRCh38
rs886891415 Chr.5: 81301511-81301511 on Build GRCh38
rs73768060 Chr.5: 81301642-81301642 on Build GRCh38
rs547617933 Chr.5: 81301488-81301488 on Build GRCh38
rs950827705 Chr.5: 81301554-81301554 on Build GRCh38
rs11955364 Chr.5: 81301545-81301545 on Build GRCh38
rs939750173 Chr.5: 81301804-81301804 on Build GRCh38
rs963314781 Chr.5: 81301584-81301584 on Build GRCh38
rs953098043 Chr.5: 81301765-81301765 on Build GRCh38
rs577472775 Chr.5: 81301612-81301612 on Build GRCh38
rs117879710 Chr.5: 81301695-81301695 on Build GRCh38
rs555045228 Chr.5: 81301558-81301558 on Build GRCh38
rs765227440 Chr.5: 81301664-81301664 on Build GRCh38
rs895148587 Chr.5: 81301659-81301659 on Build GRCh38
rs974635559 Chr.5: 81301608-81301608 on Build GRCh38
rs990944570 Chr.5: 81301905-81301905 on Build GRCh38
rs113815796 Chr.5: 81301410-81301410 on Build GRCh38
rs912582839 Chr.5: 81301569-81301569 on Build GRCh38
rs758134034 Chr.5: 81301880-81301880 on Build GRCh38
rs895120097 Chr.5: 81301518-81301518 on Build GRCh38
rs530125187 Chr.5: 81301854-81301855 on Build GRCh38
rs371498015 Chr.5: 81301838-81301838 on Build GRCh38
rs1056746125 Chr.5: 81301528-81301528 on Build GRCh38
rs554384738 Chr.5: 81301589-81301589 on Build GRCh38
rs561575820 Chr.5: 81301650-81301650 on Build GRCh38
rs944302918 Chr.5: 81301473-81301473 on Build GRCh38
rs996103484 Chr.5: 81301736-81301736 on Build GRCh38
rs549477482 Chr.5: 81301814-81301814 on Build GRCh38
rs796566218 Chr.5: 81301664-81301665 on Build GRCh38
rs540040589 Chr.5: 81301462-81301462 on Build GRCh38
rs386689530 Chr.5: 81301544-81301545 on Build GRCh38
rs954492708 Chr.5: 81301626-81301626 on Build GRCh38
rs1016090394 Chr.5: 81301578-81301578 on Build GRCh38
rs539158637 Chr.5: 81301503-81301503 on Build GRCh38
rs975851883 Chr.5: 81301719-81301719 on Build GRCh38
rs868266759 Chr.5: 81301839-81301839 on Build GRCh38
rs867838967 Chr.5: 81301716-81301716 on Build GRCh38
rs62365038 Chr.5: 81301555-81301555 on Build GRCh38
rs1001052534 Chr.5: 81301508-81301508 on Build GRCh38
rs532936998 Chr.5: 81301775-81301775 on Build GRCh38
rs978451206 Chr.5: 81301871-81301871 on Build GRCh38
rs954649754 Chr.5: 81301797-81301797 on Build GRCh38
rs1051923712 Chr.5: 81301472-81301472 on Build GRCh38
rs562936848 Chr.5: 81301667-81301667 on Build GRCh38
rs780025982 Chr.5: 81301446-81301446 on Build GRCh38
rs375984392 Chr.5: 81301545-81301545 on Build GRCh38

View Full Product Details