Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157735679-157736179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THG1L
Entrez Gene ID
54974
Gene Name
tRNA-histidine guanylyltransferase 1 like
Gene Aliases
ICF45, IHG-1, hTHG1
UniGene
Hs.353090
Gene Chromosome Location
Chr.5: 157731389-157741449 on Build GRCh38
Transcripts
NM_001317824.1 NM_001317825.1 NM_001317826.1 NM_017872.4 XM_017009603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3768269 c.604G>A p.V202I Chr.5: 157735911-157735911 on Build GRCh38
4840972 c.569T>C p.F190S Chr.5: 157735876-157735876 on Build GRCh38
1065069 c.570C>A p.F190L Chr.5: 157735877-157735877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775141934 Chr.5: 157735892-157735892 on Build GRCh38
rs112609310 Chr.5: 157735720-157735720 on Build GRCh38
rs983664331 Chr.5: 157735741-157735741 on Build GRCh38
rs757117660 Chr.5: 157735921-157735921 on Build GRCh38
rs998176879 Chr.5: 157736139-157736139 on Build GRCh38
rs771517620 Chr.5: 157735858-157735858 on Build GRCh38
rs149240662 Chr.5: 157735828-157735828 on Build GRCh38
rs759541363 Chr.5: 157735847-157735858 on Build GRCh38
rs776256094 Chr.5: 157735986-157735986 on Build GRCh38
rs111235342 Chr.5: 157735782-157735782 on Build GRCh38
rs1037299521 Chr.5: 157735851-157735851 on Build GRCh38
rs763852853 Chr.5: 157735980-157735980 on Build GRCh38
rs765257248 Chr.5: 157735924-157735924 on Build GRCh38
rs1042259041 Chr.5: 157735909-157735909 on Build GRCh38
rs767146830 Chr.5: 157735891-157735891 on Build GRCh38
rs768964844 Chr.5: 157735816-157735816 on Build GRCh38
rs769986506 Chr.5: 157735914-157735915 on Build GRCh38
rs758461118 Chr.5: 157735804-157735804 on Build GRCh38
rs563228024 Chr.5: 157735732-157735732 on Build GRCh38
rs975412371 Chr.5: 157735711-157735711 on Build GRCh38
rs112092561 Chr.5: 157735953-157735953 on Build GRCh38
rs566072255 Chr.5: 157735799-157735799 on Build GRCh38
rs976795449 Chr.5: 157735761-157735761 on Build GRCh38
rs748385893 Chr.5: 157735947-157735947 on Build GRCh38
rs146172278 Chr.5: 157735956-157735956 on Build GRCh38
rs369299694 Chr.5: 157735868-157735868 on Build GRCh38
rs377093336 Chr.5: 157735979-157735979 on Build GRCh38
rs532627483 Chr.5: 157735846-157735846 on Build GRCh38
rs770067739 Chr.5: 157735836-157735836 on Build GRCh38
rs745326323 Chr.5: 157735961-157735961 on Build GRCh38
rs1018867963 Chr.5: 157735997-157735998 on Build GRCh38
rs376892496 Chr.5: 157735839-157735841 on Build GRCh38
rs777978143 Chr.5: 157735951-157735951 on Build GRCh38
rs369630234 Chr.5: 157735938-157735938 on Build GRCh38
rs374927795 Chr.5: 157735923-157735923 on Build GRCh38
rs536956126 Chr.5: 157735960-157735960 on Build GRCh38
rs776498541 Chr.5: 157735836-157735838 on Build GRCh38
rs1055472197 Chr.5: 157735839-157735839 on Build GRCh38
rs781627342 Chr.5: 157735821-157735821 on Build GRCh38
rs374652836 Chr.5: 157735887-157735887 on Build GRCh38
rs781507428 Chr.5: 157735943-157735943 on Build GRCh38
rs763392730 Chr.5: 157735857-157735857 on Build GRCh38
rs984105393 Chr.5: 157736036-157736036 on Build GRCh38
rs747131325 Chr.5: 157735815-157735815 on Build GRCh38
rs571436596 Chr.5: 157735864-157735864 on Build GRCh38
rs756453303 Chr.5: 157735948-157735948 on Build GRCh38
rs937633490 Chr.5: 157735833-157735833 on Build GRCh38
rs372975288 Chr.5: 157735888-157735888 on Build GRCh38
rs149783002 Chr.5: 157735850-157735850 on Build GRCh38
rs753756998 Chr.5: 157735920-157735920 on Build GRCh38
rs910072166 Chr.5: 157735841-157735841 on Build GRCh38
rs763823386 Chr.5: 157735914-157735914 on Build GRCh38
rs145864613 Chr.5: 157735928-157735928 on Build GRCh38
rs760455860 Chr.5: 157735904-157735904 on Build GRCh38
rs952361565 Chr.5: 157736030-157736030 on Build GRCh38
rs375967266 Chr.5: 157735977-157735977 on Build GRCh38
rs906157772 Chr.5: 157735867-157735867 on Build GRCh38
rs572897443 Chr.5: 157736025-157736025 on Build GRCh38
rs1032063227 Chr.5: 157735721-157735721 on Build GRCh38
rs549059105 Chr.5: 157735765-157735765 on Build GRCh38
rs965988950 Chr.5: 157736041-157736041 on Build GRCh38
rs780017951 Chr.5: 157735811-157735811 on Build GRCh38
rs372623363 Chr.5: 157735978-157735978 on Build GRCh38

View Full Product Details