Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75947158-75947724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG1
Entrez Gene ID
3617
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
GP147, IPM150, SPACR, VMD4
UniGene
Hs.590893
Gene Chromosome Location
Chr.6: 75921114-76072678 on Build GRCh38
Transcripts
NM_001282368.1 NM_001563.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471389 c.1923T>A p.F641L Chr.6: 75947435-75947435 on Build GRCh38
1634940 c.1975G>T p.D659Y Chr.6: 75947383-75947383 on Build GRCh38
3875627 c.2004C>T p.L668L Chr.6: 75947354-75947354 on Build GRCh38
1644190 c.1975G>A p.D659N Chr.6: 75947383-75947383 on Build GRCh38
6488639 c.2041C>A p.P681T Chr.6: 75947317-75947317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762387361 Chr.6: 75947393-75947393 on Build GRCh38
rs775456856 Chr.6: 75947281-75947281 on Build GRCh38
rs145955265 Chr.6: 75947352-75947352 on Build GRCh38
rs773080033 Chr.6: 75947441-75947441 on Build GRCh38
rs148615908 Chr.6: 75947399-75947399 on Build GRCh38
rs951586874 Chr.6: 75947596-75947596 on Build GRCh38
rs530621330 Chr.6: 75947616-75947616 on Build GRCh38
rs376390542 Chr.6: 75947509-75947509 on Build GRCh38
rs905256579 Chr.6: 75947571-75947571 on Build GRCh38
rs999467256 Chr.6: 75947238-75947238 on Build GRCh38
rs964455329 Chr.6: 75947257-75947257 on Build GRCh38
rs1037821641 Chr.6: 75947451-75947451 on Build GRCh38
rs746872890 Chr.6: 75947466-75947474 on Build GRCh38
rs147614663 Chr.6: 75947522-75947522 on Build GRCh38
rs201823141 Chr.6: 75947310-75947310 on Build GRCh38
rs145267469 Chr.6: 75947468-75947468 on Build GRCh38
rs371118175 Chr.6: 75947517-75947517 on Build GRCh38
rs779171435 Chr.6: 75947513-75947513 on Build GRCh38
rs987255527 Chr.6: 75947695-75947695 on Build GRCh38
rs186188993 Chr.6: 75947632-75947632 on Build GRCh38
rs777740210 Chr.6: 75947337-75947340 on Build GRCh38
rs190568085 Chr.6: 75947659-75947659 on Build GRCh38
rs1055154364 Chr.6: 75947559-75947559 on Build GRCh38
rs764710111 Chr.6: 75947489-75947489 on Build GRCh38
rs757203031 Chr.6: 75947298-75947298 on Build GRCh38
rs181256434 Chr.6: 75947497-75947497 on Build GRCh38
rs202200012 Chr.6: 75947418-75947418 on Build GRCh38
rs371197189 Chr.6: 75947408-75947408 on Build GRCh38
rs371030379 Chr.6: 75947619-75947619 on Build GRCh38
rs185850526 Chr.6: 75947646-75947646 on Build GRCh38
rs376165008 Chr.6: 75947427-75947427 on Build GRCh38
rs760230012 Chr.6: 75947573-75947573 on Build GRCh38
rs780784630 Chr.6: 75947479-75947480 on Build GRCh38
rs201013836 Chr.6: 75947420-75947420 on Build GRCh38
rs768507146 Chr.6: 75947629-75947629 on Build GRCh38
rs151138934 Chr.6: 75947525-75947525 on Build GRCh38
rs906664476 Chr.6: 75947375-75947375 on Build GRCh38
rs987530179 Chr.6: 75947187-75947187 on Build GRCh38
rs759162476 Chr.6: 75947465-75947465 on Build GRCh38
rs769638626 Chr.6: 75947521-75947521 on Build GRCh38
rs140401349 Chr.6: 75947592-75947592 on Build GRCh38
rs754657889 Chr.6: 75947396-75947396 on Build GRCh38
rs1030997793 Chr.6: 75947259-75947259 on Build GRCh38
rs200128845 Chr.6: 75947376-75947376 on Build GRCh38
rs940063319 Chr.6: 75947353-75947353 on Build GRCh38
rs764106748 Chr.6: 75947286-75947286 on Build GRCh38
rs777765131 Chr.6: 75947334-75947334 on Build GRCh38
rs576132697 Chr.6: 75947322-75947322 on Build GRCh38
rs753658284 Chr.6: 75947292-75947292 on Build GRCh38
rs765195875 Chr.6: 75947390-75947390 on Build GRCh38
rs931737685 Chr.6: 75947691-75947691 on Build GRCh38
rs753298586 Chr.6: 75947328-75947328 on Build GRCh38
rs767081146 Chr.6: 75947563-75947563 on Build GRCh38
rs751273411 Chr.6: 75947395-75947395 on Build GRCh38
rs200959900 Chr.6: 75947327-75947327 on Build GRCh38
rs761693136 Chr.6: 75947377-75947377 on Build GRCh38
rs749953054 Chr.6: 75947392-75947392 on Build GRCh38
rs921619269 Chr.6: 75947690-75947690 on Build GRCh38
rs372193265 Chr.6: 75947564-75947564 on Build GRCh38
rs984685022 Chr.6: 75947515-75947515 on Build GRCh38
rs771309602 Chr.6: 75947349-75947349 on Build GRCh38
rs760497546 Chr.6: 75947283-75947283 on Build GRCh38
rs556959981 Chr.6: 75947516-75947516 on Build GRCh38
rs776335869 Chr.6: 75947421-75947421 on Build GRCh38
rs975614325 Chr.6: 75947258-75947258 on Build GRCh38
rs958100377 Chr.6: 75947262-75947262 on Build GRCh38
rs573887374 Chr.6: 75947401-75947401 on Build GRCh38
rs774647095 Chr.6: 75947363-75947363 on Build GRCh38
rs75117418 Chr.6: 75947457-75947457 on Build GRCh38
rs78305682 Chr.6: 75947402-75947402 on Build GRCh38
rs749382585 Chr.6: 75947414-75947414 on Build GRCh38
rs375810046 Chr.6: 75947551-75947551 on Build GRCh38
rs749332855 Chr.6: 75947340-75947340 on Build GRCh38
rs764618020 Chr.6: 75947575-75947575 on Build GRCh38
rs756434628 Chr.6: 75947476-75947476 on Build GRCh38
rs754222080 Chr.6: 75947583-75947583 on Build GRCh38
rs770835713 Chr.6: 75947544-75947544 on Build GRCh38
rs760558589 Chr.6: 75947373-75947373 on Build GRCh38
rs749175021 Chr.6: 75947581-75947581 on Build GRCh38
rs749901493 Chr.6: 75947303-75947303 on Build GRCh38
rs1008603589 Chr.6: 75947434-75947434 on Build GRCh38
rs118155926 Chr.6: 75947413-75947413 on Build GRCh38
rs181685622 Chr.6: 75947675-75947675 on Build GRCh38
rs139192386 Chr.6: 75947426-75947426 on Build GRCh38
rs770205977 Chr.6: 75947564-75947565 on Build GRCh38
rs781610216 Chr.6: 75947317-75947317 on Build GRCh38
rs149949442 Chr.6: 75947699-75947699 on Build GRCh38
rs761616758 Chr.6: 75947422-75947422 on Build GRCh38
rs201889175 Chr.6: 75947263-75947263 on Build GRCh38
rs751040344 Chr.6: 75947461-75947461 on Build GRCh38
rs1017194704 Chr.6: 75947623-75947623 on Build GRCh38
rs766036273 Chr.6: 75947394-75947394 on Build GRCh38
rs745936741 Chr.6: 75947518-75947518 on Build GRCh38
rs937974870 Chr.6: 75947555-75947555 on Build GRCh38
rs1051579797 Chr.6: 75947482-75947482 on Build GRCh38
rs758091142 Chr.6: 75947307-75947307 on Build GRCh38
rs767150099 Chr.6: 75947467-75947467 on Build GRCh38
rs267601131 Chr.6: 75947409-75947409 on Build GRCh38
rs746117801 Chr.6: 75947482-75947486 on Build GRCh38
rs766676537 Chr.6: 75947264-75947264 on Build GRCh38
rs748671913 Chr.6: 75947320-75947320 on Build GRCh38
rs751929839 Chr.6: 75947314-75947314 on Build GRCh38
rs3215818 Chr.6: 75947262-75947262 on Build GRCh38
rs757107031 Chr.6: 75947477-75947478 on Build GRCh38

View Full Product Details