Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111575266-111575836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF3IP2
Entrez Gene ID
10758
Gene Name
TRAF3 interacting protein 2
Gene Aliases
ACT1, C6orf2, C6orf4, C6orf5, C6orf6, CANDF8, CIKS, PSORS13
UniGene
Hs.561514
Gene Chromosome Location
Chr.6: 111555378-111606874 on Build GRCh38
Transcripts
NM_001164281.2 NM_147200.2 NM_147686.3 XM_011535386.1 NR_034108.1 NR_034109.1 NR_034110.1

Gene Symbol
TRAF3IP2-AS1
Entrez Gene ID
643749
Gene Name
TRAF3IP2 antisense RNA 1
Gene Aliases
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2
UniGene
Hs.486228
Gene Chromosome Location
Chr.6: 111483472-111602295 on Build GRCh38
Transcripts
NM_001164281.2 NM_147200.2 NM_147686.3 XM_011535386.1 NR_034108.1 NR_034109.1 NR_034110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715298 c.1028A>T p.Q343L Chr.6: 111575816-111575816 on Build GRCh38
6282234 c.1182C>T p.S394S Chr.6: 111575662-111575662 on Build GRCh38
7149297 c.1201+1G>T p.? Chr.6: 111575642-111575642 on Build GRCh38
3674523 c.1066G>T p.G356W Chr.6: 111575778-111575778 on Build GRCh38
1268456 c.1201+1G>A p.? Chr.6: 111575642-111575642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030865920 Chr.6: 111575639-111575639 on Build GRCh38
rs551559794 Chr.6: 111575682-111575682 on Build GRCh38
rs753954583 Chr.6: 111575659-111575659 on Build GRCh38
rs764269377 Chr.6: 111575782-111575782 on Build GRCh38
rs149449506 Chr.6: 111575699-111575699 on Build GRCh38
rs750939608 Chr.6: 111575805-111575805 on Build GRCh38
rs564345969 Chr.6: 111575520-111575520 on Build GRCh38
rs778029105 Chr.6: 111575587-111575590 on Build GRCh38
rs760916707 Chr.6: 111575781-111575781 on Build GRCh38
rs992697374 Chr.6: 111575461-111575461 on Build GRCh38
rs766936250 Chr.6: 111575604-111575609 on Build GRCh38
rs1056214303 Chr.6: 111575421-111575421 on Build GRCh38
rs1047245538 Chr.6: 111575384-111575384 on Build GRCh38
rs978931110 Chr.6: 111575358-111575358 on Build GRCh38
rs762591912 Chr.6: 111575773-111575773 on Build GRCh38
rs60481646 Chr.6: 111575588-111575591 on Build GRCh38
rs756345544 Chr.6: 111575646-111575646 on Build GRCh38
rs755144332 Chr.6: 111575587-111575591 on Build GRCh38
rs752970544 Chr.6: 111575644-111575644 on Build GRCh38
rs191714491 Chr.6: 111575543-111575543 on Build GRCh38
rs771040089 Chr.6: 111575589-111575592 on Build GRCh38
rs561146758 Chr.6: 111575311-111575311 on Build GRCh38
rs763843514 Chr.6: 111575796-111575796 on Build GRCh38
rs546035926 Chr.6: 111575473-111575473 on Build GRCh38
rs541607649 Chr.6: 111575354-111575354 on Build GRCh38
rs756853938 Chr.6: 111575411-111575411 on Build GRCh38
rs765306789 Chr.6: 111575626-111575626 on Build GRCh38
rs779213459 Chr.6: 111575594-111575594 on Build GRCh38
rs919911102 Chr.6: 111575689-111575689 on Build GRCh38
rs752745789 Chr.6: 111575588-111575592 on Build GRCh38
rs867800419 Chr.6: 111575481-111575481 on Build GRCh38
rs768227542 Chr.6: 111575606-111575606 on Build GRCh38
rs61585604 Chr.6: 111575565-111575565 on Build GRCh38
rs767596361 Chr.6: 111575643-111575643 on Build GRCh38
rs746189713 Chr.6: 111575671-111575671 on Build GRCh38
rs764227946 Chr.6: 111575651-111575651 on Build GRCh38
rs776065221 Chr.6: 111575622-111575622 on Build GRCh38
rs112963658 Chr.6: 111575321-111575321 on Build GRCh38
rs942712382 Chr.6: 111575325-111575325 on Build GRCh38
rs896144447 Chr.6: 111575480-111575480 on Build GRCh38
rs766180997 Chr.6: 111575774-111575774 on Build GRCh38
rs67628622 Chr.6: 111575605-111575608 on Build GRCh38
rs1022432047 Chr.6: 111575734-111575734 on Build GRCh38
rs373128911 Chr.6: 111575593-111575593 on Build GRCh38
rs940478847 Chr.6: 111575393-111575393 on Build GRCh38
rs887266199 Chr.6: 111575385-111575385 on Build GRCh38
rs775238458 Chr.6: 111575597-111575597 on Build GRCh38
rs148619849 Chr.6: 111575786-111575786 on Build GRCh38
rs948642130 Chr.6: 111575482-111575484 on Build GRCh38
rs12192705 Chr.6: 111575574-111575574 on Build GRCh38
rs187790575 Chr.6: 111575374-111575374 on Build GRCh38
rs529388792 Chr.6: 111575583-111575583 on Build GRCh38
rs143865750 Chr.6: 111575780-111575780 on Build GRCh38
rs758888448 Chr.6: 111575812-111575812 on Build GRCh38
rs139767840 Chr.6: 111575660-111575660 on Build GRCh38
rs746279732 Chr.6: 111575595-111575595 on Build GRCh38
rs975417820 Chr.6: 111575677-111575677 on Build GRCh38
rs370011231 Chr.6: 111575725-111575725 on Build GRCh38
rs903748845 Chr.6: 111575536-111575536 on Build GRCh38
rs1019234289 Chr.6: 111575303-111575303 on Build GRCh38
rs768260854 Chr.6: 111575703-111575703 on Build GRCh38
rs796093475 Chr.6: 111575542-111575542 on Build GRCh38
rs912585102 Chr.6: 111575317-111575317 on Build GRCh38
rs920150404 Chr.6: 111575364-111575364 on Build GRCh38
rs904037371 Chr.6: 111575755-111575755 on Build GRCh38
rs754308740 Chr.6: 111575608-111575609 on Build GRCh38
rs769810239 Chr.6: 111575753-111575753 on Build GRCh38
rs765663607 Chr.6: 111575798-111575798 on Build GRCh38
rs773316350 Chr.6: 111575762-111575762 on Build GRCh38
rs753164704 Chr.6: 111575344-111575344 on Build GRCh38
rs375800644 Chr.6: 111575642-111575642 on Build GRCh38
rs575543007 Chr.6: 111575415-111575415 on Build GRCh38
rs758792853 Chr.6: 111575675-111575675 on Build GRCh38
rs749779623 Chr.6: 111575592-111575592 on Build GRCh38
rs747713162 Chr.6: 111575726-111575726 on Build GRCh38
rs778945109 Chr.6: 111575590-111575592 on Build GRCh38
rs746435840 Chr.6: 111575605-111575605 on Build GRCh38
rs771574504 Chr.6: 111575596-111575596 on Build GRCh38
rs761838585 Chr.6: 111575623-111575623 on Build GRCh38
rs12206072 Chr.6: 111575589-111575589 on Build GRCh38
rs1011879971 Chr.6: 111575513-111575513 on Build GRCh38

View Full Product Details