Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23717403-23717928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK31
Entrez Gene ID
56164
Gene Name
serine/threonine kinase 31
Gene Aliases
SGK396, TDRD8
UniGene
Hs.309767
Gene Chromosome Location
Chr.7: 23710141-23832513 on Build GRCh38
Transcripts
NM_001260504.1 NM_001260505.1 NM_031414.4 NM_032944.3 NR_048542.1 XM_006715758.3 XM_011515449.2 XM_011515450.1 XM_011515452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7197957 c.169G>C p.D57H Chr.7: 23717499-23717499 on Build GRCh38
3762594 c.213G>C p.Q71H Chr.7: 23717543-23717543 on Build GRCh38
746390 c.151-2A>G p.? Chr.7: 23717479-23717479 on Build GRCh38
3394690 c.231G>T p.G77G Chr.7: 23717561-23717561 on Build GRCh38
6306141 c.161G>A p.R54K Chr.7: 23717491-23717491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200349502 Chr.7: 23717562-23717562 on Build GRCh38
rs764978791 Chr.7: 23717633-23717633 on Build GRCh38
rs753375169 Chr.7: 23717539-23717539 on Build GRCh38
rs911328971 Chr.7: 23717617-23717617 on Build GRCh38
rs756037399 Chr.7: 23717463-23717464 on Build GRCh38
rs767273579 Chr.7: 23717435-23717436 on Build GRCh38
rs766492565 Chr.7: 23717537-23717537 on Build GRCh38
rs750272264 Chr.7: 23717458-23717462 on Build GRCh38
rs565850936 Chr.7: 23717719-23717719 on Build GRCh38
rs185679959 Chr.7: 23717889-23717889 on Build GRCh38
rs980368443 Chr.7: 23717771-23717771 on Build GRCh38
rs930008918 Chr.7: 23717874-23717874 on Build GRCh38
rs904134728 Chr.7: 23717643-23717643 on Build GRCh38
rs375333272 Chr.7: 23717442-23717442 on Build GRCh38
rs142516836 Chr.7: 23717504-23717504 on Build GRCh38
rs969895270 Chr.7: 23717767-23717767 on Build GRCh38
rs1056285777 Chr.7: 23717658-23717658 on Build GRCh38
rs960699792 Chr.7: 23717744-23717744 on Build GRCh38
rs970306253 Chr.7: 23717476-23717476 on Build GRCh38
rs918216236 Chr.7: 23717849-23717849 on Build GRCh38
rs780593481 Chr.7: 23717594-23717594 on Build GRCh38
rs1031708358 Chr.7: 23717764-23717764 on Build GRCh38
rs974095532 Chr.7: 23717830-23717831 on Build GRCh38
rs760659057 Chr.7: 23717534-23717534 on Build GRCh38
rs749329174 Chr.7: 23717604-23717604 on Build GRCh38
rs933713249 Chr.7: 23717613-23717613 on Build GRCh38
rs2058995 Chr.7: 23717641-23717641 on Build GRCh38
rs768748364 Chr.7: 23717840-23717840 on Build GRCh38
rs761633249 Chr.7: 23717503-23717503 on Build GRCh38
rs939217787 Chr.7: 23717646-23717646 on Build GRCh38
rs753729762 Chr.7: 23717477-23717477 on Build GRCh38
rs527284389 Chr.7: 23717445-23717445 on Build GRCh38
rs752323769 Chr.7: 23717546-23717546 on Build GRCh38
rs6945306 Chr.7: 23717543-23717543 on Build GRCh38
rs539162374 Chr.7: 23717807-23717807 on Build GRCh38
rs746394445 Chr.7: 23717456-23717456 on Build GRCh38
rs1051293668 Chr.7: 23717614-23717614 on Build GRCh38
rs770331156 Chr.7: 23717458-23717458 on Build GRCh38
rs781632972 Chr.7: 23717454-23717454 on Build GRCh38
rs907813641 Chr.7: 23717890-23717890 on Build GRCh38
rs974063128 Chr.7: 23717622-23717622 on Build GRCh38
rs267601466 Chr.7: 23717499-23717499 on Build GRCh38
rs1011977349 Chr.7: 23717716-23717716 on Build GRCh38
rs1031048387 Chr.7: 23717730-23717730 on Build GRCh38
rs547825144 Chr.7: 23717482-23717482 on Build GRCh38
rs192396365 Chr.7: 23717639-23717639 on Build GRCh38
rs564231836 Chr.7: 23717567-23717567 on Build GRCh38
rs757567571 Chr.7: 23717553-23717553 on Build GRCh38
rs1024154603 Chr.7: 23717754-23717754 on Build GRCh38
rs76512493 Chr.7: 23717821-23717821 on Build GRCh38
rs903487063 Chr.7: 23717734-23717734 on Build GRCh38
rs372861236 Chr.7: 23717619-23717619 on Build GRCh38
rs372521231 Chr.7: 23717776-23717776 on Build GRCh38
rs763005784 Chr.7: 23717485-23717485 on Build GRCh38
rs559246818 Chr.7: 23717424-23717424 on Build GRCh38
rs113382775 Chr.7: 23717488-23717488 on Build GRCh38
rs758350725 Chr.7: 23717712-23717712 on Build GRCh38
rs368706626 Chr.7: 23717576-23717576 on Build GRCh38
rs879672661 Chr.7: 23717777-23717777 on Build GRCh38
rs771204206 Chr.7: 23717452-23717452 on Build GRCh38
rs894919718 Chr.7: 23717660-23717660 on Build GRCh38
rs774444994 Chr.7: 23717491-23717491 on Build GRCh38
rs999519739 Chr.7: 23717672-23717672 on Build GRCh38
rs569986096 Chr.7: 23717787-23717787 on Build GRCh38
rs919899406 Chr.7: 23717632-23717632 on Build GRCh38
rs767445764 Chr.7: 23717513-23717513 on Build GRCh38
rs1013639481 Chr.7: 23717751-23717751 on Build GRCh38
rs773193165 Chr.7: 23717532-23717532 on Build GRCh38
rs766234814 Chr.7: 23717471-23717475 on Build GRCh38
rs146120284 Chr.7: 23717885-23717885 on Build GRCh38
rs754654215 Chr.7: 23717441-23717441 on Build GRCh38

View Full Product Details