Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17282553-17283120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUCB2
Entrez Gene ID
4925
Gene Name
nucleobindin 2
Gene Aliases
HEL-S-109, NEFA
UniGene
Hs.654599
Gene Chromosome Location
Chr.11: 17260340-17331881 on Build GRCh38
Transcripts
NM_005013.2 XM_017017804.1 XM_017017805.1 XM_017017806.1 XM_017017807.1 XM_017017808.1 XM_017017809.1 XM_017017810.1 XM_017017811.1 XM_017017812.1 XM_017017813.1 XM_017017814.1 XM_017017815.1 XM_017017816.1 XM_017017817.1 XM_017017818.1 XM_017017819.1 XM_017017820.1 XM_017017821.1 XM_017017822.1 XM_017017823.1 XM_017017824.1 XM_017017825.1 XM_017017826.1 XM_017017827.1 XM_017017828.1 XM_017017829.1

SNP Detail

SNP ID Chromosome Location
rs577901308 Chr.11: 17282669-17282669 on Build GRCh38
rs888223670 Chr.11: 17282776-17282776 on Build GRCh38
rs377120469 Chr.11: 17282748-17282748 on Build GRCh38
rs978172883 Chr.11: 17282635-17282635 on Build GRCh38
rs760293690 Chr.11: 17282752-17282752 on Build GRCh38
rs966655710 Chr.11: 17282943-17282943 on Build GRCh38
rs959066782 Chr.11: 17282761-17282761 on Build GRCh38
rs991088219 Chr.11: 17282652-17282652 on Build GRCh38
rs115767617 Chr.11: 17282592-17282592 on Build GRCh38
rs535611484 Chr.11: 17282754-17282757 on Build GRCh38
rs749480688 Chr.11: 17282882-17282882 on Build GRCh38
rs1018676141 Chr.11: 17282987-17282987 on Build GRCh38
rs893912208 Chr.11: 17282852-17282852 on Build GRCh38
rs763938837 Chr.11: 17282792-17282792 on Build GRCh38
rs982002758 Chr.11: 17283075-17283075 on Build GRCh38
rs1053676145 Chr.11: 17282799-17282799 on Build GRCh38
rs199576740 Chr.11: 17283055-17283056 on Build GRCh38
rs1003712650 Chr.11: 17282906-17282906 on Build GRCh38
rs916385032 Chr.11: 17283056-17283056 on Build GRCh38
rs900680574 Chr.11: 17282881-17282881 on Build GRCh38
rs572700262 Chr.11: 17282798-17282798 on Build GRCh38
rs540532614 Chr.11: 17282678-17282678 on Build GRCh38
rs1004233091 Chr.11: 17282743-17282744 on Build GRCh38
rs771462771 Chr.11: 17282901-17282901 on Build GRCh38
rs1053834290 Chr.11: 17282681-17282681 on Build GRCh38
rs577029492 Chr.11: 17282579-17282579 on Build GRCh38
rs925375521 Chr.11: 17282643-17282643 on Build GRCh38
rs899870713 Chr.11: 17282637-17282637 on Build GRCh38
rs189752241 Chr.11: 17282886-17282886 on Build GRCh38
rs983322267 Chr.11: 17282771-17282772 on Build GRCh38
rs564233466 Chr.11: 17283032-17283032 on Build GRCh38
rs976053934 Chr.11: 17282581-17282581 on Build GRCh38
rs540005885 Chr.11: 17282962-17282962 on Build GRCh38
rs973056595 Chr.11: 17282765-17282765 on Build GRCh38
rs1041063074 Chr.11: 17282628-17282628 on Build GRCh38
rs983634249 Chr.11: 17282981-17282981 on Build GRCh38
rs186314980 Chr.11: 17282650-17282650 on Build GRCh38
rs970562811 Chr.11: 17283062-17283062 on Build GRCh38
rs971017997 Chr.11: 17282708-17282708 on Build GRCh38
rs747773542 Chr.11: 17282617-17282617 on Build GRCh38
rs1032413842 Chr.11: 17283011-17283011 on Build GRCh38
rs1027299253 Chr.11: 17282955-17282955 on Build GRCh38
rs944540738 Chr.11: 17282772-17282772 on Build GRCh38
rs895366345 Chr.11: 17282944-17282944 on Build GRCh38
rs1033964863 Chr.11: 17282917-17282917 on Build GRCh38
rs560443962 Chr.11: 17282877-17282877 on Build GRCh38
rs556270205 Chr.11: 17282618-17282618 on Build GRCh38
rs994505410 Chr.11: 17282948-17282948 on Build GRCh38
rs756088361 Chr.11: 17282671-17282671 on Build GRCh38
rs920339292 Chr.11: 17282578-17282578 on Build GRCh38
rs949246052 Chr.11: 17282699-17282699 on Build GRCh38
rs778161773 Chr.11: 17282680-17282680 on Build GRCh38
rs541843333 Chr.11: 17282770-17282770 on Build GRCh38
rs1048127914 Chr.11: 17282771-17282771 on Build GRCh38
rs958396209 Chr.11: 17283018-17283018 on Build GRCh38
rs966309788 Chr.11: 17283054-17283054 on Build GRCh38
rs114057699 Chr.11: 17282791-17282791 on Build GRCh38
rs546135231 Chr.11: 17283000-17283000 on Build GRCh38
rs1027413148 Chr.11: 17282767-17282767 on Build GRCh38
rs893861170 Chr.11: 17282690-17282690 on Build GRCh38
rs1046355049 Chr.11: 17282701-17282701 on Build GRCh38

View Full Product Details