Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 99819414-99819760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN5
Entrez Gene ID
53942
Gene Name
contactin 5
Gene Aliases
HNB-2s, NB-2
UniGene
Hs.656783
Gene Chromosome Location
Chr.11: 99020904-100358885 on Build GRCh38
Transcripts
NM_001243270.1 NM_001243271.1 NM_014361.3 NM_175566.2 XM_011542871.1 XM_011542873.1 XM_017017926.1 XM_017017927.1 XM_017017928.1 XM_017017929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870494 c.122C>T p.S41L Chr.11: 99819610-99819610 on Build GRCh38
4592079 c.242A>G p.N81S Chr.11: 99819730-99819730 on Build GRCh38
690850 c.125C>G p.S42C Chr.11: 99819613-99819613 on Build GRCh38
3870493 c.122C>T p.S41L Chr.11: 99819610-99819610 on Build GRCh38
5360384 c.51-10delT p.? Chr.11: 99819529-99819529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758053014 Chr.11: 99819733-99819733 on Build GRCh38
rs756903722 Chr.11: 99819666-99819666 on Build GRCh38
rs201399847 Chr.11: 99819703-99819703 on Build GRCh38
rs199786366 Chr.11: 99819484-99819484 on Build GRCh38
rs199920688 Chr.11: 99819560-99819560 on Build GRCh38
rs746426286 Chr.11: 99819542-99819542 on Build GRCh38
rs532915134 Chr.11: 99819552-99819552 on Build GRCh38
rs202035562 Chr.11: 99819564-99819564 on Build GRCh38
rs757789255 Chr.11: 99819538-99819538 on Build GRCh38
rs753301950 Chr.11: 99819725-99819725 on Build GRCh38
rs7125822 Chr.11: 99819697-99819697 on Build GRCh38
rs372476071 Chr.11: 99819533-99819533 on Build GRCh38
rs200788939 Chr.11: 99819735-99819735 on Build GRCh38
rs769441323 Chr.11: 99819551-99819551 on Build GRCh38
rs770692990 Chr.11: 99819617-99819617 on Build GRCh38
rs775243907 Chr.11: 99819553-99819554 on Build GRCh38
rs755046611 Chr.11: 99819522-99819522 on Build GRCh38
rs760918286 Chr.11: 99819571-99819571 on Build GRCh38
rs369959040 Chr.11: 99819528-99819528 on Build GRCh38
rs867939537 Chr.11: 99819705-99819705 on Build GRCh38
rs376713995 Chr.11: 99819527-99819527 on Build GRCh38
rs759322436 Chr.11: 99819625-99819625 on Build GRCh38
rs374794794 Chr.11: 99819616-99819616 on Build GRCh38
rs200572550 Chr.11: 99819702-99819702 on Build GRCh38
rs367832129 Chr.11: 99819502-99819502 on Build GRCh38
rs202073412 Chr.11: 99819736-99819736 on Build GRCh38
rs200637003 Chr.11: 99819722-99819722 on Build GRCh38
rs756168987 Chr.11: 99819461-99819461 on Build GRCh38
rs775466035 Chr.11: 99819671-99819671 on Build GRCh38
rs12292548 Chr.11: 99819481-99819481 on Build GRCh38
rs202004805 Chr.11: 99819595-99819595 on Build GRCh38
rs774266423 Chr.11: 99819619-99819619 on Build GRCh38
rs765004879 Chr.11: 99819496-99819496 on Build GRCh38
rs201682523 Chr.11: 99819726-99819726 on Build GRCh38
rs753357501 Chr.11: 99819690-99819690 on Build GRCh38
rs775378819 Chr.11: 99819630-99819630 on Build GRCh38
rs1012593226 Chr.11: 99819485-99819485 on Build GRCh38
rs369643416 Chr.11: 99819567-99819567 on Build GRCh38
rs202208723 Chr.11: 99819658-99819658 on Build GRCh38
rs764309266 Chr.11: 99819634-99819634 on Build GRCh38
rs754214109 Chr.11: 99819583-99819583 on Build GRCh38
rs746937022 Chr.11: 99819699-99819699 on Build GRCh38
rs201117922 Chr.11: 99819612-99819612 on Build GRCh38
rs200581115 Chr.11: 99819563-99819563 on Build GRCh38
rs779593509 Chr.11: 99819539-99819539 on Build GRCh38
rs12292659 Chr.11: 99819645-99819645 on Build GRCh38
rs752042635 Chr.11: 99819685-99819685 on Build GRCh38
rs7105653 Chr.11: 99819456-99819456 on Build GRCh38
rs200519976 Chr.11: 99819682-99819682 on Build GRCh38
rs768279431 Chr.11: 99819587-99819587 on Build GRCh38
rs541792239 Chr.11: 99819453-99819453 on Build GRCh38
rs776491575 Chr.11: 99819721-99819721 on Build GRCh38
rs529404054 Chr.11: 99819627-99819627 on Build GRCh38
rs768027790 Chr.11: 99819545-99819545 on Build GRCh38
rs780502497 Chr.11: 99819607-99819607 on Build GRCh38
rs201422329 Chr.11: 99819683-99819683 on Build GRCh38
rs755593251 Chr.11: 99819653-99819653 on Build GRCh38
rs200679586 Chr.11: 99819523-99819523 on Build GRCh38
rs767487705 Chr.11: 99819565-99819565 on Build GRCh38
rs777315487 Chr.11: 99819654-99819654 on Build GRCh38
rs759012669 Chr.11: 99819599-99819599 on Build GRCh38
rs777437500 Chr.11: 99819613-99819613 on Build GRCh38
rs748932737 Chr.11: 99819663-99819663 on Build GRCh38
rs944995336 Chr.11: 99819651-99819651 on Build GRCh38
rs199950154 Chr.11: 99819476-99819476 on Build GRCh38
rs747751684 Chr.11: 99819548-99819548 on Build GRCh38
rs771826877 Chr.11: 99819530-99819531 on Build GRCh38
rs745546818 Chr.11: 99819696-99819696 on Build GRCh38
rs752165707 Chr.11: 99819646-99819646 on Build GRCh38
rs762182166 Chr.11: 99819641-99819641 on Build GRCh38
rs766320825 Chr.11: 99819508-99819508 on Build GRCh38
rs749249067 Chr.11: 99819448-99819448 on Build GRCh38
rs763136607 Chr.11: 99819718-99819718 on Build GRCh38
rs202179266 Chr.11: 99819642-99819642 on Build GRCh38
rs757664109 Chr.11: 99819588-99819588 on Build GRCh38
rs745694465 Chr.11: 99819670-99819670 on Build GRCh38
rs745428164 Chr.11: 99819528-99819529 on Build GRCh38
rs764702381 Chr.11: 99819729-99819729 on Build GRCh38
rs780813895 Chr.11: 99819547-99819547 on Build GRCh38
rs768210733 Chr.11: 99819665-99819665 on Build GRCh38
rs200685098 Chr.11: 99819504-99819504 on Build GRCh38
rs199701621 Chr.11: 99819519-99819519 on Build GRCh38
rs756806886 Chr.11: 99819692-99819692 on Build GRCh38
rs754089407 Chr.11: 99819638-99819638 on Build GRCh38
rs199945392 Chr.11: 99819604-99819604 on Build GRCh38
rs760869154 Chr.11: 99819632-99819632 on Build GRCh38
rs201383460 Chr.11: 99819509-99819509 on Build GRCh38
rs10893933 Chr.11: 99819730-99819730 on Build GRCh38
rs768030519 Chr.11: 99819723-99819723 on Build GRCh38
rs185691534 Chr.11: 99819474-99819474 on Build GRCh38
rs769870676 Chr.11: 99819715-99819715 on Build GRCh38
rs10790978 Chr.11: 99819555-99819555 on Build GRCh38
rs201084279 Chr.11: 99819594-99819594 on Build GRCh38
rs200129740 Chr.11: 99819569-99819569 on Build GRCh38
rs201386179 Chr.11: 99819652-99819652 on Build GRCh38
rs761355116 Chr.11: 99819728-99819728 on Build GRCh38
rs776919813 Chr.11: 99819673-99819673 on Build GRCh38
rs552066224 Chr.11: 99819687-99819687 on Build GRCh38
rs765510767 Chr.11: 99819676-99819676 on Build GRCh38
rs537629421 Chr.11: 99819716-99819716 on Build GRCh38
rs745803839 Chr.11: 99819500-99819500 on Build GRCh38
rs778655565 Chr.11: 99819693-99819693 on Build GRCh38
rs746681632 Chr.11: 99819566-99819567 on Build GRCh38
rs565909092 Chr.11: 99819643-99819643 on Build GRCh38
rs778212750 Chr.11: 99819530-99819530 on Build GRCh38
rs1054639707 Chr.11: 99819470-99819470 on Build GRCh38
rs764432285 Chr.11: 99819580-99819580 on Build GRCh38
rs879012990 Chr.11: 99819443-99819445 on Build GRCh38
rs868836971 Chr.11: 99819577-99819577 on Build GRCh38
rs201201223 Chr.11: 99819711-99819711 on Build GRCh38
rs747707558 Chr.11: 99819611-99819611 on Build GRCh38
rs778510250 Chr.11: 99819667-99819667 on Build GRCh38

View Full Product Details