Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 33406482-33407077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT10
Entrez Gene ID
341359
Gene Name
synaptotagmin 10
Gene Aliases
-
UniGene
Hs.118703
Gene Chromosome Location
Chr.12: 33375413-33439819 on Build GRCh38
Transcripts
NM_198992.3 XM_011520644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4397591 c.1052G>A p.W351* Chr.12: 33406814-33406814 on Build GRCh38
6072435 c.908C>A p.P303H Chr.12: 33406958-33406958 on Build GRCh38
2206184 c.1013T>C p.L338S Chr.12: 33406853-33406853 on Build GRCh38
6553130 c.866G>A p.R289K Chr.12: 33407000-33407000 on Build GRCh38
6531473 c.896C>G p.T299S Chr.12: 33406970-33406970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766147514 Chr.12: 33406756-33406756 on Build GRCh38
rs751127584 Chr.12: 33406961-33406961 on Build GRCh38
rs973903820 Chr.12: 33406504-33406504 on Build GRCh38
rs752724042 Chr.12: 33406931-33406931 on Build GRCh38
rs1042570996 Chr.12: 33406996-33406996 on Build GRCh38
rs759382079 Chr.12: 33406944-33406944 on Build GRCh38
rs111268745 Chr.12: 33406578-33406578 on Build GRCh38
rs945524272 Chr.12: 33406627-33406627 on Build GRCh38
rs777388386 Chr.12: 33406793-33406793 on Build GRCh38
rs538524814 Chr.12: 33406576-33406576 on Build GRCh38
rs140528645 Chr.12: 33406662-33406662 on Build GRCh38
rs148091182 Chr.12: 33406851-33406851 on Build GRCh38
rs765148812 Chr.12: 33407035-33407035 on Build GRCh38
rs532944176 Chr.12: 33406702-33406702 on Build GRCh38
rs768575228 Chr.12: 33406635-33406637 on Build GRCh38
rs549633506 Chr.12: 33406718-33406718 on Build GRCh38
rs571810055 Chr.12: 33406766-33406766 on Build GRCh38
rs541132224 Chr.12: 33407010-33407010 on Build GRCh38
rs1905428 Chr.12: 33406540-33406540 on Build GRCh38
rs10844581 Chr.12: 33406658-33406658 on Build GRCh38
rs775643779 Chr.12: 33406764-33406764 on Build GRCh38
rs2389197 Chr.12: 33406589-33406589 on Build GRCh38
rs770240151 Chr.12: 33407004-33407004 on Build GRCh38
rs375766818 Chr.12: 33406742-33406742 on Build GRCh38
rs746248442 Chr.12: 33406837-33406837 on Build GRCh38
rs770010023 Chr.12: 33406838-33406838 on Build GRCh38
rs921605009 Chr.12: 33406597-33406597 on Build GRCh38
rs912954634 Chr.12: 33406934-33406934 on Build GRCh38
rs186865193 Chr.12: 33406721-33406721 on Build GRCh38
rs764919133 Chr.12: 33406924-33406924 on Build GRCh38
rs769167593 Chr.12: 33407007-33407007 on Build GRCh38
rs373995269 Chr.12: 33406865-33406865 on Build GRCh38
rs757469162 Chr.12: 33406745-33406745 on Build GRCh38
rs746724273 Chr.12: 33406805-33406805 on Build GRCh38
rs760778883 Chr.12: 33407029-33407029 on Build GRCh38
rs531915199 Chr.12: 33406736-33406736 on Build GRCh38
rs747135559 Chr.12: 33406997-33406997 on Build GRCh38
rs763243494 Chr.12: 33407002-33407002 on Build GRCh38
rs747987045 Chr.12: 33406771-33406771 on Build GRCh38
rs151316189 Chr.12: 33406600-33406600 on Build GRCh38
rs866101269 Chr.12: 33406959-33406959 on Build GRCh38
rs114233167 Chr.12: 33406578-33406578 on Build GRCh38
rs141819559 Chr.12: 33406912-33406912 on Build GRCh38
rs764196418 Chr.12: 33406859-33406859 on Build GRCh38
rs560234594 Chr.12: 33406711-33406711 on Build GRCh38
rs777482060 Chr.12: 33406953-33406953 on Build GRCh38
rs895139652 Chr.12: 33406661-33406661 on Build GRCh38
rs758611055 Chr.12: 33406900-33406900 on Build GRCh38
rs761826830 Chr.12: 33406883-33406883 on Build GRCh38
rs528959573 Chr.12: 33406712-33406712 on Build GRCh38
rs754823879 Chr.12: 33406916-33406916 on Build GRCh38
rs767787877 Chr.12: 33406887-33406887 on Build GRCh38
rs751501752 Chr.12: 33407051-33407051 on Build GRCh38
rs554252150 Chr.12: 33406595-33406595 on Build GRCh38
rs533896426 Chr.12: 33406594-33406594 on Build GRCh38
rs762776606 Chr.12: 33407044-33407044 on Build GRCh38
rs767336073 Chr.12: 33407008-33407008 on Build GRCh38
rs778941836 Chr.12: 33406769-33406769 on Build GRCh38
rs745458244 Chr.12: 33406984-33406984 on Build GRCh38
rs758034056 Chr.12: 33406772-33406772 on Build GRCh38
rs750658161 Chr.12: 33406848-33406851 on Build GRCh38
rs756761234 Chr.12: 33406963-33406963 on Build GRCh38
rs369857680 Chr.12: 33406852-33406852 on Build GRCh38
rs780248794 Chr.12: 33407005-33407005 on Build GRCh38
rs752525618 Chr.12: 33407039-33407039 on Build GRCh38
rs776089898 Chr.12: 33406827-33406827 on Build GRCh38
rs536056246 Chr.12: 33406515-33406515 on Build GRCh38
rs181668606 Chr.12: 33406634-33406634 on Build GRCh38
rs983301756 Chr.12: 33406577-33406577 on Build GRCh38
rs753685463 Chr.12: 33406765-33406765 on Build GRCh38
rs1018770716 Chr.12: 33406831-33406831 on Build GRCh38
rs1003831632 Chr.12: 33406651-33406651 on Build GRCh38
rs16920985 Chr.12: 33406738-33406738 on Build GRCh38
rs780107776 Chr.12: 33406960-33406960 on Build GRCh38
rs146031713 Chr.12: 33407009-33407009 on Build GRCh38
rs935726648 Chr.12: 33406601-33406601 on Build GRCh38
rs534242250 Chr.12: 33406932-33406932 on Build GRCh38
rs1054633179 Chr.12: 33406602-33406602 on Build GRCh38
rs770692294 Chr.12: 33406808-33406808 on Build GRCh38
rs539804229 Chr.12: 33406708-33406708 on Build GRCh38
rs773533809 Chr.12: 33407012-33407012 on Build GRCh38
rs764131425 Chr.12: 33407050-33407050 on Build GRCh38
rs11052684 Chr.12: 33406644-33406644 on Build GRCh38
rs541666175 Chr.12: 33406621-33406621 on Build GRCh38
rs569599951 Chr.12: 33406571-33406571 on Build GRCh38
rs764109882 Chr.12: 33406740-33406740 on Build GRCh38
rs144188198 Chr.12: 33406979-33406979 on Build GRCh38
rs965903003 Chr.12: 33406840-33406841 on Build GRCh38

View Full Product Details