Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6390071-6390471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBR
Entrez Gene ID
4055
Gene Name
lymphotoxin beta receptor
Gene Aliases
CD18, D12S370, LT-BETA-R, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFR3, TNFRSF3
UniGene
Hs.1116
Gene Chromosome Location
Chr.12: 6375368-6391571 on Build GRCh38
Transcripts
NM_001270987.1 NM_002342.2 XM_005253688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
694795 c.893C>G p.S298C Chr.12: 6390203-6390203 on Build GRCh38
1991257 c.956C>A p.P319Q Chr.12: 6390266-6390266 on Build GRCh38
5933080 c.887C>T p.P296L Chr.12: 6390197-6390197 on Build GRCh38
5747362 c.902T>C p.V301A Chr.12: 6390212-6390212 on Build GRCh38
7292302 c.963G>C p.Q321H Chr.12: 6390273-6390273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748659594 Chr.12: 6390303-6390303 on Build GRCh38
rs747211408 Chr.12: 6390190-6390190 on Build GRCh38
rs555685333 Chr.12: 6390383-6390383 on Build GRCh38
rs756591790 Chr.12: 6390131-6390131 on Build GRCh38
rs557471207 Chr.12: 6390109-6390109 on Build GRCh38
rs530984880 Chr.12: 6390263-6390263 on Build GRCh38
rs774000672 Chr.12: 6390300-6390300 on Build GRCh38
rs750574965 Chr.12: 6390158-6390158 on Build GRCh38
rs1029284604 Chr.12: 6390163-6390163 on Build GRCh38
rs2286599 Chr.12: 6390367-6390367 on Build GRCh38
rs766807673 Chr.12: 6390376-6390376 on Build GRCh38
rs370657088 Chr.12: 6390269-6390269 on Build GRCh38
rs201042007 Chr.12: 6390231-6390231 on Build GRCh38
rs758405373 Chr.12: 6390168-6390168 on Build GRCh38
rs758556929 Chr.12: 6390253-6390253 on Build GRCh38
rs757842062 Chr.12: 6390152-6390152 on Build GRCh38
rs893019225 Chr.12: 6390403-6390403 on Build GRCh38
rs990526922 Chr.12: 6390202-6390202 on Build GRCh38
rs764572877 Chr.12: 6390139-6390139 on Build GRCh38
rs74400611 Chr.12: 6390170-6390170 on Build GRCh38
rs750864837 Chr.12: 6390247-6390247 on Build GRCh38
rs535783221 Chr.12: 6390377-6390377 on Build GRCh38
rs1004227070 Chr.12: 6390429-6390429 on Build GRCh38
rs749203106 Chr.12: 6390281-6390281 on Build GRCh38
rs34407931 Chr.12: 6390237-6390237 on Build GRCh38
rs905143498 Chr.12: 6390401-6390401 on Build GRCh38
rs762069381 Chr.12: 6390243-6390243 on Build GRCh38
rs1804534 Chr.12: 6390305-6390305 on Build GRCh38
rs374220510 Chr.12: 6390315-6390315 on Build GRCh38
rs770859209 Chr.12: 6390297-6390297 on Build GRCh38
rs892152990 Chr.12: 6390205-6390205 on Build GRCh38
rs949037029 Chr.12: 6390375-6390375 on Build GRCh38
rs532797854 Chr.12: 6390313-6390313 on Build GRCh38
rs35681405 Chr.12: 6390130-6390130 on Build GRCh38
rs1011474135 Chr.12: 6390406-6390406 on Build GRCh38
rs1046355145 Chr.12: 6390218-6390218 on Build GRCh38
rs888297420 Chr.12: 6390410-6390410 on Build GRCh38
rs148530627 Chr.12: 6390267-6390267 on Build GRCh38
rs949254102 Chr.12: 6390209-6390209 on Build GRCh38
rs144056108 Chr.12: 6390212-6390212 on Build GRCh38
rs957317292 Chr.12: 6390180-6390180 on Build GRCh38
rs780363968 Chr.12: 6390171-6390171 on Build GRCh38
rs779228196 Chr.12: 6390153-6390153 on Build GRCh38
rs888567841 Chr.12: 6390227-6390227 on Build GRCh38
rs376673488 Chr.12: 6390423-6390423 on Build GRCh38
rs901602769 Chr.12: 6390370-6390370 on Build GRCh38
rs770323197 Chr.12: 6390234-6390234 on Build GRCh38
rs765840282 Chr.12: 6390244-6390244 on Build GRCh38
rs754214226 Chr.12: 6390147-6390147 on Build GRCh38
rs11555386 Chr.12: 6390196-6390196 on Build GRCh38
rs996377001 Chr.12: 6390400-6390400 on Build GRCh38
rs371667034 Chr.12: 6390350-6390350 on Build GRCh38
rs759292715 Chr.12: 6390238-6390238 on Build GRCh38
rs765050173 Chr.12: 6390091-6390091 on Build GRCh38
rs550280761 Chr.12: 6390266-6390266 on Build GRCh38
rs976931721 Chr.12: 6390265-6390265 on Build GRCh38
rs144755581 Chr.12: 6390242-6390242 on Build GRCh38
rs886168809 Chr.12: 6390108-6390108 on Build GRCh38
rs768820155 Chr.12: 6390193-6390193 on Build GRCh38
rs762314697 Chr.12: 6390119-6390119 on Build GRCh38
rs147770937 Chr.12: 6390219-6390219 on Build GRCh38
rs771908386 Chr.12: 6390314-6390314 on Build GRCh38
rs140869621 Chr.12: 6390223-6390223 on Build GRCh38
rs760040112 Chr.12: 6390385-6390386 on Build GRCh38
rs368425660 Chr.12: 6390324-6390324 on Build GRCh38
rs755041714 Chr.12: 6390379-6390379 on Build GRCh38
rs767210693 Chr.12: 6390239-6390239 on Build GRCh38
rs780130706 Chr.12: 6390260-6390260 on Build GRCh38
rs750197738 Chr.12: 6390343-6390343 on Build GRCh38
rs1028837678 Chr.12: 6390402-6390402 on Build GRCh38

View Full Product Details