Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50110848-50111442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD1
Entrez Gene ID
2819
Gene Name
glycerol-3-phosphate dehydrogenase 1
Gene Aliases
GPD-C, GPDH-C, HTGTI
UniGene
Hs.524418
Gene Chromosome Location
Chr.12: 50103819-50111320 on Build GRCh38
Transcripts
NM_001257199.1 NM_005276.3

SNP Detail

SNP ID Chromosome Location
rs748241169 Chr.12: 50111046-50111046 on Build GRCh38
rs1045222277 Chr.12: 50110907-50110907 on Build GRCh38
rs954428351 Chr.12: 50111112-50111112 on Build GRCh38
rs893784034 Chr.12: 50111296-50111296 on Build GRCh38
rs923057873 Chr.12: 50111084-50111084 on Build GRCh38
rs954579256 Chr.12: 50110967-50110967 on Build GRCh38
rs760941046 Chr.12: 50111401-50111401 on Build GRCh38
rs905382034 Chr.12: 50111287-50111287 on Build GRCh38
rs919163143 Chr.12: 50111073-50111073 on Build GRCh38
rs770421668 Chr.12: 50111211-50111212 on Build GRCh38
rs985940825 Chr.12: 50110969-50110969 on Build GRCh38
rs1013755245 Chr.12: 50110906-50110906 on Build GRCh38
rs895285941 Chr.12: 50110913-50110913 on Build GRCh38
rs115532318 Chr.12: 50111263-50111263 on Build GRCh38
rs1032576993 Chr.12: 50110929-50110929 on Build GRCh38
rs562697795 Chr.12: 50111366-50111366 on Build GRCh38
rs562021620 Chr.12: 50111380-50111380 on Build GRCh38
rs983080967 Chr.12: 50111136-50111136 on Build GRCh38
rs999563618 Chr.12: 50110946-50110946 on Build GRCh38
rs892707579 Chr.12: 50111323-50111323 on Build GRCh38
rs1030120740 Chr.12: 50110949-50110949 on Build GRCh38
rs540475457 Chr.12: 50110878-50110878 on Build GRCh38
rs963218321 Chr.12: 50111390-50111390 on Build GRCh38
rs956427939 Chr.12: 50110930-50110930 on Build GRCh38
rs545884674 Chr.12: 50111348-50111348 on Build GRCh38
rs78070861 Chr.12: 50111139-50111139 on Build GRCh38
rs144024920 Chr.12: 50110922-50110922 on Build GRCh38
rs558750445 Chr.12: 50111049-50111049 on Build GRCh38
rs185524044 Chr.12: 50111032-50111032 on Build GRCh38
rs949468279 Chr.12: 50111239-50111239 on Build GRCh38
rs566967283 Chr.12: 50110966-50110966 on Build GRCh38
rs955945053 Chr.12: 50110975-50110975 on Build GRCh38
rs531517000 Chr.12: 50111367-50111367 on Build GRCh38
rs1052455446 Chr.12: 50111291-50111291 on Build GRCh38
rs964603786 Chr.12: 50111017-50111017 on Build GRCh38
rs974963717 Chr.12: 50111050-50111050 on Build GRCh38
rs569087930 Chr.12: 50111110-50111110 on Build GRCh38
rs1007287053 Chr.12: 50111376-50111376 on Build GRCh38
rs574394824 Chr.12: 50111194-50111194 on Build GRCh38
rs558585513 Chr.12: 50111288-50111288 on Build GRCh38
rs542183223 Chr.12: 50111375-50111375 on Build GRCh38
rs930033932 Chr.12: 50111091-50111091 on Build GRCh38
rs1001021106 Chr.12: 50111290-50111290 on Build GRCh38
rs55975153 Chr.12: 50111223-50111223 on Build GRCh38
rs771044884 Chr.12: 50110931-50110931 on Build GRCh38
rs900408962 Chr.12: 50111394-50111394 on Build GRCh38
rs554834199 Chr.12: 50111177-50111177 on Build GRCh38
rs906624787 Chr.12: 50111271-50111271 on Build GRCh38
rs78153953 Chr.12: 50111297-50111297 on Build GRCh38
rs916796991 Chr.12: 50111242-50111242 on Build GRCh38
rs1009236931 Chr.12: 50111334-50111334 on Build GRCh38
rs910363186 Chr.12: 50110994-50110994 on Build GRCh38
rs948284122 Chr.12: 50111255-50111255 on Build GRCh38
rs896561709 Chr.12: 50110896-50110896 on Build GRCh38
rs776360953 Chr.12: 50111404-50111410 on Build GRCh38
rs773592374 Chr.12: 50111388-50111388 on Build GRCh38
rs1001091099 Chr.12: 50110927-50110927 on Build GRCh38
rs1011469422 Chr.12: 50110983-50110983 on Build GRCh38

View Full Product Details