Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32208093-32208663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCC
Entrez Gene ID
55721
Gene Name
IQ motif containing C
Gene Aliases
-
UniGene
Hs.274356
Gene Chromosome Location
Chr.1: 32205635-32208687 on Build GRCh38
Transcripts
NM_001160042.1 NM_018134.2 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001160042.1 NM_018134.2 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

SNP Detail

SNP ID Chromosome Location
rs930750015 Chr.1: 32208144-32208144 on Build GRCh38
rs377225080 Chr.1: 32208576-32208576 on Build GRCh38
rs886310808 Chr.1: 32208200-32208200 on Build GRCh38
rs41302746 Chr.1: 32208265-32208265 on Build GRCh38
rs760736648 Chr.1: 32208321-32208321 on Build GRCh38
rs994716142 Chr.1: 32208253-32208253 on Build GRCh38
rs939142701 Chr.1: 32208460-32208460 on Build GRCh38
rs558904316 Chr.1: 32208315-32208315 on Build GRCh38
rs565632927 Chr.1: 32208562-32208562 on Build GRCh38
rs534352550 Chr.1: 32208410-32208410 on Build GRCh38
rs939275480 Chr.1: 32208211-32208211 on Build GRCh38
rs952159929 Chr.1: 32208370-32208370 on Build GRCh38
rs763673055 Chr.1: 32208418-32208418 on Build GRCh38
rs920648632 Chr.1: 32208114-32208114 on Build GRCh38
rs920615536 Chr.1: 32208367-32208367 on Build GRCh38
rs900416176 Chr.1: 32208201-32208201 on Build GRCh38
rs541645680 Chr.1: 32208369-32208369 on Build GRCh38
rs1056277299 Chr.1: 32208214-32208214 on Build GRCh38
rs1009039658 Chr.1: 32208593-32208593 on Build GRCh38
rs886242797 Chr.1: 32208293-32208293 on Build GRCh38
rs948003858 Chr.1: 32208496-32208496 on Build GRCh38
rs1021931860 Chr.1: 32208236-32208236 on Build GRCh38
rs999205228 Chr.1: 32208523-32208523 on Build GRCh38
rs368933605 Chr.1: 32208190-32208190 on Build GRCh38
rs753346895 Chr.1: 32208518-32208518 on Build GRCh38
rs1009069899 Chr.1: 32208301-32208301 on Build GRCh38
rs1005140258 Chr.1: 32208310-32208310 on Build GRCh38
rs974727240 Chr.1: 32208344-32208344 on Build GRCh38
rs750103598 Chr.1: 32208628-32208628 on Build GRCh38
rs942702708 Chr.1: 32208149-32208149 on Build GRCh38
rs186977369 Chr.1: 32208477-32208477 on Build GRCh38
rs1047538248 Chr.1: 32208171-32208171 on Build GRCh38
rs868638897 Chr.1: 32208486-32208486 on Build GRCh38
rs778358812 Chr.1: 32208597-32208597 on Build GRCh38
rs181951464 Chr.1: 32208329-32208329 on Build GRCh38
rs924208747 Chr.1: 32208603-32208604 on Build GRCh38
rs191466560 Chr.1: 32208497-32208497 on Build GRCh38
rs956366041 Chr.1: 32208300-32208300 on Build GRCh38
rs563220918 Chr.1: 32208411-32208411 on Build GRCh38
rs895069755 Chr.1: 32208226-32208226 on Build GRCh38
rs1011845284 Chr.1: 32208232-32208232 on Build GRCh38
rs916470594 Chr.1: 32208481-32208481 on Build GRCh38
rs964946762 Chr.1: 32208612-32208612 on Build GRCh38
rs1034759437 Chr.1: 32208320-32208320 on Build GRCh38
rs530747775 Chr.1: 32208464-32208464 on Build GRCh38
rs1039760138 Chr.1: 32208191-32208191 on Build GRCh38
rs956871992 Chr.1: 32208635-32208635 on Build GRCh38
rs1011909246 Chr.1: 32208456-32208456 on Build GRCh38
rs555280358 Chr.1: 32208289-32208289 on Build GRCh38
rs903590783 Chr.1: 32208255-32208255 on Build GRCh38
rs1031839793 Chr.1: 32208280-32208280 on Build GRCh38
rs903558449 Chr.1: 32208519-32208519 on Build GRCh38

View Full Product Details