Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46041841-46042439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R3
Entrez Gene ID
8503
Gene Name
phosphoinositide-3-kinase regulatory subunit 3
Gene Aliases
p55, p55-GAMMA, p55PIK
UniGene
Hs.655387
Gene Chromosome Location
Chr.1: 46040140-46176495 on Build GRCh38
Transcripts
NM_001114172.1 NM_001303427.1 NM_001303428.1 NM_001303429.1 NM_003629.3 XM_017002614.1

SNP Detail

SNP ID Chromosome Location
rs963082879 Chr.1: 46042316-46042316 on Build GRCh38
rs574440818 Chr.1: 46041987-46041987 on Build GRCh38
rs753557871 Chr.1: 46042310-46042310 on Build GRCh38
rs1007478291 Chr.1: 46041952-46041952 on Build GRCh38
rs912723747 Chr.1: 46042102-46042102 on Build GRCh38
rs561844328 Chr.1: 46042306-46042306 on Build GRCh38
rs950347003 Chr.1: 46042365-46042365 on Build GRCh38
rs1009767820 Chr.1: 46042000-46042000 on Build GRCh38
rs112591587 Chr.1: 46042326-46042326 on Build GRCh38
rs1040227199 Chr.1: 46042184-46042184 on Build GRCh38
rs1043093495 Chr.1: 46042209-46042209 on Build GRCh38
rs933808177 Chr.1: 46042251-46042251 on Build GRCh38
rs555886041 Chr.1: 46041961-46041961 on Build GRCh38
rs959862277 Chr.1: 46041912-46041912 on Build GRCh38
rs991656886 Chr.1: 46041917-46041917 on Build GRCh38
rs999181861 Chr.1: 46042273-46042273 on Build GRCh38
rs12746446 Chr.1: 46042181-46042181 on Build GRCh38
rs998744323 Chr.1: 46041882-46041882 on Build GRCh38
rs879710063 Chr.1: 46041908-46041908 on Build GRCh38
rs778223236 Chr.1: 46042018-46042018 on Build GRCh38
rs903162343 Chr.1: 46042237-46042237 on Build GRCh38
rs889125672 Chr.1: 46041866-46041866 on Build GRCh38
rs1051802567 Chr.1: 46042298-46042298 on Build GRCh38
rs564340940 Chr.1: 46042157-46042157 on Build GRCh38
rs983784264 Chr.1: 46042168-46042168 on Build GRCh38
rs1035951828 Chr.1: 46041907-46041907 on Build GRCh38
rs11548677 Chr.1: 46041996-46041996 on Build GRCh38
rs1021523740 Chr.1: 46042024-46042024 on Build GRCh38
rs547409569 Chr.1: 46042336-46042336 on Build GRCh38
rs543654704 Chr.1: 46042214-46042214 on Build GRCh38
rs903089302 Chr.1: 46041875-46041875 on Build GRCh38
rs749860628 Chr.1: 46042193-46042193 on Build GRCh38
rs981870524 Chr.1: 46042123-46042123 on Build GRCh38
rs545659326 Chr.1: 46042098-46042098 on Build GRCh38
rs28435505 Chr.1: 46042187-46042187 on Build GRCh38
rs1022707471 Chr.1: 46041960-46041960 on Build GRCh38
rs1033395392 Chr.1: 46042133-46042133 on Build GRCh38
rs770246244 Chr.1: 46042012-46042016 on Build GRCh38
rs895675206 Chr.1: 46042291-46042291 on Build GRCh38
rs556233048 Chr.1: 46042031-46042031 on Build GRCh38
rs754538324 Chr.1: 46042312-46042312 on Build GRCh38
rs779339600 Chr.1: 46042358-46042358 on Build GRCh38
rs572194138 Chr.1: 46042073-46042073 on Build GRCh38
rs983445447 Chr.1: 46042378-46042378 on Build GRCh38
rs766172663 Chr.1: 46042272-46042272 on Build GRCh38
rs192331848 Chr.1: 46042021-46042021 on Build GRCh38
rs910635453 Chr.1: 46042299-46042299 on Build GRCh38
rs867699175 Chr.1: 46042349-46042349 on Build GRCh38
rs1057014925 Chr.1: 46042280-46042280 on Build GRCh38

View Full Product Details