Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36775486-36776059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT27
Entrez Gene ID
11020
Gene Name
intraflagellar transport 27
Gene Aliases
BBS19, RABL4, RAYL
UniGene
Hs.415172
Gene Chromosome Location
Chr.22: 36758202-36776133 on Build GRCh38
Transcripts
NM_001177701.2 NM_006860.4 NR_033531.2 XM_006724106.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532562 c.21A>T p.K7N Chr.22: 36775687-36775687 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780431589 Chr.22: 36775740-36775740 on Build GRCh38
rs1031125378 Chr.22: 36775758-36775758 on Build GRCh38
rs928347329 Chr.22: 36775592-36775592 on Build GRCh38
rs773063905 Chr.22: 36775691-36775691 on Build GRCh38
rs898969171 Chr.22: 36776013-36776013 on Build GRCh38
rs933277696 Chr.22: 36775846-36775846 on Build GRCh38
rs759207767 Chr.22: 36775716-36775716 on Build GRCh38
rs756749200 Chr.22: 36775731-36775731 on Build GRCh38
rs938423263 Chr.22: 36775593-36775593 on Build GRCh38
rs755388616 Chr.22: 36775754-36775754 on Build GRCh38
rs1034477683 Chr.22: 36776035-36776035 on Build GRCh38
rs750955002 Chr.22: 36775726-36775726 on Build GRCh38
rs773558648 Chr.22: 36776034-36776034 on Build GRCh38
rs1023926309 Chr.22: 36775564-36775564 on Build GRCh38
rs925388420 Chr.22: 36775814-36775814 on Build GRCh38
rs867141279 Chr.22: 36775888-36775888 on Build GRCh38
rs200296902 Chr.22: 36775656-36775656 on Build GRCh38
rs749520432 Chr.22: 36775657-36775657 on Build GRCh38
rs183375553 Chr.22: 36775681-36775681 on Build GRCh38
rs563140347 Chr.22: 36775835-36775835 on Build GRCh38
rs992438410 Chr.22: 36775915-36775915 on Build GRCh38
rs757917955 Chr.22: 36775720-36775720 on Build GRCh38
rs202199880 Chr.22: 36775650-36775650 on Build GRCh38
rs1011105053 Chr.22: 36775824-36775824 on Build GRCh38
rs765570338 Chr.22: 36775831-36775831 on Build GRCh38
rs2009667 Chr.22: 36775567-36775567 on Build GRCh38
rs960947639 Chr.22: 36775911-36775911 on Build GRCh38
rs749659995 Chr.22: 36775741-36775741 on Build GRCh38
rs921819475 Chr.22: 36775925-36775925 on Build GRCh38
rs117576168 Chr.22: 36775713-36775713 on Build GRCh38
rs973284504 Chr.22: 36775909-36775909 on Build GRCh38
rs542596060 Chr.22: 36775853-36775853 on Build GRCh38
rs34451817 Chr.22: 36775924-36775925 on Build GRCh38
rs979737191 Chr.22: 36775857-36775857 on Build GRCh38
rs369610277 Chr.22: 36775749-36775749 on Build GRCh38
rs766603222 Chr.22: 36775523-36775523 on Build GRCh38
rs969649704 Chr.22: 36775852-36775852 on Build GRCh38
rs768800449 Chr.22: 36775669-36775669 on Build GRCh38
rs1004669382 Chr.22: 36775637-36775637 on Build GRCh38
rs983601726 Chr.22: 36775898-36775898 on Build GRCh38
rs879197195 Chr.22: 36775834-36775834 on Build GRCh38
rs375616019 Chr.22: 36775600-36775600 on Build GRCh38
rs770175816 Chr.22: 36775811-36775811 on Build GRCh38
rs112315007 Chr.22: 36775751-36775751 on Build GRCh38
rs528112600 Chr.22: 36775924-36775924 on Build GRCh38
rs963145255 Chr.22: 36775660-36775660 on Build GRCh38
rs372071343 Chr.22: 36775641-36775641 on Build GRCh38
rs373565282 Chr.22: 36775907-36775907 on Build GRCh38
rs907648494 Chr.22: 36775898-36775898 on Build GRCh38
rs12167741 Chr.22: 36775645-36775645 on Build GRCh38
rs924153419 Chr.22: 36775905-36775905 on Build GRCh38
rs763713964 Chr.22: 36775725-36775725 on Build GRCh38
rs1037477371 Chr.22: 36775996-36775996 on Build GRCh38
rs931964336 Chr.22: 36775675-36775675 on Build GRCh38
rs918136681 Chr.22: 36775760-36775760 on Build GRCh38
rs748172586 Chr.22: 36775682-36775682 on Build GRCh38
rs959736731 Chr.22: 36775893-36775893 on Build GRCh38
rs893958395 Chr.22: 36775797-36775797 on Build GRCh38
rs143672566 Chr.22: 36775844-36775844 on Build GRCh38
rs966908882 Chr.22: 36775780-36775780 on Build GRCh38
rs1004700443 Chr.22: 36775992-36775992 on Build GRCh38
rs1051597507 Chr.22: 36775594-36775594 on Build GRCh38
rs764751587 Chr.22: 36775718-36775718 on Build GRCh38
rs369770720 Chr.22: 36775715-36775715 on Build GRCh38
rs536206116 Chr.22: 36775568-36775568 on Build GRCh38
rs886349007 Chr.22: 36775961-36775961 on Build GRCh38
rs931911899 Chr.22: 36775926-36775926 on Build GRCh38
rs1035362490 Chr.22: 36775872-36775872 on Build GRCh38
rs113426880 Chr.22: 36775522-36775522 on Build GRCh38
rs935369519 Chr.22: 36775993-36775993 on Build GRCh38
rs546807961 Chr.22: 36775974-36775974 on Build GRCh38
rs528942552 Chr.22: 36775575-36775575 on Build GRCh38
rs112886277 Chr.22: 36775973-36775973 on Build GRCh38
rs941910958 Chr.22: 36775632-36775632 on Build GRCh38
rs1051671376 Chr.22: 36775938-36775938 on Build GRCh38
rs143443220 Chr.22: 36775708-36775708 on Build GRCh38
rs916714996 Chr.22: 36775920-36775920 on Build GRCh38
rs772166556 Chr.22: 36775688-36775688 on Build GRCh38
rs1052936053 Chr.22: 36776032-36776032 on Build GRCh38

View Full Product Details