Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25791182-25791728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXSM
Entrez Gene ID
54995
Gene Name
3-oxoacyl-ACP synthase, mitochondrial
Gene Aliases
FASN2D, KASI, KS
UniGene
Hs.55781
Gene Chromosome Location
Chr.3: 25783661-25794534 on Build GRCh38
Transcripts
NM_001145391.1 NM_017897.2 NR_026937.1 XM_006713216.3 XM_006713217.3 XM_017006713.1 XM_017006714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2948885 c.613G>A p.V205I Chr.3: 25791633-25791633 on Build GRCh38
4931907 c.598G>T p.G200C Chr.3: 25791618-25791618 on Build GRCh38
6736376 c.593T>C p.L198P Chr.3: 25791613-25791613 on Build GRCh38
4847726 c.656C>G p.S219* Chr.3: 25791676-25791676 on Build GRCh38
6736375 c.466C>T p.P156S Chr.3: 25791486-25791486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756628632 Chr.3: 25791333-25791333 on Build GRCh38
rs565892479 Chr.3: 25791657-25791657 on Build GRCh38
rs376430364 Chr.3: 25791690-25791690 on Build GRCh38
rs780784598 Chr.3: 25791372-25791372 on Build GRCh38
rs770479217 Chr.3: 25791569-25791569 on Build GRCh38
rs766556779 Chr.3: 25791631-25791631 on Build GRCh38
rs983317793 Chr.3: 25791666-25791666 on Build GRCh38
rs774084673 Chr.3: 25791449-25791449 on Build GRCh38
rs772652094 Chr.3: 25791276-25791276 on Build GRCh38
rs901062791 Chr.3: 25791602-25791602 on Build GRCh38
rs367918208 Chr.3: 25791345-25791345 on Build GRCh38
rs755879820 Chr.3: 25791537-25791537 on Build GRCh38
rs139719761 Chr.3: 25791560-25791560 on Build GRCh38
rs769298430 Chr.3: 25791275-25791275 on Build GRCh38
rs753248444 Chr.3: 25791329-25791329 on Build GRCh38
rs773852165 Chr.3: 25791296-25791296 on Build GRCh38
rs111964113 Chr.3: 25791561-25791561 on Build GRCh38
rs745408317 Chr.3: 25791662-25791662 on Build GRCh38
rs765195571 Chr.3: 25791504-25791504 on Build GRCh38
rs770296939 Chr.3: 25791399-25791399 on Build GRCh38
rs150601765 Chr.3: 25791456-25791456 on Build GRCh38
rs552836917 Chr.3: 25791626-25791626 on Build GRCh38
rs777709229 Chr.3: 25791556-25791556 on Build GRCh38
rs890910019 Chr.3: 25791221-25791221 on Build GRCh38
rs780689675 Chr.3: 25791226-25791227 on Build GRCh38
rs767733189 Chr.3: 25791319-25791319 on Build GRCh38
rs372167890 Chr.3: 25791614-25791614 on Build GRCh38
rs147184138 Chr.3: 25791603-25791603 on Build GRCh38
rs372030404 Chr.3: 25791687-25791687 on Build GRCh38
rs890161265 Chr.3: 25791524-25791524 on Build GRCh38
rs775772977 Chr.3: 25791467-25791467 on Build GRCh38
rs757383964 Chr.3: 25791341-25791341 on Build GRCh38
rs777478186 Chr.3: 25791643-25791643 on Build GRCh38
rs771686624 Chr.3: 25791588-25791588 on Build GRCh38
rs745366360 Chr.3: 25791580-25791580 on Build GRCh38
rs768318040 Chr.3: 25791268-25791268 on Build GRCh38
rs755933142 Chr.3: 25791641-25791641 on Build GRCh38
rs1035805769 Chr.3: 25791469-25791469 on Build GRCh38
rs754040484 Chr.3: 25791337-25791337 on Build GRCh38
rs775254154 Chr.3: 25791598-25791598 on Build GRCh38
rs769783256 Chr.3: 25791388-25791388 on Build GRCh38
rs759967237 Chr.3: 25791299-25791299 on Build GRCh38
rs949475573 Chr.3: 25791392-25791392 on Build GRCh38
rs769399379 Chr.3: 25791523-25791523 on Build GRCh38
rs748092794 Chr.3: 25791378-25791378 on Build GRCh38
rs963384566 Chr.3: 25791628-25791628 on Build GRCh38
rs746032938 Chr.3: 25791355-25791355 on Build GRCh38
rs747995353 Chr.3: 25791272-25791272 on Build GRCh38
rs1046913530 Chr.3: 25791209-25791209 on Build GRCh38
rs893750825 Chr.3: 25791404-25791404 on Build GRCh38
rs761113265 Chr.3: 25791471-25791471 on Build GRCh38
rs991552536 Chr.3: 25791586-25791586 on Build GRCh38
rs769097304 Chr.3: 25791604-25791604 on Build GRCh38
rs1046349441 Chr.3: 25791393-25791393 on Build GRCh38
rs764740644 Chr.3: 25791483-25791483 on Build GRCh38
rs753096401 Chr.3: 25791226-25791226 on Build GRCh38
rs201690414 Chr.3: 25791240-25791240 on Build GRCh38
rs1020803238 Chr.3: 25791515-25791515 on Build GRCh38
rs778224046 Chr.3: 25791574-25791574 on Build GRCh38
rs751729784 Chr.3: 25791633-25791633 on Build GRCh38
rs908598417 Chr.3: 25791381-25791381 on Build GRCh38
rs142653321 Chr.3: 25791262-25791262 on Build GRCh38
rs202016313 Chr.3: 25791222-25791222 on Build GRCh38
rs558426183 Chr.3: 25791395-25791395 on Build GRCh38
rs752557791 Chr.3: 25791528-25791528 on Build GRCh38
rs770363947 Chr.3: 25791611-25791611 on Build GRCh38
rs754377713 Chr.3: 25791484-25791484 on Build GRCh38
rs777929634 Chr.3: 25791239-25791239 on Build GRCh38
rs929865831 Chr.3: 25791668-25791668 on Build GRCh38
rs773100693 Chr.3: 25791394-25791394 on Build GRCh38
rs999000662 Chr.3: 25791421-25791421 on Build GRCh38
rs189087941 Chr.3: 25791624-25791624 on Build GRCh38
rs1009783957 Chr.3: 25791405-25791405 on Build GRCh38
rs747586021 Chr.3: 25791279-25791279 on Build GRCh38
rs150811011 Chr.3: 25791346-25791346 on Build GRCh38
rs764681649 Chr.3: 25791335-25791335 on Build GRCh38
rs375858422 Chr.3: 25791265-25791265 on Build GRCh38
rs746624121 Chr.3: 25791699-25791699 on Build GRCh38
rs754833833 Chr.3: 25791358-25791358 on Build GRCh38
rs757195424 Chr.3: 25791656-25791656 on Build GRCh38
rs756289273 Chr.3: 25791238-25791238 on Build GRCh38
rs780310939 Chr.3: 25791517-25791517 on Build GRCh38
rs543480319 Chr.3: 25791508-25791508 on Build GRCh38
rs766058410 Chr.3: 25791291-25791291 on Build GRCh38
rs561407254 Chr.3: 25791460-25791460 on Build GRCh38
rs919865897 Chr.3: 25791665-25791665 on Build GRCh38
rs762438629 Chr.3: 25791288-25791288 on Build GRCh38
rs1012250425 Chr.3: 25791338-25791338 on Build GRCh38
rs143066680 Chr.3: 25791270-25791270 on Build GRCh38
rs1008658789 Chr.3: 25791543-25791543 on Build GRCh38
rs763086339 Chr.3: 25791630-25791630 on Build GRCh38
rs938055145 Chr.3: 25791398-25791398 on Build GRCh38
rs201522040 Chr.3: 25791487-25791487 on Build GRCh38
rs753783337 Chr.3: 25791653-25791653 on Build GRCh38
rs757609014 Chr.3: 25791260-25791260 on Build GRCh38
rs572272317 Chr.3: 25791439-25791439 on Build GRCh38
rs758493035 Chr.3: 25791516-25791516 on Build GRCh38

View Full Product Details