Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 62222249-62222842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT2
Entrez Gene ID
10678
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2
Gene Aliases
3-Gn-T1, 3-Gn-T2, B3GN-T2, B3GNT, B3GNT-2, B3GNT1, BETA3GNT, BGNT2, BGnT-2, beta-1, beta3Gn-T1, beta3Gn-T2
UniGene
Hs.173203
Gene Chromosome Location
Chr.2: 62196127-62224731 on Build GRCh38
Transcripts
NM_001319075.1 NM_006577.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609305 c.182G>A p.R61Q Chr.2: 62222402-62222402 on Build GRCh38
4517972 c.483_484CC>TT p.R162W Chr.2: 62222703-62222704 on Build GRCh38
6609309 c.202C>T p.R68W Chr.2: 62222422-62222422 on Build GRCh38
4979968 c.594G>A p.L198L Chr.2: 62222814-62222814 on Build GRCh38
6609307 c.165C>A p.P55P Chr.2: 62222385-62222385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200035804 Chr.2: 62222401-62222401 on Build GRCh38
rs754971906 Chr.2: 62222300-62222300 on Build GRCh38
rs537607171 Chr.2: 62222727-62222727 on Build GRCh38
rs35622576 Chr.2: 62222733-62222734 on Build GRCh38
rs913095459 Chr.2: 62222720-62222720 on Build GRCh38
rs76769447 Chr.2: 62222658-62222658 on Build GRCh38
rs767012986 Chr.2: 62222753-62222753 on Build GRCh38
rs573730013 Chr.2: 62222745-62222745 on Build GRCh38
rs772113870 Chr.2: 62222595-62222595 on Build GRCh38
rs770122947 Chr.2: 62222478-62222478 on Build GRCh38
rs199523056 Chr.2: 62222582-62222582 on Build GRCh38
rs773173323 Chr.2: 62222679-62222679 on Build GRCh38
rs763016470 Chr.2: 62222391-62222391 on Build GRCh38
rs72893877 Chr.2: 62222529-62222529 on Build GRCh38
rs764153622 Chr.2: 62222600-62222600 on Build GRCh38
rs760222661 Chr.2: 62222775-62222775 on Build GRCh38
rs777861571 Chr.2: 62222812-62222812 on Build GRCh38
rs369888496 Chr.2: 62222577-62222577 on Build GRCh38
rs151141269 Chr.2: 62222279-62222279 on Build GRCh38
rs775458740 Chr.2: 62222619-62222619 on Build GRCh38
rs925776577 Chr.2: 62222441-62222441 on Build GRCh38
rs765061417 Chr.2: 62222614-62222614 on Build GRCh38
rs760937734 Chr.2: 62222580-62222580 on Build GRCh38
rs756441040 Chr.2: 62222558-62222558 on Build GRCh38
rs756777348 Chr.2: 62222627-62222627 on Build GRCh38
rs750648231 Chr.2: 62222511-62222511 on Build GRCh38
rs748225022 Chr.2: 62222588-62222588 on Build GRCh38
rs760273324 Chr.2: 62222548-62222556 on Build GRCh38
rs980888736 Chr.2: 62222364-62222364 on Build GRCh38
rs761889404 Chr.2: 62222497-62222497 on Build GRCh38
rs535480021 Chr.2: 62222624-62222624 on Build GRCh38
rs937006976 Chr.2: 62222810-62222810 on Build GRCh38
rs202087907 Chr.2: 62222631-62222631 on Build GRCh38
rs138422692 Chr.2: 62222381-62222381 on Build GRCh38
rs778882641 Chr.2: 62222429-62222429 on Build GRCh38
rs377129789 Chr.2: 62222796-62222796 on Build GRCh38
rs746222194 Chr.2: 62222476-62222476 on Build GRCh38
rs200219699 Chr.2: 62222297-62222297 on Build GRCh38
rs755325938 Chr.2: 62222780-62222780 on Build GRCh38
rs141787237 Chr.2: 62222446-62222446 on Build GRCh38
rs1039816222 Chr.2: 62222818-62222818 on Build GRCh38
rs749920688 Chr.2: 62222774-62222774 on Build GRCh38
rs765186281 Chr.2: 62222526-62222526 on Build GRCh38
rs780850209 Chr.2: 62222752-62222752 on Build GRCh38
rs749110175 Chr.2: 62222299-62222299 on Build GRCh38
rs139589693 Chr.2: 62222471-62222471 on Build GRCh38
rs745565189 Chr.2: 62222549-62222549 on Build GRCh38
rs563186939 Chr.2: 62222449-62222449 on Build GRCh38
rs751721470 Chr.2: 62222415-62222415 on Build GRCh38
rs148241288 Chr.2: 62222781-62222781 on Build GRCh38
rs771751289 Chr.2: 62222337-62222337 on Build GRCh38
rs768857235 Chr.2: 62222568-62222568 on Build GRCh38
rs777380168 Chr.2: 62222531-62222531 on Build GRCh38
rs774443407 Chr.2: 62222490-62222490 on Build GRCh38
rs771301412 Chr.2: 62222704-62222704 on Build GRCh38
rs1026753717 Chr.2: 62222512-62222512 on Build GRCh38
rs140564887 Chr.2: 62222623-62222623 on Build GRCh38
rs771144883 Chr.2: 62222368-62222368 on Build GRCh38
rs34019499 Chr.2: 62222704-62222705 on Build GRCh38
rs747217983 Chr.2: 62222687-62222687 on Build GRCh38
rs755888058 Chr.2: 62222280-62222280 on Build GRCh38
rs780344242 Chr.2: 62222632-62222632 on Build GRCh38
rs751137645 Chr.2: 62222532-62222532 on Build GRCh38
rs147249508 Chr.2: 62222385-62222385 on Build GRCh38
rs921516752 Chr.2: 62222718-62222718 on Build GRCh38
rs142907987 Chr.2: 62222730-62222730 on Build GRCh38
rs760823706 Chr.2: 62222599-62222599 on Build GRCh38
rs759375083 Chr.2: 62222518-62222518 on Build GRCh38
rs749628332 Chr.2: 62222649-62222649 on Build GRCh38
rs767753065 Chr.2: 62222509-62222509 on Build GRCh38
rs751492881 Chr.2: 62222625-62222625 on Build GRCh38
rs78968142 Chr.2: 62222628-62222628 on Build GRCh38
rs781363819 Chr.2: 62222472-62222472 on Build GRCh38
rs747635960 Chr.2: 62222321-62222321 on Build GRCh38
rs370583197 Chr.2: 62222797-62222797 on Build GRCh38
rs772134926 Chr.2: 62222677-62222677 on Build GRCh38
rs773474610 Chr.2: 62222596-62222596 on Build GRCh38
rs755371226 Chr.2: 62222650-62222650 on Build GRCh38
rs957166825 Chr.2: 62222661-62222661 on Build GRCh38
rs376025052 Chr.2: 62222426-62222426 on Build GRCh38
rs763462600 Chr.2: 62222484-62222484 on Build GRCh38
rs766045424 Chr.2: 62222422-62222422 on Build GRCh38
rs752626069 Chr.2: 62222528-62222528 on Build GRCh38
rs34012259 Chr.2: 62222294-62222295 on Build GRCh38
rs777567753 Chr.2: 62222341-62222341 on Build GRCh38
rs776017170 Chr.2: 62222386-62222386 on Build GRCh38
rs966314133 Chr.2: 62222567-62222567 on Build GRCh38
rs777061198 Chr.2: 62222377-62222377 on Build GRCh38
rs765982851 Chr.2: 62222779-62222779 on Build GRCh38
rs1005107278 Chr.2: 62222371-62222371 on Build GRCh38
rs138894863 Chr.2: 62222356-62222356 on Build GRCh38
rs917542505 Chr.2: 62222354-62222354 on Build GRCh38
rs775962401 Chr.2: 62222483-62222483 on Build GRCh38
rs374000666 Chr.2: 62222508-62222508 on Build GRCh38
rs752752682 Chr.2: 62222432-62222432 on Build GRCh38
rs767688621 Chr.2: 62222462-62222462 on Build GRCh38
rs530600752 Chr.2: 62222463-62222463 on Build GRCh38
rs780399465 Chr.2: 62222559-62222559 on Build GRCh38
rs768654582 Chr.2: 62222486-62222486 on Build GRCh38
rs778233916 Chr.2: 62222303-62222303 on Build GRCh38
rs201194611 Chr.2: 62222660-62222660 on Build GRCh38
rs749751253 Chr.2: 62222561-62222561 on Build GRCh38
rs747038487 Chr.2: 62222816-62222816 on Build GRCh38
rs377454562 Chr.2: 62222539-62222539 on Build GRCh38
rs144486810 Chr.2: 62222293-62222293 on Build GRCh38
rs370993124 Chr.2: 62222716-62222716 on Build GRCh38
rs200996349 Chr.2: 62222421-62222421 on Build GRCh38
rs780891543 Chr.2: 62222540-62222540 on Build GRCh38
rs149533514 Chr.2: 62222744-62222744 on Build GRCh38

View Full Product Details