Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65014331-65014880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA13
Entrez Gene ID
10672
Gene Name
G protein subunit alpha 13
Gene Aliases
G13
UniGene
Hs.515018
Gene Chromosome Location
Chr.17: 65009289-65056802 on Build GRCh38
Transcripts
NM_001282425.1 NM_006572.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
982989 c.778C>T p.R260* Chr.17: 65014613-65014613 on Build GRCh38
220594 c.1034A>C p.H345P Chr.17: 65014357-65014357 on Build GRCh38
4687372 c.634G>A p.E212K Chr.17: 65014757-65014757 on Build GRCh38
982987 c.946C>A p.P316T Chr.17: 65014445-65014445 on Build GRCh38
220590 c.875A>G p.K292R Chr.17: 65014516-65014516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984042342 Chr.17: 65014800-65014800 on Build GRCh38
rs747088179 Chr.17: 65014843-65014843 on Build GRCh38
rs551449071 Chr.17: 65014399-65014399 on Build GRCh38
rs563262712 Chr.17: 65014565-65014565 on Build GRCh38
rs1020629751 Chr.17: 65014745-65014745 on Build GRCh38
rs745917724 Chr.17: 65014603-65014603 on Build GRCh38
rs773072737 Chr.17: 65014664-65014664 on Build GRCh38
rs769688760 Chr.17: 65014493-65014493 on Build GRCh38
rs756057026 Chr.17: 65014855-65014855 on Build GRCh38
rs757715203 Chr.17: 65014390-65014390 on Build GRCh38
rs775043719 Chr.17: 65014746-65014746 on Build GRCh38
rs375578952 Chr.17: 65014543-65014543 on Build GRCh38
rs941409424 Chr.17: 65014461-65014461 on Build GRCh38
rs2230312 Chr.17: 65014752-65014752 on Build GRCh38
rs376356001 Chr.17: 65014412-65014412 on Build GRCh38
rs543576786 Chr.17: 65014536-65014536 on Build GRCh38
rs199660009 Chr.17: 65014555-65014555 on Build GRCh38
rs749049765 Chr.17: 65014679-65014679 on Build GRCh38
rs757910798 Chr.17: 65014428-65014428 on Build GRCh38
rs756503153 Chr.17: 65014441-65014441 on Build GRCh38
rs779924076 Chr.17: 65014535-65014535 on Build GRCh38
rs758704526 Chr.17: 65014804-65014804 on Build GRCh38
rs765670502 Chr.17: 65014780-65014780 on Build GRCh38
rs112783387 Chr.17: 65014674-65014674 on Build GRCh38
rs61754625 Chr.17: 65014590-65014590 on Build GRCh38
rs113503987 Chr.17: 65014562-65014562 on Build GRCh38
rs368514878 Chr.17: 65014491-65014491 on Build GRCh38
rs528941392 Chr.17: 65014601-65014601 on Build GRCh38
rs542259871 Chr.17: 65014604-65014604 on Build GRCh38
rs779049963 Chr.17: 65014460-65014460 on Build GRCh38
rs769918719 Chr.17: 65014646-65014646 on Build GRCh38
rs778151842 Chr.17: 65014443-65014443 on Build GRCh38
rs765950706 Chr.17: 65014529-65014529 on Build GRCh38
rs749493486 Chr.17: 65014445-65014445 on Build GRCh38
rs750066119 Chr.17: 65014387-65014387 on Build GRCh38
rs1037001980 Chr.17: 65014469-65014469 on Build GRCh38
rs754269916 Chr.17: 65014573-65014573 on Build GRCh38
rs754149325 Chr.17: 65014763-65014763 on Build GRCh38
rs764938914 Chr.17: 65014365-65014365 on Build GRCh38
rs957774501 Chr.17: 65014691-65014691 on Build GRCh38
rs766915568 Chr.17: 65014733-65014733 on Build GRCh38
rs773199144 Chr.17: 65014501-65014501 on Build GRCh38
rs759273182 Chr.17: 65014731-65014731 on Build GRCh38
rs528393244 Chr.17: 65014680-65014680 on Build GRCh38
rs750584124 Chr.17: 65014781-65014781 on Build GRCh38
rs745945642 Chr.17: 65014462-65014462 on Build GRCh38
rs533239233 Chr.17: 65014758-65014758 on Build GRCh38
rs780276984 Chr.17: 65014810-65014810 on Build GRCh38
rs757591251 Chr.17: 65014764-65014764 on Build GRCh38
rs778102560 Chr.17: 65014568-65014568 on Build GRCh38
rs780326315 Chr.17: 65014608-65014608 on Build GRCh38
rs753013543 Chr.17: 65014427-65014427 on Build GRCh38
rs751094630 Chr.17: 65014407-65014407 on Build GRCh38
rs765899545 Chr.17: 65014396-65014396 on Build GRCh38
rs373006520 Chr.17: 65014557-65014557 on Build GRCh38
rs376225936 Chr.17: 65014818-65014818 on Build GRCh38
rs1062597 Chr.17: 65014730-65014730 on Build GRCh38
rs768587363 Chr.17: 65014466-65014466 on Build GRCh38
rs201673014 Chr.17: 65014488-65014488 on Build GRCh38
rs1001695741 Chr.17: 65014566-65014566 on Build GRCh38
rs200032909 Chr.17: 65014400-65014400 on Build GRCh38
rs778424474 Chr.17: 65014839-65014839 on Build GRCh38
rs868396276 Chr.17: 65014619-65014619 on Build GRCh38
rs371081033 Chr.17: 65014518-65014518 on Build GRCh38
rs145562983 Chr.17: 65014452-65014452 on Build GRCh38
rs762741889 Chr.17: 65014391-65014391 on Build GRCh38
rs748083796 Chr.17: 65014645-65014645 on Build GRCh38

View Full Product Details