Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36555895-36556417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF529
Entrez Gene ID
57711
Gene Name
zinc finger protein 529
Gene Aliases
-
UniGene
Hs.654960
Gene Chromosome Location
Chr.19: 36534491-36605411 on Build GRCh38
Transcripts
NM_001145649.1 NM_001145650.1 NM_001321351.1 NM_020951.4 XM_006723302.3 XM_006723304.2 XM_011527164.2 XM_011527165.2 XM_011527166.2 XM_011527167.2 XM_011527169.2 XM_011527170.1 XM_017027039.1 XM_017027040.1 XM_017027041.1 XM_017027042.1 XM_017027043.1 XM_017027044.1 XM_017027045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
995507 c.29A>G p.H10R Chr.19: 36556183-36556183 on Build GRCh38
995505 c.29A>G p.H10R Chr.19: 36556183-36556183 on Build GRCh38
1393102 c.73C>T p.P25S Chr.19: 36556139-36556139 on Build GRCh38
1393101 c.73C>T p.P25S Chr.19: 36556139-36556139 on Build GRCh38
3388904 c.15-2A>G p.? Chr.19: 36556199-36556199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934182846 Chr.19: 36556135-36556135 on Build GRCh38
rs150333116 Chr.19: 36556298-36556298 on Build GRCh38
rs951708015 Chr.19: 36555945-36555945 on Build GRCh38
rs1051117803 Chr.19: 36556002-36556002 on Build GRCh38
rs995704912 Chr.19: 36556080-36556080 on Build GRCh38
rs557725841 Chr.19: 36556196-36556196 on Build GRCh38
rs971200384 Chr.19: 36556334-36556334 on Build GRCh38
rs138960313 Chr.19: 36556306-36556306 on Build GRCh38
rs780160827 Chr.19: 36556131-36556131 on Build GRCh38
rs778403820 Chr.19: 36556210-36556210 on Build GRCh38
rs572916104 Chr.19: 36556223-36556223 on Build GRCh38
rs183650399 Chr.19: 36556341-36556341 on Build GRCh38
rs777756916 Chr.19: 36556203-36556203 on Build GRCh38
rs990015473 Chr.19: 36556319-36556319 on Build GRCh38
rs546079791 Chr.19: 36556192-36556192 on Build GRCh38
rs986790157 Chr.19: 36556331-36556331 on Build GRCh38
rs573550464 Chr.19: 36555978-36555978 on Build GRCh38
rs971283936 Chr.19: 36556293-36556293 on Build GRCh38
rs1054881575 Chr.19: 36556261-36556261 on Build GRCh38
rs940295639 Chr.19: 36556012-36556012 on Build GRCh38
rs553126328 Chr.19: 36555946-36555946 on Build GRCh38
rs1041784200 Chr.19: 36556171-36556171 on Build GRCh38
rs898627533 Chr.19: 36556053-36556053 on Build GRCh38
rs141193291 Chr.19: 36556247-36556247 on Build GRCh38
rs191077147 Chr.19: 36556001-36556001 on Build GRCh38
rs1027793517 Chr.19: 36556090-36556090 on Build GRCh38
rs990605473 Chr.19: 36555982-36555982 on Build GRCh38
rs544453010 Chr.19: 36556335-36556335 on Build GRCh38
rs922753442 Chr.19: 36556128-36556128 on Build GRCh38
rs188063855 Chr.19: 36556370-36556370 on Build GRCh38
rs769813092 Chr.19: 36556318-36556318 on Build GRCh38
rs921328160 Chr.19: 36556320-36556320 on Build GRCh38
rs775775443 Chr.19: 36556086-36556086 on Build GRCh38
rs993295391 Chr.19: 36556375-36556375 on Build GRCh38
rs371354285 Chr.19: 36555964-36555964 on Build GRCh38
rs1031594693 Chr.19: 36556306-36556310 on Build GRCh38
rs932749904 Chr.19: 36556321-36556321 on Build GRCh38
rs185963997 Chr.19: 36555920-36555920 on Build GRCh38
rs746657704 Chr.19: 36555998-36555998 on Build GRCh38
rs924478919 Chr.19: 36555956-36555956 on Build GRCh38
rs912529047 Chr.19: 36556385-36556385 on Build GRCh38
rs575495745 Chr.19: 36556060-36556060 on Build GRCh38
rs776214137 Chr.19: 36556181-36556181 on Build GRCh38
rs77122130 Chr.19: 36556009-36556009 on Build GRCh38
rs1039979773 Chr.19: 36556125-36556125 on Build GRCh38
rs775272232 Chr.19: 36556371-36556371 on Build GRCh38
rs1037290875 Chr.19: 36556019-36556019 on Build GRCh38

View Full Product Details