Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133828685-133828989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB6B
Entrez Gene ID
51560
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.715344
Gene Chromosome Location
Chr.3: 133824235-133895847 on Build GRCh38
Transcripts
NM_016577.3 XM_011512893.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1693493 c.602C>T p.A201V Chr.3: 133828813-133828813 on Build GRCh38
7426349 c.595C>A p.P199T Chr.3: 133828820-133828820 on Build GRCh38
3759773 c.567C>T p.I189I Chr.3: 133828848-133828848 on Build GRCh38
3587670 c.620C>T p.S207F Chr.3: 133828795-133828795 on Build GRCh38
6761605 c.577C>G p.L193V Chr.3: 133828838-133828838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771112288 Chr.3: 133828775-133828775 on Build GRCh38
rs772722900 Chr.3: 133828897-133828897 on Build GRCh38
rs369228908 Chr.3: 133828744-133828744 on Build GRCh38
rs1026296297 Chr.3: 133828887-133828887 on Build GRCh38
rs749338361 Chr.3: 133828845-133828845 on Build GRCh38
rs774666750 Chr.3: 133828713-133828713 on Build GRCh38
rs771114953 Chr.3: 133828847-133828847 on Build GRCh38
rs762372552 Chr.3: 133828747-133828747 on Build GRCh38
rs202080888 Chr.3: 133828809-133828809 on Build GRCh38
rs748174713 Chr.3: 133828763-133828763 on Build GRCh38
rs373337169 Chr.3: 133828808-133828808 on Build GRCh38
rs749716053 Chr.3: 133828732-133828732 on Build GRCh38
rs145404929 Chr.3: 133828864-133828864 on Build GRCh38
rs761260243 Chr.3: 133828877-133828877 on Build GRCh38
rs759980284 Chr.3: 133828779-133828779 on Build GRCh38
rs949304456 Chr.3: 133828969-133828969 on Build GRCh38
rs373274918 Chr.3: 133828865-133828865 on Build GRCh38
rs200553268 Chr.3: 133828754-133828754 on Build GRCh38
rs553369916 Chr.3: 133828821-133828821 on Build GRCh38
rs949419368 Chr.3: 133828718-133828718 on Build GRCh38
rs990632276 Chr.3: 133828935-133828935 on Build GRCh38
rs536858692 Chr.3: 133828710-133828710 on Build GRCh38
rs915781944 Chr.3: 133828922-133828922 on Build GRCh38
rs2293372 Chr.3: 133828951-133828951 on Build GRCh38
rs769842460 Chr.3: 133828767-133828767 on Build GRCh38
rs925019615 Chr.3: 133828914-133828914 on Build GRCh38
rs768100530 Chr.3: 133828787-133828787 on Build GRCh38
rs1037002397 Chr.3: 133828759-133828759 on Build GRCh38
rs749591613 Chr.3: 133828906-133828906 on Build GRCh38
rs761317503 Chr.3: 133828802-133828802 on Build GRCh38
rs749969544 Chr.3: 133828803-133828803 on Build GRCh38
rs917835881 Chr.3: 133828944-133828945 on Build GRCh38
rs550352284 Chr.3: 133828720-133828720 on Build GRCh38
rs756660568 Chr.3: 133828817-133828817 on Build GRCh38
rs149351332 Chr.3: 133828795-133828795 on Build GRCh38
rs774717347 Chr.3: 133828778-133828778 on Build GRCh38
rs758028508 Chr.3: 133828743-133828743 on Build GRCh38
rs115194527 Chr.3: 133828899-133828899 on Build GRCh38
rs750071125 Chr.3: 133828737-133828737 on Build GRCh38
rs369390537 Chr.3: 133828815-133828815 on Build GRCh38
rs1020325020 Chr.3: 133828816-133828816 on Build GRCh38
rs1052690305 Chr.3: 133828714-133828714 on Build GRCh38
rs769101137 Chr.3: 133828879-133828879 on Build GRCh38
rs773203969 Chr.3: 133828771-133828771 on Build GRCh38
rs2293373 Chr.3: 133828848-133828848 on Build GRCh38
rs539333768 Chr.3: 133828818-133828818 on Build GRCh38
rs766006340 Chr.3: 133828900-133828900 on Build GRCh38
rs754789249 Chr.3: 133828812-133828812 on Build GRCh38
rs751189291 Chr.3: 133828901-133828901 on Build GRCh38
rs777891226 Chr.3: 133828839-133828839 on Build GRCh38
rs748045973 Chr.3: 133828820-133828820 on Build GRCh38
rs754852978 Chr.3: 133828750-133828750 on Build GRCh38
rs377316788 Chr.3: 133828915-133828915 on Build GRCh38

View Full Product Details