Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 152299986-152300506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBNL1
Entrez Gene ID
4154
Gene Name
muscleblind like splicing regulator 1
Gene Aliases
EXP, MBNL
UniGene
Hs.201858
Gene Chromosome Location
Chr.3: 152243656-152465780 on Build GRCh38
Transcripts
NM_001314057.1 NM_021038.4 NM_207292.2 NM_207293.1 NM_207294.1 NM_207295.1 NM_207296.1 NM_207297.1 XM_005247457.4 XM_005247458.4 XM_005247460.4 XM_005247461.4 XM_005247462.4 XM_005247463.4 XM_005247464.4 XM_005247465.4 XM_005247466.4 XM_005247467.4 XM_005247468.4 XM_005247471.4 XM_005247474.3 XM_005247475.3 XM_005247476.4 XM_005247477.4 XM_006713639.1 XM_011512843.2 XM_011512845.2 XM_011512846.2 XM_011512847.2 XM_011512848.2 XM_011512849.2 XM_017006423.1 XM_017006424.1 XM_017006425.1 XM_017006426.1 XM_017006427.1 XM_017006428.1 XM_017006429.1 XM_017006430.1 XM_017006431.1 XM_017006432.1 XM_017006433.1 XM_017006434.1 XM_017006435.1 XM_017006436.1 XM_017006437.1 XM_017006438.1 XM_017006439.1 XM_017006440.1 XM_017006441.1 XM_017006442.1 XM_017006443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6262310 c.7G>T p.V3F Chr.3: 152300200-152300200 on Build GRCh38
392231 c.41delT p.L14fs*37 Chr.3: 152300234-152300234 on Build GRCh38
1693624 c.158G>T p.C53F Chr.3: 152300351-152300351 on Build GRCh38
5708509 c.70_71insG p.T26fs*10 Chr.3: 152300263-152300264 on Build GRCh38
361253 c.130C>G p.Q44E Chr.3: 152300323-152300323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760116131 Chr.3: 152300210-152300210 on Build GRCh38
rs763828262 Chr.3: 152300143-152300143 on Build GRCh38
rs770696564 Chr.3: 152300278-152300278 on Build GRCh38
rs867524518 Chr.3: 152300235-152300235 on Build GRCh38
rs771223286 Chr.3: 152300030-152300030 on Build GRCh38
rs750463887 Chr.3: 152300177-152300177 on Build GRCh38
rs189258831 Chr.3: 152300045-152300045 on Build GRCh38
rs750601139 Chr.3: 152300379-152300379 on Build GRCh38
rs773981630 Chr.3: 152300280-152300280 on Build GRCh38
rs17368446 Chr.3: 152300434-152300434 on Build GRCh38
rs941257182 Chr.3: 152300463-152300463 on Build GRCh38
rs1001880076 Chr.3: 152300168-152300168 on Build GRCh38
rs765381450 Chr.3: 152300346-152300346 on Build GRCh38
rs755516336 Chr.3: 152300401-152300401 on Build GRCh38
rs960413604 Chr.3: 152300437-152300437 on Build GRCh38
rs971856430 Chr.3: 152300176-152300176 on Build GRCh38
rs757442403 Chr.3: 152300167-152300167 on Build GRCh38
rs763578642 Chr.3: 152300275-152300275 on Build GRCh38
rs971889006 Chr.3: 152300473-152300473 on Build GRCh38
rs769170921 Chr.3: 152300226-152300226 on Build GRCh38
rs933849756 Chr.3: 152300410-152300410 on Build GRCh38
rs764837742 Chr.3: 152300183-152300183 on Build GRCh38
rs927714800 Chr.3: 152300141-152300141 on Build GRCh38
rs185894411 Chr.3: 152300288-152300288 on Build GRCh38
rs370857002 Chr.3: 152300169-152300169 on Build GRCh38
rs141393481 Chr.3: 152300214-152300214 on Build GRCh38
rs766438725 Chr.3: 152300385-152300385 on Build GRCh38
rs370778181 Chr.3: 152300402-152300402 on Build GRCh38
rs376731460 Chr.3: 152300238-152300238 on Build GRCh38
rs753410588 Chr.3: 152300312-152300312 on Build GRCh38
rs747466168 Chr.3: 152300215-152300215 on Build GRCh38
rs933819462 Chr.3: 152300068-152300068 on Build GRCh38
rs980523097 Chr.3: 152300105-152300105 on Build GRCh38
rs758976742 Chr.3: 152300282-152300282 on Build GRCh38
rs758425774 Chr.3: 152300381-152300381 on Build GRCh38
rs748521232 Chr.3: 152300419-152300419 on Build GRCh38
rs767870615 Chr.3: 152300094-152300094 on Build GRCh38
rs1022960687 Chr.3: 152300378-152300378 on Build GRCh38
rs375041852 Chr.3: 152300199-152300199 on Build GRCh38
rs753615521 Chr.3: 152300339-152300339 on Build GRCh38
rs919056569 Chr.3: 152300484-152300484 on Build GRCh38
rs180834822 Chr.3: 152300083-152300083 on Build GRCh38
rs565783469 Chr.3: 152300386-152300386 on Build GRCh38
rs775976769 Chr.3: 152300133-152300133 on Build GRCh38
rs1021924098 Chr.3: 152300073-152300073 on Build GRCh38
rs919179667 Chr.3: 152300187-152300187 on Build GRCh38
rs775313136 Chr.3: 152300302-152300302 on Build GRCh38
rs758436405 Chr.3: 152300209-152300209 on Build GRCh38
rs969065151 Chr.3: 152300382-152300382 on Build GRCh38
rs200628264 Chr.3: 152300201-152300201 on Build GRCh38
rs942739473 Chr.3: 152300026-152300026 on Build GRCh38
rs750142332 Chr.3: 152300377-152300381 on Build GRCh38
rs1039573057 Chr.3: 152300038-152300038 on Build GRCh38
rs1037961420 Chr.3: 152300138-152300138 on Build GRCh38
rs960549915 Chr.3: 152300163-152300163 on Build GRCh38
rs368507124 Chr.3: 152300109-152300109 on Build GRCh38
rs555382989 Chr.3: 152300148-152300148 on Build GRCh38
rs956948999 Chr.3: 152300019-152300019 on Build GRCh38
rs909920090 Chr.3: 152300268-152300268 on Build GRCh38
rs202116667 Chr.3: 152300193-152300193 on Build GRCh38
rs1039837012 Chr.3: 152300277-152300277 on Build GRCh38
rs762088936 Chr.3: 152300343-152300343 on Build GRCh38
rs774554649 Chr.3: 152300032-152300032 on Build GRCh38
rs74497536 Chr.3: 152300313-152300313 on Build GRCh38
rs764968337 Chr.3: 152300333-152300333 on Build GRCh38

View Full Product Details