Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48304029-48304225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
UniGene
Hs.408528
Gene Chromosome Location
Chr.13: 48303747-48481890 on Build GRCh38
Transcripts
NM_000321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5685203 c.137G>A p.R46K Chr.13: 48304049-48304049 on Build GRCh38
19545 c.1_*del p.0 Chr.13: 48303913-48480071 on Build GRCh38
6911720 c.137+1G>C p.? Chr.13: 48304050-48304050 on Build GRCh38
6913560 c.137+2T>A p.? Chr.13: 48304051-48304051 on Build GRCh38
6969592 c.128_129delCT p.L44fs*4 Chr.13: 48304040-48304041 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546745714 Chr.13: 48304114-48304114 on Build GRCh38
rs1032172153 Chr.13: 48304081-48304081 on Build GRCh38
rs763972315 Chr.13: 48304064-48304064 on Build GRCh38
rs999325303 Chr.13: 48304068-48304068 on Build GRCh38
rs762766187 Chr.13: 48304062-48304062 on Build GRCh38
rs957863843 Chr.13: 48304091-48304091 on Build GRCh38
rs191994935 Chr.13: 48304143-48304143 on Build GRCh38
rs2252544 Chr.13: 48304156-48304156 on Build GRCh38
rs569133870 Chr.13: 48304161-48304161 on Build GRCh38
rs975089524 Chr.13: 48304105-48304105 on Build GRCh38
rs1028898036 Chr.13: 48304109-48304109 on Build GRCh38
rs868243630 Chr.13: 48304155-48304155 on Build GRCh38
rs574145040 Chr.13: 48304061-48304061 on Build GRCh38
rs940455223 Chr.13: 48304137-48304137 on Build GRCh38
rs773624735 Chr.13: 48304080-48304080 on Build GRCh38
rs2854351 Chr.13: 48304135-48304135 on Build GRCh38
rs937509494 Chr.13: 48304189-48304189 on Build GRCh38
rs987114647 Chr.13: 48304126-48304126 on Build GRCh38
rs1037709289 Chr.13: 48304096-48304096 on Build GRCh38
rs1007315913 Chr.13: 48304129-48304129 on Build GRCh38
rs961608512 Chr.13: 48304108-48304137 on Build GRCh38

View Full Product Details