Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1511990-1512480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3A
Entrez Gene ID
55210
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
-
UniGene
Hs.23413
Gene Chromosome Location
Chr.1: 1512143-1534687 on Build GRCh38
Transcripts
NM_001170535.1 NM_018188.3 XM_017001628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429320 c.108G>C p.G36G Chr.1: 1512376-1512376 on Build GRCh38
3688961 c.44G>A p.G15D Chr.1: 1512312-1512312 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs973316367 Chr.1: 1512217-1512217 on Build GRCh38
rs906901418 Chr.1: 1512235-1512235 on Build GRCh38
rs762349361 Chr.1: 1512270-1512270 on Build GRCh38
rs1004732117 Chr.1: 1512373-1512373 on Build GRCh38
rs547280903 Chr.1: 1512045-1512045 on Build GRCh38
rs1030637854 Chr.1: 1512083-1512083 on Build GRCh38
rs953298933 Chr.1: 1512459-1512459 on Build GRCh38
rs777769663 Chr.1: 1512336-1512336 on Build GRCh38
rs535854315 Chr.1: 1512121-1512121 on Build GRCh38
rs1045795660 Chr.1: 1512229-1512229 on Build GRCh38
rs550272997 Chr.1: 1512113-1512113 on Build GRCh38
rs940585674 Chr.1: 1512051-1512051 on Build GRCh38
rs564012773 Chr.1: 1512436-1512436 on Build GRCh38
rs561570704 Chr.1: 1512355-1512355 on Build GRCh38
rs980876413 Chr.1: 1512101-1512102 on Build GRCh38
rs770645672 Chr.1: 1512259-1512259 on Build GRCh38
rs867724166 Chr.1: 1512021-1512021 on Build GRCh38
rs1037943846 Chr.1: 1512058-1512058 on Build GRCh38
rs778779075 Chr.1: 1512150-1512150 on Build GRCh38
rs775436875 Chr.1: 1512259-1512260 on Build GRCh38
rs558471838 Chr.1: 1512204-1512204 on Build GRCh38
rs750610176 Chr.1: 1512222-1512222 on Build GRCh38
rs578231625 Chr.1: 1512230-1512230 on Build GRCh38
rs962726816 Chr.1: 1512216-1512216 on Build GRCh38
rs541906634 Chr.1: 1512313-1512313 on Build GRCh38
rs769862110 Chr.1: 1512249-1512251 on Build GRCh38
rs767886117 Chr.1: 1512219-1512219 on Build GRCh38
rs776278284 Chr.1: 1512260-1512260 on Build GRCh38
rs982886083 Chr.1: 1512208-1512208 on Build GRCh38
rs72468212 Chr.1: 1512170-1512170 on Build GRCh38
rs201314162 Chr.1: 1512287-1512287 on Build GRCh38
rs1057517687 Chr.1: 1512426-1512426 on Build GRCh38
rs2274435 Chr.1: 1512312-1512312 on Build GRCh38
rs944435980 Chr.1: 1512168-1512168 on Build GRCh38
rs560185737 Chr.1: 1512120-1512120 on Build GRCh38
rs748857128 Chr.1: 1512078-1512078 on Build GRCh38
rs908501559 Chr.1: 1512118-1512118 on Build GRCh38
rs931795726 Chr.1: 1512151-1512151 on Build GRCh38
rs1028244940 Chr.1: 1512447-1512447 on Build GRCh38
rs570348876 Chr.1: 1512097-1512097 on Build GRCh38
rs905453932 Chr.1: 1512324-1512324 on Build GRCh38
rs961079072 Chr.1: 1512205-1512205 on Build GRCh38
rs1050193092 Chr.1: 1512152-1512152 on Build GRCh38
rs758407786 Chr.1: 1512245-1512245 on Build GRCh38
rs749293250 Chr.1: 1512231-1512231 on Build GRCh38
rs757647963 Chr.1: 1512218-1512218 on Build GRCh38
rs892717394 Chr.1: 1512084-1512084 on Build GRCh38
rs1002420548 Chr.1: 1512349-1512349 on Build GRCh38
rs762276120 Chr.1: 1512283-1512283 on Build GRCh38
rs905421314 Chr.1: 1512200-1512200 on Build GRCh38
rs944493370 Chr.1: 1512296-1512296 on Build GRCh38
rs753971450 Chr.1: 1512319-1512319 on Build GRCh38
rs770716948 Chr.1: 1512013-1512013 on Build GRCh38
rs1050119026 Chr.1: 1512068-1512068 on Build GRCh38
rs746833842 Chr.1: 1512256-1512256 on Build GRCh38
rs1052533262 Chr.1: 1512169-1512169 on Build GRCh38
rs745576974 Chr.1: 1512261-1512261 on Build GRCh38
rs555425516 Chr.1: 1512127-1512127 on Build GRCh38
rs995479116 Chr.1: 1512399-1512399 on Build GRCh38
rs1035123193 Chr.1: 1512110-1512110 on Build GRCh38
rs777517973 Chr.1: 1512250-1512250 on Build GRCh38
rs1002388048 Chr.1: 1512201-1512201 on Build GRCh38
rs751438870 Chr.1: 1512343-1512343 on Build GRCh38
rs940641770 Chr.1: 1512132-1512132 on Build GRCh38
rs745668318 Chr.1: 1512213-1512213 on Build GRCh38
rs960755628 Chr.1: 1512114-1512114 on Build GRCh38
rs963092143 Chr.1: 1512394-1512394 on Build GRCh38
rs898999921 Chr.1: 1512060-1512060 on Build GRCh38
rs911686477 Chr.1: 1512293-1512293 on Build GRCh38
rs113982948 Chr.1: 1512325-1512325 on Build GRCh38
rs754970619 Chr.1: 1512321-1512321 on Build GRCh38
rs769431090 Chr.1: 1512263-1512263 on Build GRCh38
rs539230501 Chr.1: 1512107-1512107 on Build GRCh38
rs890216984 Chr.1: 1512167-1512167 on Build GRCh38
rs780037943 Chr.1: 1512019-1512019 on Build GRCh38
rs775107741 Chr.1: 1512267-1512267 on Build GRCh38
rs756278855 Chr.1: 1512225-1512225 on Build GRCh38
rs1013193508 Chr.1: 1512183-1512183 on Build GRCh38
rs969533977 Chr.1: 1512100-1512100 on Build GRCh38
rs1022506559 Chr.1: 1512096-1512096 on Build GRCh38
rs892123263 Chr.1: 1512172-1512172 on Build GRCh38
rs1008998356 Chr.1: 1512082-1512082 on Build GRCh38
rs973333524 Chr.1: 1512143-1512144 on Build GRCh38
rs745949447 Chr.1: 1512238-1512252 on Build GRCh38
rs781409895 Chr.1: 1512229-1512230 on Build GRCh38
rs182127890 Chr.1: 1512117-1512117 on Build GRCh38
rs774681303 Chr.1: 1512277-1512277 on Build GRCh38
rs556239318 Chr.1: 1512307-1512307 on Build GRCh38
rs1240736 Chr.1: 1512376-1512376 on Build GRCh38
rs769229163 Chr.1: 1512274-1512274 on Build GRCh38
rs1022535760 Chr.1: 1512190-1512190 on Build GRCh38
rs931764757 Chr.1: 1512064-1512064 on Build GRCh38
rs535379248 Chr.1: 1512181-1512181 on Build GRCh38
rs1052177288 Chr.1: 1512300-1512300 on Build GRCh38

View Full Product Details