Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17743794-17744334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BID
Entrez Gene ID
637
Gene Name
BH3 interacting domain death agonist
Gene Aliases
FP497
UniGene
Hs.517145
Gene Chromosome Location
Chr.22: 17734140-17774665 on Build GRCh38
Transcripts
NM_001196.3 NM_001244567.1 NM_001244569.1 NM_001244570.1 NM_001244572.1 NM_197966.2 NM_197967.2 XM_017028906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3842153 c.40G>A p.E14K Chr.22: 17743986-17743986 on Build GRCh38
116387 c.329G>C p.S110T Chr.22: 17743835-17743835 on Build GRCh38
3785603 c.201C>T p.S67S Chr.22: 17743825-17743825 on Build GRCh38
3964075 c.223+6C>T p.? Chr.22: 17743797-17743797 on Build GRCh38
7230209 c.159C>T p.N53N Chr.22: 17744005-17744005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977290161 Chr.22: 17744313-17744313 on Build GRCh38
rs189923680 Chr.22: 17743942-17743942 on Build GRCh38
rs772763406 Chr.22: 17743887-17743887 on Build GRCh38
rs747016844 Chr.22: 17743899-17743899 on Build GRCh38
rs764876536 Chr.22: 17744059-17744059 on Build GRCh38
rs1029613671 Chr.22: 17744291-17744291 on Build GRCh38
rs776679558 Chr.22: 17743905-17743905 on Build GRCh38
rs750006117 Chr.22: 17743852-17743852 on Build GRCh38
rs759103109 Chr.22: 17744049-17744049 on Build GRCh38
rs547029088 Chr.22: 17743864-17743864 on Build GRCh38
rs747539715 Chr.22: 17743977-17743977 on Build GRCh38
rs200409062 Chr.22: 17743912-17743912 on Build GRCh38
rs903232389 Chr.22: 17744076-17744076 on Build GRCh38
rs768886503 Chr.22: 17743835-17743836 on Build GRCh38
rs535265881 Chr.22: 17743989-17743989 on Build GRCh38
rs760132280 Chr.22: 17743925-17743925 on Build GRCh38
rs761738989 Chr.22: 17743918-17743918 on Build GRCh38
rs767379821 Chr.22: 17743922-17743922 on Build GRCh38
rs748577760 Chr.22: 17743881-17743881 on Build GRCh38
rs755255176 Chr.22: 17743869-17743869 on Build GRCh38
rs771880230 Chr.22: 17744023-17744023 on Build GRCh38
rs940864168 Chr.22: 17743961-17743961 on Build GRCh38
rs888197362 Chr.22: 17744169-17744169 on Build GRCh38
rs773096522 Chr.22: 17744035-17744035 on Build GRCh38
rs771344517 Chr.22: 17743838-17743838 on Build GRCh38
rs753179702 Chr.22: 17743954-17743954 on Build GRCh38
rs982436538 Chr.22: 17744176-17744176 on Build GRCh38
rs779396721 Chr.22: 17744008-17744009 on Build GRCh38
rs1032783427 Chr.22: 17744142-17744142 on Build GRCh38
rs1005185292 Chr.22: 17744227-17744227 on Build GRCh38
rs11542850 Chr.22: 17743878-17743878 on Build GRCh38
rs1023958067 Chr.22: 17744152-17744152 on Build GRCh38
rs574200539 Chr.22: 17744228-17744228 on Build GRCh38
rs61757628 Chr.22: 17743824-17743824 on Build GRCh38
rs779372119 Chr.22: 17743872-17743872 on Build GRCh38
rs781383435 Chr.22: 17743992-17743992 on Build GRCh38
rs372024838 Chr.22: 17743911-17743911 on Build GRCh38
rs749551829 Chr.22: 17743819-17743819 on Build GRCh38
rs747883996 Chr.22: 17744017-17744017 on Build GRCh38
rs754705127 Chr.22: 17743936-17743936 on Build GRCh38
rs543546745 Chr.22: 17744132-17744132 on Build GRCh38
rs922447901 Chr.22: 17743854-17743854 on Build GRCh38
rs1020789560 Chr.22: 17744045-17744045 on Build GRCh38
rs932255856 Chr.22: 17744287-17744287 on Build GRCh38
rs1054197372 Chr.22: 17743984-17743984 on Build GRCh38
rs113033070 Chr.22: 17743970-17743970 on Build GRCh38
rs753398808 Chr.22: 17743933-17743933 on Build GRCh38
rs753553004 Chr.22: 17743868-17743868 on Build GRCh38
rs756092459 Chr.22: 17743841-17743841 on Build GRCh38
rs781725256 Chr.22: 17744166-17744166 on Build GRCh38
rs899436087 Chr.22: 17743985-17743985 on Build GRCh38
rs8190313 Chr.22: 17744311-17744311 on Build GRCh38
rs2072392 Chr.22: 17743846-17743846 on Build GRCh38
rs577608009 Chr.22: 17744129-17744129 on Build GRCh38
rs201438109 Chr.22: 17743835-17743835 on Build GRCh38
rs8190314 Chr.22: 17744005-17744005 on Build GRCh38
rs759844569 Chr.22: 17743910-17743910 on Build GRCh38
rs184637515 Chr.22: 17744265-17744265 on Build GRCh38
rs559996997 Chr.22: 17744232-17744232 on Build GRCh38
rs760965442 Chr.22: 17743837-17743837 on Build GRCh38
rs764590275 Chr.22: 17743926-17743926 on Build GRCh38
rs777994982 Chr.22: 17743889-17743889 on Build GRCh38
rs769081918 Chr.22: 17743823-17743823 on Build GRCh38
rs557777819 Chr.22: 17744041-17744041 on Build GRCh38
rs971466394 Chr.22: 17744159-17744159 on Build GRCh38
rs776259151 Chr.22: 17744042-17744042 on Build GRCh38
rs778365897 Chr.22: 17743969-17743969 on Build GRCh38
rs199617244 Chr.22: 17743923-17743923 on Build GRCh38
rs8190315 Chr.22: 17743998-17743998 on Build GRCh38
rs760643580 Chr.22: 17744040-17744040 on Build GRCh38
rs773469812 Chr.22: 17743836-17743836 on Build GRCh38
rs182040518 Chr.22: 17744026-17744026 on Build GRCh38
rs879161693 Chr.22: 17743856-17743856 on Build GRCh38
rs778333007 Chr.22: 17744312-17744312 on Build GRCh38
rs557015605 Chr.22: 17744149-17744149 on Build GRCh38
rs958636408 Chr.22: 17744146-17744146 on Build GRCh38
rs406388 Chr.22: 17744231-17744231 on Build GRCh38
rs908303517 Chr.22: 17744223-17744223 on Build GRCh38
rs545703573 Chr.22: 17744289-17744289 on Build GRCh38
rs908249052 Chr.22: 17743939-17743939 on Build GRCh38
rs1000449662 Chr.22: 17744131-17744131 on Build GRCh38
rs770121452 Chr.22: 17744004-17744004 on Build GRCh38
rs748717824 Chr.22: 17743831-17743831 on Build GRCh38
rs758837211 Chr.22: 17743962-17743962 on Build GRCh38
rs150608132 Chr.22: 17743900-17743900 on Build GRCh38
rs377243261 Chr.22: 17743838-17743839 on Build GRCh38
rs755629886 Chr.22: 17743853-17743853 on Build GRCh38

View Full Product Details