Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 283167-283662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
UniGene
Hs.432397
Gene Chromosome Location
Chr.10: 274190-689710 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_005252430.2 XM_011519428.2 XM_011519429.2 XM_011519430.2 XM_011519431.2 XM_011519432.1 XM_011519433.2 XM_017015977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3437334 c.4260C>T p.F1420F Chr.10: 283306-283306 on Build GRCh38
4930462 c.4240C>A p.R1414S Chr.10: 283326-283326 on Build GRCh38
4430335 c.4193A>T p.H1398L Chr.10: 283373-283373 on Build GRCh38
3769109 c.4290T>A p.N1430K Chr.10: 283276-283276 on Build GRCh38
1250096 c.4205G>A p.R1402K Chr.10: 283361-283361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565662601 Chr.10: 283206-283206 on Build GRCh38
rs751088556 Chr.10: 283319-283319 on Build GRCh38
rs761410153 Chr.10: 283496-283496 on Build GRCh38
rs917913518 Chr.10: 283573-283573 on Build GRCh38
rs779629838 Chr.10: 283246-283247 on Build GRCh38
rs757875245 Chr.10: 283316-283316 on Build GRCh38
rs116933626 Chr.10: 283214-283214 on Build GRCh38
rs373743484 Chr.10: 283237-283237 on Build GRCh38
rs201157609 Chr.10: 283434-283434 on Build GRCh38
rs533187925 Chr.10: 283386-283386 on Build GRCh38
rs755878866 Chr.10: 283345-283345 on Build GRCh38
rs543293681 Chr.10: 283259-283259 on Build GRCh38
rs200158282 Chr.10: 283398-283398 on Build GRCh38
rs72653030 Chr.10: 283252-283252 on Build GRCh38
rs148820305 Chr.10: 283424-283424 on Build GRCh38
rs757565216 Chr.10: 283555-283555 on Build GRCh38
rs747153975 Chr.10: 283473-283473 on Build GRCh38
rs752219612 Chr.10: 283244-283244 on Build GRCh38
rs371053708 Chr.10: 283406-283406 on Build GRCh38
rs781669452 Chr.10: 283477-283477 on Build GRCh38
rs892326419 Chr.10: 283638-283638 on Build GRCh38
rs377557232 Chr.10: 283256-283256 on Build GRCh38
rs766691147 Chr.10: 283226-283226 on Build GRCh38
rs150522756 Chr.10: 283591-283591 on Build GRCh38
rs540557030 Chr.10: 283202-283202 on Build GRCh38
rs528054364 Chr.10: 283488-283488 on Build GRCh38
rs537220720 Chr.10: 283236-283236 on Build GRCh38
rs534481067 Chr.10: 283326-283326 on Build GRCh38
rs545182516 Chr.10: 283329-283329 on Build GRCh38
rs757914975 Chr.10: 283245-283245 on Build GRCh38
rs115894082 Chr.10: 283200-283200 on Build GRCh38
rs748363942 Chr.10: 283369-283369 on Build GRCh38
rs398074835 Chr.10: 283250-283250 on Build GRCh38
rs931377121 Chr.10: 283515-283515 on Build GRCh38
rs551082156 Chr.10: 283495-283495 on Build GRCh38
rs936605033 Chr.10: 283609-283609 on Build GRCh38
rs962935746 Chr.10: 283509-283509 on Build GRCh38
rs748824439 Chr.10: 283250-283252 on Build GRCh38
rs763031005 Chr.10: 283435-283435 on Build GRCh38
rs751178524 Chr.10: 283416-283416 on Build GRCh38
rs768827016 Chr.10: 283251-283251 on Build GRCh38
rs192450400 Chr.10: 283225-283225 on Build GRCh38
rs201616323 Chr.10: 283265-283265 on Build GRCh38
rs777967580 Chr.10: 283464-283464 on Build GRCh38
rs763608800 Chr.10: 283318-283318 on Build GRCh38
rs536546291 Chr.10: 283595-283595 on Build GRCh38
rs1030444345 Chr.10: 283240-283240 on Build GRCh38
rs2303991 Chr.10: 283547-283547 on Build GRCh38
rs3815985 Chr.10: 283553-283553 on Build GRCh38
rs764279202 Chr.10: 283532-283533 on Build GRCh38
rs755738558 Chr.10: 283243-283244 on Build GRCh38
rs557792743 Chr.10: 283217-283217 on Build GRCh38
rs149699277 Chr.10: 283404-283404 on Build GRCh38
rs373727129 Chr.10: 283301-283301 on Build GRCh38
rs17158883 Chr.10: 283190-283190 on Build GRCh38
rs917951931 Chr.10: 283300-283300 on Build GRCh38
rs373902857 Chr.10: 283458-283458 on Build GRCh38
rs368335581 Chr.10: 283452-283452 on Build GRCh38
rs529214854 Chr.10: 283423-283423 on Build GRCh38
rs748025955 Chr.10: 283455-283458 on Build GRCh38
rs746334386 Chr.10: 283480-283480 on Build GRCh38
rs371065533 Chr.10: 283252-283252 on Build GRCh38
rs1025803260 Chr.10: 283521-283521 on Build GRCh38
rs759895121 Chr.10: 283269-283269 on Build GRCh38
rs770942306 Chr.10: 283475-283475 on Build GRCh38
rs766848763 Chr.10: 283262-283262 on Build GRCh38
rs763219718 Chr.10: 283492-283492 on Build GRCh38
rs571037399 Chr.10: 283500-283500 on Build GRCh38
rs554187484 Chr.10: 283454-283454 on Build GRCh38
rs553588565 Chr.10: 283605-283605 on Build GRCh38
rs371940286 Chr.10: 283455-283455 on Build GRCh38
rs888807976 Chr.10: 283468-283468 on Build GRCh38
rs980816440 Chr.10: 283604-283604 on Build GRCh38
rs779006699 Chr.10: 283368-283368 on Build GRCh38
rs753512753 Chr.10: 283234-283234 on Build GRCh38
rs771498229 Chr.10: 283393-283393 on Build GRCh38
rs1052263552 Chr.10: 283630-283630 on Build GRCh38
rs777113194 Chr.10: 283396-283396 on Build GRCh38
rs1047143536 Chr.10: 283544-283544 on Build GRCh38
rs45445697 Chr.10: 283248-283248 on Build GRCh38
rs573142305 Chr.10: 283291-283291 on Build GRCh38
rs368114814 Chr.10: 283481-283481 on Build GRCh38
rs59023111 Chr.10: 283251-283251 on Build GRCh38
rs768480547 Chr.10: 283494-283494 on Build GRCh38
rs377450275 Chr.10: 283399-283399 on Build GRCh38
rs867411915 Chr.10: 283274-283274 on Build GRCh38
rs867251302 Chr.10: 283336-283336 on Build GRCh38
rs974178640 Chr.10: 283510-283510 on Build GRCh38
rs757296632 Chr.10: 283321-283321 on Build GRCh38
rs565346725 Chr.10: 283374-283374 on Build GRCh38
rs770097196 Chr.10: 283392-283392 on Build GRCh38
rs757137220 Chr.10: 283246-283246 on Build GRCh38
rs202195587 Chr.10: 283250-283250 on Build GRCh38
rs368272835 Chr.10: 283254-283254 on Build GRCh38
rs918631780 Chr.10: 283258-283258 on Build GRCh38
rs66861796 Chr.10: 283243-283243 on Build GRCh38
rs759909740 Chr.10: 283397-283397 on Build GRCh38
rs145443667 Chr.10: 283417-283417 on Build GRCh38
rs1008491754 Chr.10: 283285-283285 on Build GRCh38
rs893944866 Chr.10: 283617-283617 on Build GRCh38
rs747061908 Chr.10: 283390-283390 on Build GRCh38
rs745975770 Chr.10: 283241-283241 on Build GRCh38
rs574026213 Chr.10: 283253-283253 on Build GRCh38
rs768083759 Chr.10: 283251-283252 on Build GRCh38
rs764859549 Chr.10: 283279-283279 on Build GRCh38

View Full Product Details