Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245123876-245124411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB2
Entrez Gene ID
84288
Gene Name
EF-hand calcium binding domain 2
Gene Aliases
CFAP200, DRC8
UniGene
Hs.134857
Gene Chromosome Location
Chr.1: 244969739-245125228 on Build GRCh38
Transcripts
NM_001143943.1 NR_026587.1 XM_017002539.1 XM_017002540.1

SNP Detail

SNP ID Chromosome Location
rs865905706 Chr.1: 245124367-245124367 on Build GRCh38
rs1057504659 Chr.1: 245124384-245124384 on Build GRCh38
rs960224260 Chr.1: 245123963-245123963 on Build GRCh38
rs771928817 Chr.1: 245124332-245124332 on Build GRCh38
rs954779061 Chr.1: 245124021-245124021 on Build GRCh38
rs191041134 Chr.1: 245124329-245124329 on Build GRCh38
rs922254403 Chr.1: 245124061-245124061 on Build GRCh38
rs113267665 Chr.1: 245124303-245124303 on Build GRCh38
rs116173775 Chr.1: 245124390-245124390 on Build GRCh38
rs1056283413 Chr.1: 245124186-245124186 on Build GRCh38
rs998470257 Chr.1: 245124276-245124276 on Build GRCh38
rs975158180 Chr.1: 245124292-245124292 on Build GRCh38
rs1001665149 Chr.1: 245124185-245124187 on Build GRCh38
rs967175011 Chr.1: 245124004-245124004 on Build GRCh38
rs559664468 Chr.1: 245124215-245124215 on Build GRCh38
rs929578713 Chr.1: 245124373-245124373 on Build GRCh38
rs933661399 Chr.1: 245124082-245124082 on Build GRCh38
rs535437607 Chr.1: 245124330-245124330 on Build GRCh38
rs986211844 Chr.1: 245124023-245124023 on Build GRCh38
rs899277083 Chr.1: 245123916-245123916 on Build GRCh38
rs1001926022 Chr.1: 245123921-245123921 on Build GRCh38
rs976631699 Chr.1: 245124007-245124007 on Build GRCh38
rs558110247 Chr.1: 245123993-245123993 on Build GRCh38
rs986241548 Chr.1: 245124328-245124328 on Build GRCh38
rs577666158 Chr.1: 245124120-245124120 on Build GRCh38
rs147768586 Chr.1: 245124370-245124370 on Build GRCh38
rs910588846 Chr.1: 245124025-245124025 on Build GRCh38
rs12117676 Chr.1: 245124206-245124206 on Build GRCh38
rs113565574 Chr.1: 245123905-245123906 on Build GRCh38
rs868816033 Chr.1: 245124225-245124225 on Build GRCh38
rs765137288 Chr.1: 245123906-245123907 on Build GRCh38
rs1057306268 Chr.1: 245124185-245124185 on Build GRCh38
rs775409971 Chr.1: 245124374-245124374 on Build GRCh38
rs1044118868 Chr.1: 245124213-245124213 on Build GRCh38
rs111320109 Chr.1: 245124262-245124262 on Build GRCh38
rs564978572 Chr.1: 245124267-245124267 on Build GRCh38
rs528410523 Chr.1: 245124250-245124250 on Build GRCh38
rs1034386963 Chr.1: 245123944-245123944 on Build GRCh38
rs919533952 Chr.1: 245124126-245124126 on Build GRCh38
rs1012589084 Chr.1: 245123954-245123954 on Build GRCh38
rs145905528 Chr.1: 245124351-245124351 on Build GRCh38
rs12125365 Chr.1: 245124172-245124172 on Build GRCh38
rs978684974 Chr.1: 245123984-245123984 on Build GRCh38
rs936444005 Chr.1: 245124020-245124020 on Build GRCh38
rs111806462 Chr.1: 245124122-245124122 on Build GRCh38
rs956090885 Chr.1: 245123994-245123994 on Build GRCh38
rs955046547 Chr.1: 245124310-245124310 on Build GRCh38
rs113753800 Chr.1: 245124205-245124205 on Build GRCh38
rs10803267 Chr.1: 245124290-245124290 on Build GRCh38
rs114927989 Chr.1: 245123965-245123965 on Build GRCh38
rs940846412 Chr.1: 245124145-245124145 on Build GRCh38
rs968354882 Chr.1: 245123966-245123966 on Build GRCh38

View Full Product Details