Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 68782228-68782723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122A
Entrez Gene ID
116224
Gene Name
family with sequence similarity 122A
Gene Aliases
C9orf42
UniGene
Hs.655572
Gene Chromosome Location
Chr.9: 68780006-68783879 on Build GRCh38
Transcripts
NM_138333.4 NM_001278253.1 NM_003558.3 XM_005252261.3 XM_005252262.4 XM_006717300.2 XM_006717301.1 XM_011519082.2 XM_011519083.2 XM_011519084.2 XM_017015188.1 XM_017015189.1 XM_017015190.1 XM_017015191.1

Gene Symbol
PIP5K1B
Entrez Gene ID
8395
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
Gene Aliases
MSS4, STM7
UniGene
Hs.534371
Gene Chromosome Location
Chr.9: 68705217-69009176 on Build GRCh38
Transcripts
NM_138333.4 NM_001278253.1 NM_003558.3 XM_005252261.3 XM_005252262.4 XM_006717300.2 XM_006717301.1 XM_011519082.2 XM_011519083.2 XM_011519084.2 XM_017015188.1 XM_017015189.1 XM_017015190.1 XM_017015191.1

SNP Detail

SNP ID Chromosome Location
rs191856518 Chr.9: 68782439-68782439 on Build GRCh38
rs944117058 Chr.9: 68782614-68782614 on Build GRCh38
rs758494430 Chr.9: 68782463-68782463 on Build GRCh38
rs1008288685 Chr.9: 68782272-68782272 on Build GRCh38
rs182485993 Chr.9: 68782582-68782582 on Build GRCh38
rs1024672151 Chr.9: 68782411-68782411 on Build GRCh38
rs148768467 Chr.9: 68782576-68782576 on Build GRCh38
rs995683745 Chr.9: 68782358-68782358 on Build GRCh38
rs1051570433 Chr.9: 68782598-68782598 on Build GRCh38
rs560468460 Chr.9: 68782404-68782404 on Build GRCh38
rs142133645 Chr.9: 68782590-68782590 on Build GRCh38
rs974696633 Chr.9: 68782539-68782545 on Build GRCh38
rs1036301736 Chr.9: 68782424-68782424 on Build GRCh38
rs1027225842 Chr.9: 68782407-68782407 on Build GRCh38
rs1018048671 Chr.9: 68782301-68782301 on Build GRCh38
rs1001186345 Chr.9: 68782324-68782324 on Build GRCh38
rs991256147 Chr.9: 68782512-68782512 on Build GRCh38
rs781208996 Chr.9: 68782409-68782409 on Build GRCh38
rs75064770 Chr.9: 68782271-68782271 on Build GRCh38
rs899696341 Chr.9: 68782314-68782314 on Build GRCh38
rs527450469 Chr.9: 68782419-68782419 on Build GRCh38
rs1032259737 Chr.9: 68782357-68782357 on Build GRCh38
rs934608540 Chr.9: 68782591-68782591 on Build GRCh38
rs1047106022 Chr.9: 68782268-68782268 on Build GRCh38
rs767122987 Chr.9: 68782366-68782366 on Build GRCh38
rs759615932 Chr.9: 68782350-68782350 on Build GRCh38
rs1039773887 Chr.9: 68782687-68782687 on Build GRCh38
rs956537429 Chr.9: 68782396-68782396 on Build GRCh38
rs752196730 Chr.9: 68782416-68782416 on Build GRCh38
rs944100759 Chr.9: 68782625-68782625 on Build GRCh38
rs915552212 Chr.9: 68782514-68782514 on Build GRCh38
rs545242190 Chr.9: 68782294-68782294 on Build GRCh38
rs752038530 Chr.9: 68782282-68782282 on Build GRCh38
rs980552092 Chr.9: 68782417-68782417 on Build GRCh38
rs899859951 Chr.9: 68782340-68782340 on Build GRCh38
rs567500481 Chr.9: 68782551-68782551 on Build GRCh38
rs779407291 Chr.9: 68782363-68782367 on Build GRCh38
rs890287719 Chr.9: 68782609-68782609 on Build GRCh38
rs765728652 Chr.9: 68782518-68782518 on Build GRCh38
rs927444607 Chr.9: 68782492-68782492 on Build GRCh38
rs755495763 Chr.9: 68782335-68782335 on Build GRCh38
rs1018190258 Chr.9: 68782326-68782326 on Build GRCh38
rs920202931 Chr.9: 68782563-68782564 on Build GRCh38
rs186561008 Chr.9: 68782655-68782655 on Build GRCh38
rs1008272306 Chr.9: 68782323-68782323 on Build GRCh38
rs947023064 Chr.9: 68782547-68782547 on Build GRCh38

View Full Product Details