Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55981767-55982272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
UniGene
Hs.591069
Gene Chromosome Location
Chr.7: 55964577-56000182 on Build GRCh38
Transcripts
NM_001202469.1 NM_001483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1158003 c.403C>T p.P135S Chr.7: 55982239-55982239 on Build GRCh38
5031521 c.391G>A p.E131K Chr.7: 55982227-55982227 on Build GRCh38
5662314 c.411C>T p.L137L Chr.7: 55982247-55982247 on Build GRCh38
6044085 c.402T>A p.Y134* Chr.7: 55982238-55982238 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs954731292 Chr.7: 55982101-55982101 on Build GRCh38
rs73343724 Chr.7: 55981861-55981861 on Build GRCh38
rs954463302 Chr.7: 55981890-55981890 on Build GRCh38
rs564003419 Chr.7: 55982164-55982164 on Build GRCh38
rs543451902 Chr.7: 55981885-55981885 on Build GRCh38
rs796263965 Chr.7: 55981954-55981954 on Build GRCh38
rs777773662 Chr.7: 55982229-55982229 on Build GRCh38
rs943041733 Chr.7: 55981913-55981913 on Build GRCh38
rs1007001853 Chr.7: 55981988-55981988 on Build GRCh38
rs751411756 Chr.7: 55981866-55981866 on Build GRCh38
rs1041864333 Chr.7: 55981995-55981995 on Build GRCh38
rs557957801 Chr.7: 55981852-55981852 on Build GRCh38
rs1019697188 Chr.7: 55982004-55982004 on Build GRCh38
rs766350531 Chr.7: 55981855-55981855 on Build GRCh38
rs914861962 Chr.7: 55981965-55981965 on Build GRCh38
rs755954356 Chr.7: 55982226-55982226 on Build GRCh38
rs1043200579 Chr.7: 55981976-55981976 on Build GRCh38
rs527457367 Chr.7: 55982103-55982103 on Build GRCh38
rs947627480 Chr.7: 55981968-55981968 on Build GRCh38
rs573706111 Chr.7: 55981930-55981930 on Build GRCh38
rs371480418 Chr.7: 55982020-55982020 on Build GRCh38
rs41298773 Chr.7: 55981834-55981834 on Build GRCh38
rs1006907052 Chr.7: 55981809-55981810 on Build GRCh38
rs199850047 Chr.7: 55982237-55982237 on Build GRCh38
rs182076369 Chr.7: 55982177-55982177 on Build GRCh38
rs754791002 Chr.7: 55981908-55981908 on Build GRCh38
rs184028368 Chr.7: 55981803-55981803 on Build GRCh38
rs1053817103 Chr.7: 55981961-55981961 on Build GRCh38
rs1016225359 Chr.7: 55982049-55982049 on Build GRCh38
rs560035825 Chr.7: 55982083-55982083 on Build GRCh38
rs376936934 Chr.7: 55982072-55982072 on Build GRCh38
rs994166692 Chr.7: 55982047-55982047 on Build GRCh38
rs6947109 Chr.7: 55982027-55982027 on Build GRCh38
rs749781522 Chr.7: 55981878-55981878 on Build GRCh38
rs758441332 Chr.7: 55981845-55981845 on Build GRCh38
rs1039978684 Chr.7: 55981928-55981928 on Build GRCh38
rs535466489 Chr.7: 55982238-55982238 on Build GRCh38
rs552383188 Chr.7: 55982120-55982120 on Build GRCh38
rs983932587 Chr.7: 55981871-55981871 on Build GRCh38
rs111432465 Chr.7: 55981909-55981909 on Build GRCh38
rs540039567 Chr.7: 55981810-55981810 on Build GRCh38
rs987641342 Chr.7: 55981899-55981899 on Build GRCh38
rs1019602314 Chr.7: 55981831-55981831 on Build GRCh38
rs1026119647 Chr.7: 55982088-55982088 on Build GRCh38
rs984375337 Chr.7: 55982116-55982116 on Build GRCh38
rs903360980 Chr.7: 55981996-55981996 on Build GRCh38
rs993841531 Chr.7: 55982087-55982087 on Build GRCh38
rs368639343 Chr.7: 55982204-55982204 on Build GRCh38
rs1003386568 Chr.7: 55981796-55981796 on Build GRCh38
rs144366493 Chr.7: 55982225-55982225 on Build GRCh38
rs889891938 Chr.7: 55981980-55981980 on Build GRCh38
rs765942404 Chr.7: 55982175-55982175 on Build GRCh38
rs748414809 Chr.7: 55982185-55982185 on Build GRCh38
rs754457934 Chr.7: 55982188-55982188 on Build GRCh38
rs919088216 Chr.7: 55981886-55981886 on Build GRCh38
rs757643077 Chr.7: 55982165-55982165 on Build GRCh38
rs963224611 Chr.7: 55982071-55982071 on Build GRCh38
rs549792039 Chr.7: 55982199-55982199 on Build GRCh38
rs1002944742 Chr.7: 55981999-55981999 on Build GRCh38
rs927659203 Chr.7: 55981934-55981934 on Build GRCh38
rs376656557 Chr.7: 55982240-55982240 on Build GRCh38
rs1057467970 Chr.7: 55982013-55982013 on Build GRCh38
rs532821922 Chr.7: 55982012-55982012 on Build GRCh38

View Full Product Details