Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134960847-134961393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBS1L
Entrez Gene ID
10767
Gene Name
HBS1 like translational GTPase
Gene Aliases
EF-1a, ERFS, HBS1, HSPC276, eRF3c
UniGene
Hs.378532
Gene Chromosome Location
Chr.6: 134960378-135054898 on Build GRCh38
Transcripts
NM_001145158.1 NM_006620.3

SNP Detail

SNP ID Chromosome Location
rs905670086 Chr.6: 134961118-134961118 on Build GRCh38
rs71547020 Chr.6: 134961109-134961110 on Build GRCh38
rs185326013 Chr.6: 134961040-134961040 on Build GRCh38
rs563561479 Chr.6: 134961157-134961157 on Build GRCh38
rs1026166588 Chr.6: 134961078-134961078 on Build GRCh38
rs1017021252 Chr.6: 134961026-134961026 on Build GRCh38
rs1035578333 Chr.6: 134961191-134961191 on Build GRCh38
rs892069415 Chr.6: 134960944-134960944 on Build GRCh38
rs139904739 Chr.6: 134961261-134961261 on Build GRCh38
rs561327250 Chr.6: 134960983-134960983 on Build GRCh38
rs995395795 Chr.6: 134961085-134961085 on Build GRCh38
rs533684561 Chr.6: 134961136-134961136 on Build GRCh38
rs111288345 Chr.6: 134960999-134960999 on Build GRCh38
rs779500956 Chr.6: 134961092-134961093 on Build GRCh38
rs765421871 Chr.6: 134960907-134960907 on Build GRCh38
rs913981186 Chr.6: 134960936-134960936 on Build GRCh38
rs1002711695 Chr.6: 134961180-134961180 on Build GRCh38
rs74684002 Chr.6: 134961359-134961359 on Build GRCh38
rs928976231 Chr.6: 134961022-134961022 on Build GRCh38
rs73774524 Chr.6: 134960889-134960889 on Build GRCh38
rs766744280 Chr.6: 134961203-134961203 on Build GRCh38
rs866568536 Chr.6: 134960992-134960992 on Build GRCh38
rs77051210 Chr.6: 134961089-134961089 on Build GRCh38
rs398048877 Chr.6: 134961109-134961109 on Build GRCh38
rs7739722 Chr.6: 134961087-134961087 on Build GRCh38
rs964813581 Chr.6: 134961300-134961302 on Build GRCh38
rs1021399528 Chr.6: 134961244-134961244 on Build GRCh38
rs994142833 Chr.6: 134961041-134961041 on Build GRCh38
rs528452763 Chr.6: 134961317-134961317 on Build GRCh38
rs770452760 Chr.6: 134961216-134961216 on Build GRCh38
rs912929725 Chr.6: 134961260-134961260 on Build GRCh38
rs865970599 Chr.6: 134961027-134961027 on Build GRCh38
rs923542896 Chr.6: 134961328-134961328 on Build GRCh38
rs756132081 Chr.6: 134960882-134960882 on Build GRCh38
rs764186519 Chr.6: 134961090-134961091 on Build GRCh38
rs1051686810 Chr.6: 134960972-134960972 on Build GRCh38
rs539211566 Chr.6: 134960881-134960881 on Build GRCh38
rs200927478 Chr.6: 134960975-134960975 on Build GRCh38
rs1051388629 Chr.6: 134961264-134961264 on Build GRCh38
rs566152659 Chr.6: 134961088-134961090 on Build GRCh38
rs776801268 Chr.6: 134960871-134960871 on Build GRCh38
rs923524651 Chr.6: 134961012-134961012 on Build GRCh38
rs193251469 Chr.6: 134960925-134960925 on Build GRCh38
rs943555561 Chr.6: 134961007-134961007 on Build GRCh38
rs189581322 Chr.6: 134960911-134960911 on Build GRCh38
rs976687956 Chr.6: 134961325-134961325 on Build GRCh38
rs58274929 Chr.6: 134961107-134961109 on Build GRCh38
rs898604164 Chr.6: 134961324-134961324 on Build GRCh38
rs752003632 Chr.6: 134961093-134961093 on Build GRCh38
rs141763394 Chr.6: 134961284-134961284 on Build GRCh38
rs1024553502 Chr.6: 134961142-134961142 on Build GRCh38
rs1044180730 Chr.6: 134961088-134961088 on Build GRCh38
rs41286236 Chr.6: 134961010-134961010 on Build GRCh38
rs988456022 Chr.6: 134961220-134961220 on Build GRCh38
rs879176859 Chr.6: 134961091-134961094 on Build GRCh38
rs75109239 Chr.6: 134961271-134961271 on Build GRCh38
rs1014202659 Chr.6: 134961098-134961098 on Build GRCh38
rs7757811 Chr.6: 134961197-134961197 on Build GRCh38
rs942890234 Chr.6: 134961279-134961279 on Build GRCh38
rs925736321 Chr.6: 134961205-134961205 on Build GRCh38
rs934253242 Chr.6: 134960947-134960947 on Build GRCh38
rs541943520 Chr.6: 134960915-134960915 on Build GRCh38
rs77865145 Chr.6: 134961207-134961207 on Build GRCh38

View Full Product Details