Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236551247-236551796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR1
Entrez Gene ID
55127
Gene Name
HEAT repeat containing 1
Gene Aliases
BAP28, UTP10
UniGene
Hs.708114
Gene Chromosome Location
Chr.1: 236549005-236604541 on Build GRCh38
Transcripts
NM_018072.5 NM_006499.4 NM_201543.2 NM_201544.2 NM_201545.2

Gene Symbol
LGALS8
Entrez Gene ID
3964
Gene Name
galectin 8
Gene Aliases
Gal-8, PCTA-1, PCTA1, Po66-CBP
UniGene
Hs.4082
Gene Chromosome Location
Chr.1: 236518214-236552981 on Build GRCh38
Transcripts
NM_018072.5 NM_006499.4 NM_201543.2 NM_201544.2 NM_201545.2

SNP Detail

SNP ID Chromosome Location
rs990438162 Chr.1: 236551738-236551738 on Build GRCh38
rs867111103 Chr.1: 236551628-236551628 on Build GRCh38
rs984404496 Chr.1: 236551538-236551538 on Build GRCh38
rs906749775 Chr.1: 236551750-236551750 on Build GRCh38
rs540999366 Chr.1: 236551769-236551769 on Build GRCh38
rs1034569060 Chr.1: 236551705-236551705 on Build GRCh38
rs1042437808 Chr.1: 236551269-236551269 on Build GRCh38
rs1024709765 Chr.1: 236551375-236551375 on Build GRCh38
rs753370562 Chr.1: 236551588-236551588 on Build GRCh38
rs1012751735 Chr.1: 236551718-236551718 on Build GRCh38
rs1038902731 Chr.1: 236551525-236551527 on Build GRCh38
rs1001540340 Chr.1: 236551686-236551686 on Build GRCh38
rs1046436698 Chr.1: 236551749-236551749 on Build GRCh38
rs936678251 Chr.1: 236551679-236551679 on Build GRCh38
rs895117263 Chr.1: 236551706-236551706 on Build GRCh38
rs528811013 Chr.1: 236551357-236551357 on Build GRCh38
rs994646777 Chr.1: 236551349-236551349 on Build GRCh38
rs72632867 Chr.1: 236551410-236551410 on Build GRCh38
rs563809746 Chr.1: 236551367-236551367 on Build GRCh38
rs970836265 Chr.1: 236551429-236551429 on Build GRCh38
rs571593849 Chr.1: 236551409-236551409 on Build GRCh38
rs1038954646 Chr.1: 236551295-236551295 on Build GRCh38
rs115774627 Chr.1: 236551452-236551452 on Build GRCh38
rs546957114 Chr.1: 236551404-236551404 on Build GRCh38
rs567594512 Chr.1: 236551319-236551319 on Build GRCh38
rs2540 Chr.1: 236551616-236551616 on Build GRCh38
rs1005008552 Chr.1: 236551373-236551373 on Build GRCh38
rs1032454879 Chr.1: 236551367-236551367 on Build GRCh38
rs900429821 Chr.1: 236551525-236551525 on Build GRCh38
rs923102816 Chr.1: 236551596-236551596 on Build GRCh38
rs1005364749 Chr.1: 236551611-236551611 on Build GRCh38
rs148891348 Chr.1: 236551550-236551550 on Build GRCh38
rs758755122 Chr.1: 236551301-236551301 on Build GRCh38
rs867476038 Chr.1: 236551723-236551723 on Build GRCh38
rs950870861 Chr.1: 236551372-236551372 on Build GRCh38
rs752247258 Chr.1: 236551399-236551399 on Build GRCh38
rs957603864 Chr.1: 236551708-236551708 on Build GRCh38
rs1025575187 Chr.1: 236551412-236551412 on Build GRCh38
rs200962357 Chr.1: 236551408-236551408 on Build GRCh38
rs762445456 Chr.1: 236551409-236551410 on Build GRCh38
rs1054213259 Chr.1: 236551693-236551693 on Build GRCh38
rs530653200 Chr.1: 236551421-236551421 on Build GRCh38
rs945115406 Chr.1: 236551449-236551449 on Build GRCh38
rs990326858 Chr.1: 236551433-236551434 on Build GRCh38
rs944389433 Chr.1: 236551455-236551455 on Build GRCh38
rs374737742 Chr.1: 236551669-236551669 on Build GRCh38
rs1042275 Chr.1: 236551447-236551447 on Build GRCh38
rs923842686 Chr.1: 236551652-236551652 on Build GRCh38
rs1025848998 Chr.1: 236551450-236551450 on Build GRCh38
rs964831400 Chr.1: 236551575-236551575 on Build GRCh38
rs537207716 Chr.1: 236551586-236551586 on Build GRCh38
rs971452638 Chr.1: 236551379-236551379 on Build GRCh38
rs1042291346 Chr.1: 236551272-236551272 on Build GRCh38
rs3835675 Chr.1: 236551410-236551410 on Build GRCh38
rs549183623 Chr.1: 236551273-236551273 on Build GRCh38
rs755410907 Chr.1: 236551760-236551760 on Build GRCh38
rs575611598 Chr.1: 236551770-236551770 on Build GRCh38
rs913370208 Chr.1: 236551748-236551748 on Build GRCh38
rs186030072 Chr.1: 236551699-236551699 on Build GRCh38

View Full Product Details