Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215575904-215576429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD3
Entrez Gene ID
51133
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
NY-REN-45
UniGene
Hs.335139
Gene Chromosome Location
Chr.1: 215567185-215621821 on Build GRCh38
Transcripts
NM_001319294.1 NM_001319295.1 NM_016121.4 XM_005273158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5115746 c.190A>T p.I64F Chr.1: 215575907-215575907 on Build GRCh38
7138901 c.235C>G p.R79G Chr.1: 215575952-215575952 on Build GRCh38
7283115 c.208G>A p.A70T Chr.1: 215575925-215575925 on Build GRCh38
6692094 c.236G>A p.R79Q Chr.1: 215575953-215575953 on Build GRCh38
903805 c.241A>G p.K81E Chr.1: 215575958-215575958 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780303348 Chr.1: 215576086-215576086 on Build GRCh38
rs991808177 Chr.1: 215575989-215575989 on Build GRCh38
rs887553543 Chr.1: 215576354-215576354 on Build GRCh38
rs117699637 Chr.1: 215576286-215576286 on Build GRCh38
rs770468530 Chr.1: 215575983-215575983 on Build GRCh38
rs966910328 Chr.1: 215576019-215576020 on Build GRCh38
rs375849456 Chr.1: 215576269-215576269 on Build GRCh38
rs766396691 Chr.1: 215576019-215576019 on Build GRCh38
rs575919110 Chr.1: 215576063-215576063 on Build GRCh38
rs549631966 Chr.1: 215576274-215576274 on Build GRCh38
rs769079043 Chr.1: 215576002-215576002 on Build GRCh38
rs79889598 Chr.1: 215576074-215576074 on Build GRCh38
rs1047409177 Chr.1: 215576353-215576353 on Build GRCh38
rs773400266 Chr.1: 215576010-215576010 on Build GRCh38
rs183535121 Chr.1: 215576194-215576194 on Build GRCh38
rs541754775 Chr.1: 215576064-215576064 on Build GRCh38
rs1013669744 Chr.1: 215576178-215576178 on Build GRCh38
rs760755487 Chr.1: 215576011-215576011 on Build GRCh38
rs549726898 Chr.1: 215576366-215576366 on Build GRCh38
rs749264060 Chr.1: 215576208-215576208 on Build GRCh38
rs1026081348 Chr.1: 215576157-215576157 on Build GRCh38
rs551483992 Chr.1: 215576360-215576360 on Build GRCh38
rs116986800 Chr.1: 215576294-215576294 on Build GRCh38
rs976020792 Chr.1: 215576230-215576230 on Build GRCh38
rs1056724434 Chr.1: 215576329-215576329 on Build GRCh38
rs988075305 Chr.1: 215576278-215576278 on Build GRCh38
rs1048956057 Chr.1: 215576352-215576352 on Build GRCh38
rs560180992 Chr.1: 215576114-215576114 on Build GRCh38
rs887595403 Chr.1: 215576124-215576124 on Build GRCh38
rs768176131 Chr.1: 215576007-215576007 on Build GRCh38
rs897745153 Chr.1: 215576095-215576095 on Build GRCh38
rs755084873 Chr.1: 215576155-215576155 on Build GRCh38
rs749335442 Chr.1: 215575994-215575995 on Build GRCh38
rs1033857674 Chr.1: 215576151-215576151 on Build GRCh38
rs928973509 Chr.1: 215576345-215576345 on Build GRCh38
rs1001909239 Chr.1: 215575931-215575931 on Build GRCh38
rs896230948 Chr.1: 215576393-215576393 on Build GRCh38
rs140349105 Chr.1: 215576369-215576369 on Build GRCh38
rs925433722 Chr.1: 215576050-215576051 on Build GRCh38
rs780355090 Chr.1: 215576315-215576315 on Build GRCh38
rs528645407 Chr.1: 215576400-215576400 on Build GRCh38
rs867383137 Chr.1: 215575932-215575932 on Build GRCh38
rs954179922 Chr.1: 215576281-215576281 on Build GRCh38
rs896156620 Chr.1: 215576142-215576142 on Build GRCh38
rs578050703 Chr.1: 215576292-215576292 on Build GRCh38
rs776298076 Chr.1: 215575987-215575987 on Build GRCh38
rs1033376217 Chr.1: 215575966-215575966 on Build GRCh38
rs751556344 Chr.1: 215576060-215576060 on Build GRCh38
rs756702176 Chr.1: 215576229-215576229 on Build GRCh38
rs373023035 Chr.1: 215576005-215576005 on Build GRCh38
rs918587450 Chr.1: 215576313-215576313 on Build GRCh38
rs571881399 Chr.1: 215576154-215576154 on Build GRCh38
rs1017423016 Chr.1: 215576221-215576221 on Build GRCh38
rs553561436 Chr.1: 215575957-215575957 on Build GRCh38
rs1048924212 Chr.1: 215576120-215576120 on Build GRCh38
rs73085420 Chr.1: 215576050-215576050 on Build GRCh38
rs955385411 Chr.1: 215576257-215576257 on Build GRCh38
rs1001940298 Chr.1: 215576149-215576149 on Build GRCh38
rs555118616 Chr.1: 215576067-215576067 on Build GRCh38
rs912621174 Chr.1: 215576056-215576059 on Build GRCh38
rs556169235 Chr.1: 215576059-215576059 on Build GRCh38
rs529597253 Chr.1: 215576332-215576332 on Build GRCh38
rs147593861 Chr.1: 215576293-215576293 on Build GRCh38
rs926803576 Chr.1: 215576299-215576299 on Build GRCh38
rs746571322 Chr.1: 215575974-215575974 on Build GRCh38
rs951751181 Chr.1: 215576193-215576193 on Build GRCh38
rs1054924714 Chr.1: 215576376-215576376 on Build GRCh38
rs570545531 Chr.1: 215575998-215575998 on Build GRCh38
rs929219751 Chr.1: 215576106-215576106 on Build GRCh38
rs1036995815 Chr.1: 215576092-215576092 on Build GRCh38
rs997727236 Chr.1: 215576389-215576389 on Build GRCh38
rs1043878080 Chr.1: 215576052-215576052 on Build GRCh38
rs923154504 Chr.1: 215576237-215576237 on Build GRCh38
rs770838275 Chr.1: 215576209-215576209 on Build GRCh38
rs896710710 Chr.1: 215576339-215576339 on Build GRCh38

View Full Product Details