Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 16178663-16179170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARCH11
Entrez Gene ID
441061
Gene Name
membrane associated ring-CH-type finger 11
Gene Aliases
MARCH-XI
UniGene
Hs.662216
Gene Chromosome Location
Chr.5: 16067365-16179788 on Build GRCh38
Transcripts
NM_001102562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7281760 c.486G>A p.Q162Q Chr.5: 16179090-16179090 on Build GRCh38
4793149 c.435C>T p.C145C Chr.5: 16179141-16179141 on Build GRCh38
5679129 c.526G>T p.G176C Chr.5: 16179050-16179050 on Build GRCh38
6400812 c.415C>A p.P139T Chr.5: 16179161-16179161 on Build GRCh38
5670576 c.531G>T p.A177A Chr.5: 16179045-16179045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888050383 Chr.5: 16179010-16179010 on Build GRCh38
rs971880977 Chr.5: 16178740-16178740 on Build GRCh38
rs550664498 Chr.5: 16178920-16178920 on Build GRCh38
rs747021686 Chr.5: 16179081-16179081 on Build GRCh38
rs997267445 Chr.5: 16178777-16178777 on Build GRCh38
rs529800936 Chr.5: 16178931-16178931 on Build GRCh38
rs342549 Chr.5: 16178811-16178811 on Build GRCh38
rs919157374 Chr.5: 16179015-16179015 on Build GRCh38
rs933425652 Chr.5: 16178750-16178750 on Build GRCh38
rs570383869 Chr.5: 16179027-16179027 on Build GRCh38
rs909160706 Chr.5: 16178697-16178697 on Build GRCh38
rs765057383 Chr.5: 16179113-16179113 on Build GRCh38
rs999086121 Chr.5: 16178845-16178845 on Build GRCh38
rs980847876 Chr.5: 16178951-16178951 on Build GRCh38
rs1043060672 Chr.5: 16178814-16178814 on Build GRCh38
rs564899428 Chr.5: 16178821-16178821 on Build GRCh38
rs924461506 Chr.5: 16178919-16178919 on Build GRCh38
rs528675303 Chr.5: 16178956-16178956 on Build GRCh38
rs1024763647 Chr.5: 16178911-16178911 on Build GRCh38
rs760778073 Chr.5: 16179031-16179031 on Build GRCh38
rs969161854 Chr.5: 16178950-16178950 on Build GRCh38
rs953095345 Chr.5: 16179034-16179034 on Build GRCh38
rs1030460619 Chr.5: 16178807-16178807 on Build GRCh38
rs1043461826 Chr.5: 16179078-16179078 on Build GRCh38
rs766565284 Chr.5: 16179121-16179123 on Build GRCh38
rs779258187 Chr.5: 16178993-16178993 on Build GRCh38
rs746054318 Chr.5: 16179009-16179009 on Build GRCh38
rs531971685 Chr.5: 16178888-16178888 on Build GRCh38
rs1056556226 Chr.5: 16178943-16178943 on Build GRCh38
rs774414851 Chr.5: 16179120-16179121 on Build GRCh38
rs889187805 Chr.5: 16178790-16178790 on Build GRCh38
rs1021299453 Chr.5: 16179086-16179086 on Build GRCh38
rs898000183 Chr.5: 16179020-16179020 on Build GRCh38
rs373613667 Chr.5: 16178730-16178730 on Build GRCh38
rs987259962 Chr.5: 16179019-16179019 on Build GRCh38
rs934628673 Chr.5: 16179148-16179148 on Build GRCh38
rs962139965 Chr.5: 16178775-16178775 on Build GRCh38
rs567857766 Chr.5: 16179011-16179011 on Build GRCh38
rs1051823202 Chr.5: 16178761-16178761 on Build GRCh38
rs977374844 Chr.5: 16179123-16179123 on Build GRCh38
rs149404393 Chr.5: 16178921-16178921 on Build GRCh38
rs1015898043 Chr.5: 16178952-16178952 on Build GRCh38
rs914521371 Chr.5: 16178864-16178864 on Build GRCh38
rs768683978 Chr.5: 16179059-16179059 on Build GRCh38
rs896007916 Chr.5: 16178904-16178904 on Build GRCh38
rs371438187 Chr.5: 16178973-16178973 on Build GRCh38
rs968605107 Chr.5: 16178881-16178881 on Build GRCh38
rs948072111 Chr.5: 16178961-16178961 on Build GRCh38
rs985769690 Chr.5: 16178819-16178819 on Build GRCh38
rs904779461 Chr.5: 16178830-16178830 on Build GRCh38
rs549639843 Chr.5: 16178949-16178949 on Build GRCh38
rs547260968 Chr.5: 16179102-16179102 on Build GRCh38
rs560024548 Chr.5: 16178940-16178940 on Build GRCh38
rs763153621 Chr.5: 16179064-16179064 on Build GRCh38
rs917979917 Chr.5: 16178948-16178948 on Build GRCh38
rs1014417788 Chr.5: 16178910-16178910 on Build GRCh38
rs342550 Chr.5: 16179016-16179016 on Build GRCh38
rs375619178 Chr.5: 16179030-16179030 on Build GRCh38
rs779938124 Chr.5: 16179018-16179018 on Build GRCh38
rs998052881 Chr.5: 16179024-16179024 on Build GRCh38
rs1015073467 Chr.5: 16178741-16178741 on Build GRCh38

View Full Product Details