Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21557051-21557570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLLT10
Entrez Gene ID
8028
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Gene Aliases
AF10
UniGene
Hs.30385
Gene Chromosome Location
Chr.10: 21533349-21743630 on Build GRCh38
Transcripts
NM_001195626.1 NM_001195627.1 NM_001195628.1 NM_001195630.1 NM_001324296.1 NM_001324297.1 NM_004641.3 NR_136736.1

SNP Detail

SNP ID Chromosome Location
rs958524183 Chr.10: 21557327-21557331 on Build GRCh38
rs1039104996 Chr.10: 21557346-21557346 on Build GRCh38
rs762951862 Chr.10: 21557312-21557312 on Build GRCh38
rs751425149 Chr.10: 21557460-21557460 on Build GRCh38
rs959327444 Chr.10: 21557427-21557427 on Build GRCh38
rs941328841 Chr.10: 21557162-21557162 on Build GRCh38
rs765428910 Chr.10: 21557175-21557175 on Build GRCh38
rs944839200 Chr.10: 21557339-21557339 on Build GRCh38
rs1032718706 Chr.10: 21557319-21557319 on Build GRCh38
rs897020968 Chr.10: 21557226-21557226 on Build GRCh38
rs938027235 Chr.10: 21557086-21557086 on Build GRCh38
rs1014015309 Chr.10: 21557203-21557203 on Build GRCh38
rs1022686819 Chr.10: 21557210-21557210 on Build GRCh38
rs558051099 Chr.10: 21557185-21557185 on Build GRCh38
rs941264317 Chr.10: 21557474-21557474 on Build GRCh38
rs1037316520 Chr.10: 21557424-21557424 on Build GRCh38
rs535584822 Chr.10: 21557098-21557098 on Build GRCh38
rs1000338199 Chr.10: 21557538-21557538 on Build GRCh38
rs1028062557 Chr.10: 21557140-21557140 on Build GRCh38
rs922055472 Chr.10: 21557365-21557365 on Build GRCh38
rs868615806 Chr.10: 21557208-21557208 on Build GRCh38
rs548763480 Chr.10: 21557119-21557119 on Build GRCh38
rs190674793 Chr.10: 21557395-21557395 on Build GRCh38
rs918393488 Chr.10: 21557491-21557491 on Build GRCh38
rs546931308 Chr.10: 21557279-21557279 on Build GRCh38
rs978489085 Chr.10: 21557266-21557266 on Build GRCh38
rs1050843214 Chr.10: 21557254-21557254 on Build GRCh38
rs7092670 Chr.10: 21557252-21557252 on Build GRCh38
rs146988779 Chr.10: 21557539-21557539 on Build GRCh38
rs1015456299 Chr.10: 21557173-21557173 on Build GRCh38
rs989437311 Chr.10: 21557329-21557329 on Build GRCh38
rs74120975 Chr.10: 21557074-21557074 on Build GRCh38
rs761326214 Chr.10: 21557137-21557138 on Build GRCh38
rs569666270 Chr.10: 21557083-21557083 on Build GRCh38
rs113966224 Chr.10: 21557388-21557388 on Build GRCh38
rs1004151186 Chr.10: 21557422-21557422 on Build GRCh38
rs1022800014 Chr.10: 21557500-21557500 on Build GRCh38
rs886426876 Chr.10: 21557421-21557421 on Build GRCh38
rs1011185208 Chr.10: 21557343-21557343 on Build GRCh38
rs1014129789 Chr.10: 21557476-21557476 on Build GRCh38
rs368089141 Chr.10: 21557160-21557160 on Build GRCh38
rs994764160 Chr.10: 21557138-21557138 on Build GRCh38
rs543114479 Chr.10: 21557350-21557350 on Build GRCh38
rs577552662 Chr.10: 21557247-21557247 on Build GRCh38
rs933818314 Chr.10: 21557531-21557534 on Build GRCh38
rs750359767 Chr.10: 21557196-21557198 on Build GRCh38
rs573405771 Chr.10: 21557448-21557448 on Build GRCh38
rs969816515 Chr.10: 21557518-21557518 on Build GRCh38
rs912084390 Chr.10: 21557337-21557337 on Build GRCh38
rs1055017561 Chr.10: 21557134-21557134 on Build GRCh38
rs56865732 Chr.10: 21557418-21557418 on Build GRCh38
rs930728081 Chr.10: 21557394-21557394 on Build GRCh38
rs573686055 Chr.10: 21557323-21557323 on Build GRCh38
rs909921386 Chr.10: 21557469-21557469 on Build GRCh38
rs900515051 Chr.10: 21557137-21557137 on Build GRCh38
rs1021060205 Chr.10: 21557367-21557367 on Build GRCh38
rs201296732 Chr.10: 21557161-21557161 on Build GRCh38

View Full Product Details