Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 57600756-57601293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NARS
Entrez Gene ID
4677
Gene Name
asparaginyl-tRNA synthetase
Gene Aliases
ASNRS, NARS1
UniGene
Hs.465224
Gene Chromosome Location
Chr.18: 57600656-57621945 on Build GRCh38
Transcripts
NM_004539.3 XM_005266700.1

SNP Detail

SNP ID Chromosome Location
rs147559695 Chr.18: 57601181-57601181 on Build GRCh38
rs923375292 Chr.18: 57600981-57600981 on Build GRCh38
rs946495535 Chr.18: 57600880-57600880 on Build GRCh38
rs528719755 Chr.18: 57601182-57601182 on Build GRCh38
rs751413287 Chr.18: 57601124-57601124 on Build GRCh38
rs892160895 Chr.18: 57601031-57601031 on Build GRCh38
rs1043980527 Chr.18: 57601225-57601225 on Build GRCh38
rs535944092 Chr.18: 57601133-57601133 on Build GRCh38
rs1026781677 Chr.18: 57600857-57600857 on Build GRCh38
rs150135633 Chr.18: 57600829-57600830 on Build GRCh38
rs987047691 Chr.18: 57601051-57601051 on Build GRCh38
rs1059648 Chr.18: 57600800-57600800 on Build GRCh38
rs983097961 Chr.18: 57600898-57600898 on Build GRCh38
rs187385712 Chr.18: 57601012-57601012 on Build GRCh38
rs571404920 Chr.18: 57601215-57601215 on Build GRCh38
rs969100272 Chr.18: 57600842-57600842 on Build GRCh38
rs1002111832 Chr.18: 57600932-57600932 on Build GRCh38
rs907910330 Chr.18: 57600904-57600904 on Build GRCh38
rs936976463 Chr.18: 57601023-57601023 on Build GRCh38
rs373921197 Chr.18: 57601070-57601070 on Build GRCh38
rs947484089 Chr.18: 57601187-57601187 on Build GRCh38
rs574211704 Chr.18: 57600830-57600830 on Build GRCh38
rs549672934 Chr.18: 57601162-57601162 on Build GRCh38
rs1002058879 Chr.18: 57600855-57600855 on Build GRCh38
rs772614070 Chr.18: 57601039-57601039 on Build GRCh38
rs142285081 Chr.18: 57601208-57601208 on Build GRCh38
rs937943187 Chr.18: 57601266-57601266 on Build GRCh38
rs1043566158 Chr.18: 57600894-57600894 on Build GRCh38
rs376424032 Chr.18: 57601212-57601212 on Build GRCh38
rs180883037 Chr.18: 57600896-57600896 on Build GRCh38
rs141508224 Chr.18: 57600829-57600829 on Build GRCh38
rs976026204 Chr.18: 57600968-57600968 on Build GRCh38
rs559827463 Chr.18: 57600929-57600929 on Build GRCh38
rs573007756 Chr.18: 57600838-57600838 on Build GRCh38
rs955992604 Chr.18: 57601082-57601083 on Build GRCh38
rs58950020 Chr.18: 57601014-57601014 on Build GRCh38
rs146942918 Chr.18: 57601055-57601055 on Build GRCh38
rs1059645 Chr.18: 57600815-57600815 on Build GRCh38
rs952668155 Chr.18: 57600897-57600897 on Build GRCh38
rs369573428 Chr.18: 57600966-57600966 on Build GRCh38
rs757339017 Chr.18: 57601029-57601029 on Build GRCh38
rs766432068 Chr.18: 57601105-57601105 on Build GRCh38
rs776107004 Chr.18: 57601128-57601128 on Build GRCh38
rs954466251 Chr.18: 57601040-57601040 on Build GRCh38
rs11356 Chr.18: 57601254-57601254 on Build GRCh38
rs183653451 Chr.18: 57601180-57601180 on Build GRCh38
rs1013273442 Chr.18: 57601039-57601039 on Build GRCh38
rs925434380 Chr.18: 57601017-57601017 on Build GRCh38
rs3200342 Chr.18: 57601134-57601134 on Build GRCh38
rs904946359 Chr.18: 57601113-57601113 on Build GRCh38
rs1035120575 Chr.18: 57600961-57600961 on Build GRCh38
rs538747287 Chr.18: 57601238-57601238 on Build GRCh38

View Full Product Details