Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13617711-13618212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL7
Entrez Gene ID
51406
Gene Name
nucleolar protein 7
Gene Aliases
C6orf90, PQBP3, RARG-1, dJ223E5.2
UniGene
Hs.306242
Gene Chromosome Location
Chr.6: 13615327-13632470 on Build GRCh38
Transcripts
NM_001317724.1 NM_016167.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073649 c.483A>C p.Q161H Chr.6: 13618122-13618122 on Build GRCh38
4450250 c.500+1G>A p.? Chr.6: 13618140-13618140 on Build GRCh38
5869370 c.455G>A p.G152E Chr.6: 13618094-13618094 on Build GRCh38
6727715 c.399G>A p.S133S Chr.6: 13617782-13617782 on Build GRCh38
3726479 c.418+5G>C p.? Chr.6: 13617806-13617806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770972860 Chr.6: 13618068-13618068 on Build GRCh38
rs1004685353 Chr.6: 13617977-13617977 on Build GRCh38
rs747111074 Chr.6: 13618057-13618058 on Build GRCh38
rs945259082 Chr.6: 13617773-13617773 on Build GRCh38
rs758753516 Chr.6: 13618039-13618039 on Build GRCh38
rs768240174 Chr.6: 13617832-13617832 on Build GRCh38
rs760576189 Chr.6: 13618014-13618014 on Build GRCh38
rs896029488 Chr.6: 13617844-13617844 on Build GRCh38
rs371855659 Chr.6: 13617796-13617796 on Build GRCh38
rs371620475 Chr.6: 13617813-13617813 on Build GRCh38
rs372106357 Chr.6: 13618114-13618114 on Build GRCh38
rs751217139 Chr.6: 13618024-13618024 on Build GRCh38
rs747793701 Chr.6: 13618106-13618106 on Build GRCh38
rs766181289 Chr.6: 13617865-13617865 on Build GRCh38
rs1046742673 Chr.6: 13618085-13618085 on Build GRCh38
rs767114023 Chr.6: 13618144-13618144 on Build GRCh38
rs770744144 Chr.6: 13618119-13618119 on Build GRCh38
rs6937316 Chr.6: 13617750-13617750 on Build GRCh38
rs537582496 Chr.6: 13618040-13618040 on Build GRCh38
rs779832921 Chr.6: 13618088-13618088 on Build GRCh38
rs752994875 Chr.6: 13617805-13617805 on Build GRCh38
rs1029951394 Chr.6: 13617866-13617866 on Build GRCh38
rs762722604 Chr.6: 13617842-13617842 on Build GRCh38
rs199636122 Chr.6: 13618125-13618125 on Build GRCh38
rs752892517 Chr.6: 13618128-13618128 on Build GRCh38
rs372783344 Chr.6: 13617746-13617746 on Build GRCh38
rs771749772 Chr.6: 13618111-13618111 on Build GRCh38
rs750301041 Chr.6: 13617781-13617781 on Build GRCh38
rs1044480809 Chr.6: 13617882-13617882 on Build GRCh38
rs112804444 Chr.6: 13617814-13617814 on Build GRCh38
rs751859334 Chr.6: 13618143-13618143 on Build GRCh38
rs748903190 Chr.6: 13618143-13618144 on Build GRCh38
rs866988704 Chr.6: 13617881-13617881 on Build GRCh38
rs190938573 Chr.6: 13618187-13618187 on Build GRCh38
rs185492293 Chr.6: 13617853-13617853 on Build GRCh38
rs779686051 Chr.6: 13617823-13617823 on Build GRCh38
rs748694360 Chr.6: 13617826-13617826 on Build GRCh38
rs759536441 Chr.6: 13618121-13618121 on Build GRCh38
rs563589049 Chr.6: 13617775-13617775 on Build GRCh38
rs921256523 Chr.6: 13617762-13617762 on Build GRCh38
rs749949750 Chr.6: 13618058-13618061 on Build GRCh38
rs752251351 Chr.6: 13617921-13617922 on Build GRCh38
rs747707859 Chr.6: 13618179-13618179 on Build GRCh38
rs767933519 Chr.6: 13618157-13618157 on Build GRCh38
rs763700292 Chr.6: 13617763-13617763 on Build GRCh38
rs753277108 Chr.6: 13617828-13617834 on Build GRCh38
rs778543171 Chr.6: 13618101-13618101 on Build GRCh38
rs139252019 Chr.6: 13618020-13618020 on Build GRCh38
rs752808210 Chr.6: 13618038-13618038 on Build GRCh38
rs765222106 Chr.6: 13618126-13618126 on Build GRCh38
rs548592826 Chr.6: 13617963-13617963 on Build GRCh38
rs367832387 Chr.6: 13617806-13617806 on Build GRCh38
rs760941711 Chr.6: 13617753-13617755 on Build GRCh38
rs371738827 Chr.6: 13618161-13618161 on Build GRCh38
rs376901906 Chr.6: 13617767-13617767 on Build GRCh38
rs560011476 Chr.6: 13617902-13617902 on Build GRCh38
rs949767860 Chr.6: 13617851-13617851 on Build GRCh38
rs767993129 Chr.6: 13618092-13618092 on Build GRCh38
rs551143186 Chr.6: 13617752-13617755 on Build GRCh38
rs755873110 Chr.6: 13618084-13618085 on Build GRCh38
rs771839553 Chr.6: 13617860-13617860 on Build GRCh38
rs776365553 Chr.6: 13618120-13618120 on Build GRCh38
rs201992657 Chr.6: 13617755-13617756 on Build GRCh38
rs762632474 Chr.6: 13617761-13617761 on Build GRCh38
rs765367644 Chr.6: 13618037-13618037 on Build GRCh38
rs368449251 Chr.6: 13618030-13618030 on Build GRCh38
rs905516734 Chr.6: 13617901-13617901 on Build GRCh38
rs759155299 Chr.6: 13618064-13618064 on Build GRCh38
rs761619335 Chr.6: 13617750-13617754 on Build GRCh38
rs976601152 Chr.6: 13617983-13617997 on Build GRCh38
rs142715266 Chr.6: 13618045-13618045 on Build GRCh38
rs778252721 Chr.6: 13618167-13618167 on Build GRCh38
rs954201438 Chr.6: 13617909-13617909 on Build GRCh38
rs111485145 Chr.6: 13617945-13617945 on Build GRCh38
rs772200497 Chr.6: 13617751-13617753 on Build GRCh38
rs757715477 Chr.6: 13618049-13618049 on Build GRCh38
rs201805498 Chr.6: 13618057-13618057 on Build GRCh38
rs778953813 Chr.6: 13617801-13617801 on Build GRCh38
rs781692387 Chr.6: 13618087-13618087 on Build GRCh38
rs754836062 Chr.6: 13618177-13618178 on Build GRCh38
rs766441029 Chr.6: 13617782-13617782 on Build GRCh38
rs775200795 Chr.6: 13617760-13617760 on Build GRCh38
rs757625538 Chr.6: 13618153-13618153 on Build GRCh38
rs896065842 Chr.6: 13617794-13617794 on Build GRCh38
rs551165584 Chr.6: 13617819-13617822 on Build GRCh38
rs1005605806 Chr.6: 13618172-13618172 on Build GRCh38
rs769375225 Chr.6: 13617756-13617756 on Build GRCh38
rs989259744 Chr.6: 13617986-13617986 on Build GRCh38
rs771634763 Chr.6: 13617822-13617822 on Build GRCh38
rs375111860 Chr.6: 13617738-13617738 on Build GRCh38
rs746387374 Chr.6: 13618084-13618084 on Build GRCh38
rs112007505 Chr.6: 13617747-13617747 on Build GRCh38
rs774072411 Chr.6: 13617764-13617764 on Build GRCh38
rs768764465 Chr.6: 13617737-13617738 on Build GRCh38
rs753837757 Chr.6: 13618021-13618021 on Build GRCh38
rs780813467 Chr.6: 13618034-13618037 on Build GRCh38
rs747509053 Chr.6: 13617817-13617817 on Build GRCh38
rs888561858 Chr.6: 13617840-13617840 on Build GRCh38
rs754238917 Chr.6: 13617756-13617756 on Build GRCh38
rs774370483 Chr.6: 13617741-13617744 on Build GRCh38
rs112933240 Chr.6: 13617974-13617974 on Build GRCh38
rs761539330 Chr.6: 13617858-13617858 on Build GRCh38
rs764360512 Chr.6: 13618140-13618140 on Build GRCh38
rs758764579 Chr.6: 13617836-13617837 on Build GRCh38

View Full Product Details