Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121328537-121329076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QRFPR
Entrez Gene ID
84109
Gene Name
pyroglutamylated RFamide peptide receptor
Gene Aliases
AQ27, GPR103, SP9155
UniGene
Hs.368977
Gene Chromosome Location
Chr.4: 121328642-121381029 on Build GRCh38
Transcripts
NM_198179.2

SNP Detail

SNP ID Chromosome Location
rs566255993 Chr.4: 121328711-121328711 on Build GRCh38
rs1013345580 Chr.4: 121328563-121328563 on Build GRCh38
rs937808092 Chr.4: 121328891-121328891 on Build GRCh38
rs535860691 Chr.4: 121328916-121328916 on Build GRCh38
rs534096076 Chr.4: 121328997-121328997 on Build GRCh38
rs1000612439 Chr.4: 121328797-121328797 on Build GRCh38
rs888494771 Chr.4: 121328929-121328929 on Build GRCh38
rs757300348 Chr.4: 121328780-121328780 on Build GRCh38
rs573240719 Chr.4: 121328860-121328860 on Build GRCh38
rs535159755 Chr.4: 121328763-121328763 on Build GRCh38
rs13101518 Chr.4: 121328596-121328596 on Build GRCh38
rs753214005 Chr.4: 121328661-121328661 on Build GRCh38
rs542366561 Chr.4: 121328879-121328879 on Build GRCh38
rs966400087 Chr.4: 121328619-121328619 on Build GRCh38
rs867517078 Chr.4: 121328824-121328824 on Build GRCh38
rs992942830 Chr.4: 121328800-121328800 on Build GRCh38
rs141452691 Chr.4: 121328956-121328956 on Build GRCh38
rs11721375 Chr.4: 121328588-121328588 on Build GRCh38
rs1026958683 Chr.4: 121328764-121328764 on Build GRCh38
rs966084256 Chr.4: 121328819-121328819 on Build GRCh38
rs562118015 Chr.4: 121328885-121328885 on Build GRCh38
rs909048990 Chr.4: 121328695-121328695 on Build GRCh38
rs959215093 Chr.4: 121329023-121329023 on Build GRCh38
rs184359090 Chr.4: 121328859-121328859 on Build GRCh38
rs146007405 Chr.4: 121328973-121328973 on Build GRCh38
rs796275354 Chr.4: 121328917-121328917 on Build GRCh38
rs986371823 Chr.4: 121328590-121328590 on Build GRCh38
rs999861353 Chr.4: 121328982-121328982 on Build GRCh38
rs932689342 Chr.4: 121328914-121328914 on Build GRCh38
rs867325847 Chr.4: 121328740-121328740 on Build GRCh38
rs919305683 Chr.4: 121328636-121328636 on Build GRCh38
rs185593423 Chr.4: 121329001-121329001 on Build GRCh38
rs531022706 Chr.4: 121328886-121328886 on Build GRCh38
rs188794945 Chr.4: 121328953-121328953 on Build GRCh38
rs554082636 Chr.4: 121329000-121329000 on Build GRCh38
rs929380038 Chr.4: 121328637-121328637 on Build GRCh38
rs551148490 Chr.4: 121328890-121328890 on Build GRCh38
rs866174166 Chr.4: 121328848-121328848 on Build GRCh38
rs867877072 Chr.4: 121328760-121328760 on Build GRCh38
rs577859262 Chr.4: 121328825-121328825 on Build GRCh38
rs182203774 Chr.4: 121328957-121328957 on Build GRCh38
rs752859490 Chr.4: 121328756-121328756 on Build GRCh38
rs796190189 Chr.4: 121328568-121328568 on Build GRCh38
rs1016272580 Chr.4: 121329014-121329014 on Build GRCh38
rs984612512 Chr.4: 121328641-121328641 on Build GRCh38
rs566145007 Chr.4: 121328943-121328943 on Build GRCh38
rs547301906 Chr.4: 121328841-121328841 on Build GRCh38
rs965925572 Chr.4: 121329006-121329006 on Build GRCh38
rs917743152 Chr.4: 121329032-121329032 on Build GRCh38
rs546188718 Chr.4: 121328632-121328632 on Build GRCh38
rs138344431 Chr.4: 121328805-121328805 on Build GRCh38
rs934574185 Chr.4: 121328851-121328851 on Build GRCh38
rs944532595 Chr.4: 121328704-121328704 on Build GRCh38
rs575693079 Chr.4: 121329044-121329044 on Build GRCh38
rs902292749 Chr.4: 121328796-121328796 on Build GRCh38
rs558278709 Chr.4: 121328970-121328970 on Build GRCh38
rs997770678 Chr.4: 121328979-121328979 on Build GRCh38
rs879168796 Chr.4: 121328580-121328580 on Build GRCh38
rs143218015 Chr.4: 121328905-121328905 on Build GRCh38
rs1042483084 Chr.4: 121328928-121328928 on Build GRCh38
rs764101530 Chr.4: 121328776-121328776 on Build GRCh38
rs1041181093 Chr.4: 121328758-121328758 on Build GRCh38
rs138655937 Chr.4: 121329043-121329043 on Build GRCh38
rs893273198 Chr.4: 121328813-121328813 on Build GRCh38
rs372537877 Chr.4: 121328989-121328989 on Build GRCh38
rs571485228 Chr.4: 121328996-121328996 on Build GRCh38
rs944548397 Chr.4: 121328867-121328867 on Build GRCh38
rs6821123 Chr.4: 121328818-121328818 on Build GRCh38
rs765503930 Chr.4: 121328738-121328738 on Build GRCh38
rs974581663 Chr.4: 121329025-121329025 on Build GRCh38
rs148328506 Chr.4: 121328937-121328937 on Build GRCh38
rs936412010 Chr.4: 121328948-121328948 on Build GRCh38
rs1007816139 Chr.4: 121328817-121328817 on Build GRCh38
rs759872770 Chr.4: 121328631-121328631 on Build GRCh38
rs550996087 Chr.4: 121328569-121328569 on Build GRCh38
rs990771589 Chr.4: 121328904-121328904 on Build GRCh38
rs1004431070 Chr.4: 121328775-121328775 on Build GRCh38
rs991081573 Chr.4: 121329028-121329028 on Build GRCh38
rs11935582 Chr.4: 121328764-121328764 on Build GRCh38
rs115107652 Chr.4: 121328777-121328777 on Build GRCh38
rs1033157465 Chr.4: 121328582-121328582 on Build GRCh38

View Full Product Details